Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43305659-43306162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIP3
Entrez Gene ID
401262
Gene Name
cysteine rich protein 3
Gene Aliases
TLP, TLP-A, bA480N24.2
UniGene
Hs.653165
Gene Chromosome Location
Chr.6: 43305473-43310705 on Build GRCh38
Transcripts
NM_206922.2 XM_005249103.3 XM_011514609.2 XM_017010856.1 XM_017010857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6547277 c.537C>A p.Y179* Chr.6: 43306083-43306083 on Build GRCh38
7335788 c.562_563CC>TT p.P188F Chr.6: 43306057-43306058 on Build GRCh38
3686346 c.554-4_554-3delTC p.? Chr.6: 43305878-43305879 on Build GRCh38
4802403 c.539T>C p.L180P Chr.6: 43306081-43306081 on Build GRCh38
7189806 c.496-13_496-10delCTCT p.? Chr.6: 43306134-43306137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767743121 Chr.6: 43306041-43306041 on Build GRCh38
rs778669001 Chr.6: 43305854-43305854 on Build GRCh38
rs377417533 Chr.6: 43305811-43305811 on Build GRCh38
rs754990737 Chr.6: 43306059-43306059 on Build GRCh38
rs771902071 Chr.6: 43305863-43305863 on Build GRCh38
rs758075887 Chr.6: 43305950-43305950 on Build GRCh38
rs1018148108 Chr.6: 43306067-43306067 on Build GRCh38
rs777408235 Chr.6: 43306100-43306100 on Build GRCh38
rs760394473 Chr.6: 43305873-43305873 on Build GRCh38
rs760826223 Chr.6: 43305755-43305755 on Build GRCh38
rs970604415 Chr.6: 43305727-43305727 on Build GRCh38
rs756963871 Chr.6: 43306113-43306113 on Build GRCh38
rs1004761361 Chr.6: 43306088-43306088 on Build GRCh38
rs764519667 Chr.6: 43305810-43305810 on Build GRCh38
rs747962560 Chr.6: 43305861-43305861 on Build GRCh38
rs780089544 Chr.6: 43305925-43305925 on Build GRCh38
rs895215195 Chr.6: 43306028-43306028 on Build GRCh38
rs763708321 Chr.6: 43305878-43305879 on Build GRCh38
rs540905070 Chr.6: 43305908-43305908 on Build GRCh38
rs776320119 Chr.6: 43305884-43305884 on Build GRCh38
rs142092139 Chr.6: 43306093-43306093 on Build GRCh38
rs752684319 Chr.6: 43306135-43306135 on Build GRCh38
rs753206379 Chr.6: 43305798-43305798 on Build GRCh38
rs756050567 Chr.6: 43306003-43306004 on Build GRCh38
rs767947806 Chr.6: 43305906-43305906 on Build GRCh38
rs202173523 Chr.6: 43305882-43305882 on Build GRCh38
rs541222968 Chr.6: 43305953-43305953 on Build GRCh38
rs953096780 Chr.6: 43306131-43306131 on Build GRCh38
rs200169587 Chr.6: 43306045-43306045 on Build GRCh38
rs780923110 Chr.6: 43306128-43306128 on Build GRCh38
rs556539517 Chr.6: 43305766-43305766 on Build GRCh38
rs372092478 Chr.6: 43306023-43306023 on Build GRCh38
rs918173515 Chr.6: 43305893-43305893 on Build GRCh38
rs371132803 Chr.6: 43305910-43305910 on Build GRCh38
rs780874089 Chr.6: 43305947-43305948 on Build GRCh38
rs769444881 Chr.6: 43306011-43306011 on Build GRCh38
rs150965756 Chr.6: 43306089-43306089 on Build GRCh38
rs756372630 Chr.6: 43305921-43305921 on Build GRCh38
rs770755699 Chr.6: 43305960-43305960 on Build GRCh38
rs777651019 Chr.6: 43305952-43305952 on Build GRCh38
rs745781004 Chr.6: 43305948-43305948 on Build GRCh38
rs994213974 Chr.6: 43305702-43305702 on Build GRCh38
rs374665833 Chr.6: 43306064-43306064 on Build GRCh38
rs370836390 Chr.6: 43305791-43305791 on Build GRCh38
rs1018115148 Chr.6: 43305760-43305760 on Build GRCh38
rs778752973 Chr.6: 43305948-43305948 on Build GRCh38
rs562856023 Chr.6: 43306050-43306050 on Build GRCh38
rs762413810 Chr.6: 43306037-43306037 on Build GRCh38
rs774491969 Chr.6: 43306134-43306137 on Build GRCh38
rs753979239 Chr.6: 43305931-43305931 on Build GRCh38
rs2242416 Chr.6: 43305866-43305866 on Build GRCh38
rs371312241 Chr.6: 43306095-43306095 on Build GRCh38
rs370256460 Chr.6: 43305827-43305827 on Build GRCh38
rs1053774567 Chr.6: 43305954-43305954 on Build GRCh38
rs370582150 Chr.6: 43306048-43306048 on Build GRCh38
rs201423675 Chr.6: 43305775-43305775 on Build GRCh38
rs374667415 Chr.6: 43305886-43305886 on Build GRCh38
rs944295933 Chr.6: 43305868-43305868 on Build GRCh38
rs745436154 Chr.6: 43305993-43305993 on Build GRCh38
rs750581496 Chr.6: 43305698-43305698 on Build GRCh38
rs139840752 Chr.6: 43306055-43306055 on Build GRCh38
rs768633761 Chr.6: 43305846-43305846 on Build GRCh38
rs370374796 Chr.6: 43305875-43305875 on Build GRCh38
rs746611330 Chr.6: 43305958-43305958 on Build GRCh38
rs748127034 Chr.6: 43305949-43305949 on Build GRCh38
rs983806161 Chr.6: 43305747-43305747 on Build GRCh38
rs780142848 Chr.6: 43305842-43305842 on Build GRCh38
rs746878328 Chr.6: 43306107-43306107 on Build GRCh38
rs536286394 Chr.6: 43305754-43305754 on Build GRCh38
rs980939942 Chr.6: 43305923-43305923 on Build GRCh38
rs867455206 Chr.6: 43305985-43305985 on Build GRCh38
rs763581229 Chr.6: 43305800-43305800 on Build GRCh38
rs758061806 Chr.6: 43305851-43305852 on Build GRCh38
rs903709473 Chr.6: 43306101-43306101 on Build GRCh38
rs1029593035 Chr.6: 43305726-43305726 on Build GRCh38
rs1024789557 Chr.6: 43305697-43305697 on Build GRCh38
rs368241763 Chr.6: 43306046-43306046 on Build GRCh38
rs762028136 Chr.6: 43305904-43305904 on Build GRCh38
rs201397271 Chr.6: 43305891-43305891 on Build GRCh38
rs754908036 Chr.6: 43305935-43305935 on Build GRCh38
rs774673052 Chr.6: 43306018-43306018 on Build GRCh38
rs551756582 Chr.6: 43306133-43306133 on Build GRCh38
rs781207961 Chr.6: 43305813-43305813 on Build GRCh38
rs756757403 Chr.6: 43305916-43305916 on Build GRCh38
rs560991651 Chr.6: 43305966-43305966 on Build GRCh38
rs750674418 Chr.6: 43305816-43305816 on Build GRCh38
rs929665317 Chr.6: 43305977-43305977 on Build GRCh38
rs543114859 Chr.6: 43306051-43306051 on Build GRCh38
rs948304317 Chr.6: 43305902-43305902 on Build GRCh38
rs1006728651 Chr.6: 43305759-43305759 on Build GRCh38
rs1051159689 Chr.6: 43306005-43306005 on Build GRCh38
rs779898788 Chr.6: 43306004-43306025 on Build GRCh38
rs749095914 Chr.6: 43306023-43306024 on Build GRCh38
rs751268471 Chr.6: 43305877-43305878 on Build GRCh38

View Full Product Details