Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43307520-43308009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIP3
Entrez Gene ID
401262
Gene Name
cysteine rich protein 3
Gene Aliases
TLP, TLP-A, bA480N24.2
UniGene
Hs.653165
Gene Chromosome Location
Chr.6: 43305473-43310705 on Build GRCh38
Transcripts
NM_206922.2 XM_005249103.3 XM_011514609.2 XM_017010856.1 XM_017010857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6174568 c.39-1G>T p.? Chr.6: 43307744-43307744 on Build GRCh38
7362681 c.244C>T p.P82S Chr.6: 43307696-43307696 on Build GRCh38
4860823 c.261C>T p.T87T Chr.6: 43307679-43307679 on Build GRCh38
7309152 c.196+1delG p.? Chr.6: 43307838-43307838 on Build GRCh38
6335152 c.182_183insT p.F62fs*74 Chr.6: 43307852-43307853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773829961 Chr.6: 43307862-43307862 on Build GRCh38
rs771406786 Chr.6: 43307577-43307579 on Build GRCh38
rs762623605 Chr.6: 43307672-43307672 on Build GRCh38
rs774898490 Chr.6: 43307563-43307563 on Build GRCh38
rs981945068 Chr.6: 43307553-43307553 on Build GRCh38
rs374123253 Chr.6: 43307815-43307815 on Build GRCh38
rs771047248 Chr.6: 43307710-43307710 on Build GRCh38
rs972108937 Chr.6: 43307782-43307782 on Build GRCh38
rs1009142862 Chr.6: 43307614-43307614 on Build GRCh38
rs140362028 Chr.6: 43307869-43307869 on Build GRCh38
rs944602379 Chr.6: 43307596-43307596 on Build GRCh38
rs76547503 Chr.6: 43307628-43307628 on Build GRCh38
rs541337690 Chr.6: 43307976-43307976 on Build GRCh38
rs779822326 Chr.6: 43307840-43307840 on Build GRCh38
rs924567580 Chr.6: 43307887-43307887 on Build GRCh38
rs765672147 Chr.6: 43307777-43307777 on Build GRCh38
rs760104744 Chr.6: 43307892-43307892 on Build GRCh38
rs767080671 Chr.6: 43307773-43307773 on Build GRCh38
rs973223931 Chr.6: 43307896-43307896 on Build GRCh38
rs763574850 Chr.6: 43307674-43307674 on Build GRCh38
rs770827989 Chr.6: 43307633-43307633 on Build GRCh38
rs767108396 Chr.6: 43307842-43307842 on Build GRCh38
rs769640838 Chr.6: 43307721-43307721 on Build GRCh38
rs764540933 Chr.6: 43307933-43307933 on Build GRCh38
rs986252775 Chr.6: 43307832-43307832 on Build GRCh38
rs765906933 Chr.6: 43307914-43307914 on Build GRCh38
rs376815023 Chr.6: 43307625-43307625 on Build GRCh38
rs752932709 Chr.6: 43307701-43307701 on Build GRCh38
rs750025283 Chr.6: 43307857-43307857 on Build GRCh38
rs199501421 Chr.6: 43307741-43307741 on Build GRCh38
rs749682698 Chr.6: 43307605-43307605 on Build GRCh38
rs761559938 Chr.6: 43307864-43307864 on Build GRCh38
rs775072627 Chr.6: 43307665-43307665 on Build GRCh38
rs759757459 Chr.6: 43307778-43307784 on Build GRCh38
rs1017810683 Chr.6: 43307623-43307623 on Build GRCh38
rs377179764 Chr.6: 43307827-43307827 on Build GRCh38
rs568254475 Chr.6: 43307622-43307622 on Build GRCh38
rs769537251 Chr.6: 43307735-43307735 on Build GRCh38
rs138077538 Chr.6: 43307707-43307707 on Build GRCh38
rs751090342 Chr.6: 43307678-43307678 on Build GRCh38
rs750913287 Chr.6: 43307597-43307597 on Build GRCh38
rs776141581 Chr.6: 43307930-43307930 on Build GRCh38
rs781362757 Chr.6: 43307808-43307808 on Build GRCh38
rs771453914 Chr.6: 43307873-43307873 on Build GRCh38
rs765636137 Chr.6: 43307832-43307833 on Build GRCh38
rs757235928 Chr.6: 43307796-43307796 on Build GRCh38
rs926516414 Chr.6: 43307857-43307857 on Build GRCh38
rs768510046 Chr.6: 43307859-43307859 on Build GRCh38
rs761609893 Chr.6: 43307837-43307838 on Build GRCh38
rs201516256 Chr.6: 43307781-43307781 on Build GRCh38
rs756192716 Chr.6: 43307944-43307944 on Build GRCh38
rs370402239 Chr.6: 43307705-43307705 on Build GRCh38
rs910346181 Chr.6: 43307833-43307833 on Build GRCh38
rs866124822 Chr.6: 43307787-43307787 on Build GRCh38
rs576850505 Chr.6: 43307700-43307700 on Build GRCh38
rs545458904 Chr.6: 43307740-43307740 on Build GRCh38
rs930737787 Chr.6: 43307550-43307550 on Build GRCh38
rs761240730 Chr.6: 43307685-43307685 on Build GRCh38
rs747742993 Chr.6: 43307565-43307565 on Build GRCh38
rs763169199 Chr.6: 43307932-43307932 on Build GRCh38
rs772894256 Chr.6: 43307882-43307882 on Build GRCh38
rs751689823 Chr.6: 43307936-43307936 on Build GRCh38
rs1020611499 Chr.6: 43307813-43307813 on Build GRCh38
rs144504876 Chr.6: 43307554-43307554 on Build GRCh38
rs764936237 Chr.6: 43307568-43307568 on Build GRCh38
rs777787546 Chr.6: 43307795-43307795 on Build GRCh38
rs779910262 Chr.6: 43307948-43307948 on Build GRCh38
rs750568290 Chr.6: 43307942-43307942 on Build GRCh38
rs892871519 Chr.6: 43307699-43307699 on Build GRCh38
rs757538519 Chr.6: 43307937-43307937 on Build GRCh38
rs538248259 Chr.6: 43307987-43307988 on Build GRCh38
rs756631381 Chr.6: 43307598-43307598 on Build GRCh38
rs777891175 Chr.6: 43307706-43307706 on Build GRCh38
rs777037026 Chr.6: 43307676-43307677 on Build GRCh38
rs768260799 Chr.6: 43307742-43307742 on Build GRCh38
rs761173880 Chr.6: 43307756-43307756 on Build GRCh38
rs758685639 Chr.6: 43307784-43307784 on Build GRCh38
rs757843861 Chr.6: 43307574-43307574 on Build GRCh38
rs536850980 Chr.6: 43307646-43307646 on Build GRCh38
rs755402534 Chr.6: 43307607-43307607 on Build GRCh38
rs774058253 Chr.6: 43307744-43307744 on Build GRCh38
rs557047417 Chr.6: 43307649-43307649 on Build GRCh38
rs949301568 Chr.6: 43307540-43307540 on Build GRCh38
rs749016624 Chr.6: 43307850-43307850 on Build GRCh38
rs780933627 Chr.6: 43307716-43307716 on Build GRCh38
rs374377263 Chr.6: 43307613-43307613 on Build GRCh38
rs371142557 Chr.6: 43307686-43307686 on Build GRCh38
rs962325208 Chr.6: 43307778-43307778 on Build GRCh38
rs754262519 Chr.6: 43307776-43307776 on Build GRCh38
rs763615442 Chr.6: 43307586-43307586 on Build GRCh38
rs751213599 Chr.6: 43307833-43307834 on Build GRCh38
rs148395811 Chr.6: 43307600-43307600 on Build GRCh38
rs998542868 Chr.6: 43307640-43307640 on Build GRCh38
rs759259605 Chr.6: 43307651-43307651 on Build GRCh38
rs765029410 Chr.6: 43307661-43307661 on Build GRCh38

View Full Product Details