Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 55399090-55399599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFRAL
Entrez Gene ID
389400
Gene Name
GDNF family receptor alpha like
Gene Aliases
C6orf144, GRAL, UNQ9356, bA360D14.1
UniGene
Hs.526967
Gene Chromosome Location
Chr.6: 55327469-55402493 on Build GRCh38
Transcripts
NM_207410.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
228444 c.1033C>T p.H345Y Chr.6: 55399260-55399260 on Build GRCh38
3830806 c.1087T>C p.C363R Chr.6: 55399407-55399407 on Build GRCh38
225240 c.1091G>A p.G364E Chr.6: 55399411-55399411 on Build GRCh38
3874868 c.1029A>T p.G343G Chr.6: 55399256-55399256 on Build GRCh38
3158182 c.1049G>A p.G350E Chr.6: 55399369-55399369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200921457 Chr.6: 55399249-55399249 on Build GRCh38
rs887610355 Chr.6: 55399554-55399554 on Build GRCh38
rs775012001 Chr.6: 55399356-55399356 on Build GRCh38
rs763958116 Chr.6: 55399255-55399255 on Build GRCh38
rs1010021499 Chr.6: 55399122-55399122 on Build GRCh38
rs371425482 Chr.6: 55399332-55399332 on Build GRCh38
rs201980240 Chr.6: 55399540-55399540 on Build GRCh38
rs374135154 Chr.6: 55399195-55399195 on Build GRCh38
rs901571648 Chr.6: 55399533-55399533 on Build GRCh38
rs368127361 Chr.6: 55399339-55399339 on Build GRCh38
rs992540787 Chr.6: 55399386-55399386 on Build GRCh38
rs925623012 Chr.6: 55399436-55399436 on Build GRCh38
rs901285199 Chr.6: 55399545-55399545 on Build GRCh38
rs199881300 Chr.6: 55399298-55399298 on Build GRCh38
rs746131173 Chr.6: 55399189-55399189 on Build GRCh38
rs769159751 Chr.6: 55399291-55399291 on Build GRCh38
rs368319488 Chr.6: 55399458-55399458 on Build GRCh38
rs958843110 Chr.6: 55399338-55399338 on Build GRCh38
rs759366088 Chr.6: 55399447-55399448 on Build GRCh38
rs6936510 Chr.6: 55399564-55399564 on Build GRCh38
rs1028745674 Chr.6: 55399552-55399552 on Build GRCh38
rs1015245466 Chr.6: 55399568-55399568 on Build GRCh38
rs747091669 Chr.6: 55399278-55399278 on Build GRCh38
rs891488432 Chr.6: 55399441-55399441 on Build GRCh38
rs948221832 Chr.6: 55399357-55399357 on Build GRCh38
rs748644642 Chr.6: 55399462-55399462 on Build GRCh38
rs766054229 Chr.6: 55399137-55399137 on Build GRCh38
rs757320637 Chr.6: 55399411-55399411 on Build GRCh38
rs980907727 Chr.6: 55399405-55399405 on Build GRCh38
rs762189285 Chr.6: 55399220-55399220 on Build GRCh38
rs770832351 Chr.6: 55399272-55399272 on Build GRCh38
rs879043079 Chr.6: 55399140-55399140 on Build GRCh38
rs774973224 Chr.6: 55399486-55399486 on Build GRCh38
rs935930435 Chr.6: 55399164-55399164 on Build GRCh38
rs758608419 Chr.6: 55399435-55399435 on Build GRCh38
rs770142998 Chr.6: 55399316-55399316 on Build GRCh38
rs746075168 Chr.6: 55399268-55399268 on Build GRCh38
rs62419126 Chr.6: 55399252-55399252 on Build GRCh38
rs41271314 Chr.6: 55399123-55399123 on Build GRCh38
rs201803352 Chr.6: 55399197-55399197 on Build GRCh38
rs753873571 Chr.6: 55399260-55399260 on Build GRCh38
rs758708163 Chr.6: 55399272-55399272 on Build GRCh38
rs769048075 Chr.6: 55399213-55399213 on Build GRCh38
rs756015631 Chr.6: 55399160-55399160 on Build GRCh38
rs781318787 Chr.6: 55399461-55399461 on Build GRCh38
rs773965514 Chr.6: 55399227-55399227 on Build GRCh38
rs891452622 Chr.6: 55399117-55399117 on Build GRCh38
rs760078856 Chr.6: 55399399-55399399 on Build GRCh38
rs910404867 Chr.6: 55399511-55399511 on Build GRCh38
rs776984587 Chr.6: 55399297-55399297 on Build GRCh38
rs760441499 Chr.6: 55399493-55399493 on Build GRCh38
rs754465875 Chr.6: 55399146-55399146 on Build GRCh38
rs780164083 Chr.6: 55399437-55399437 on Build GRCh38
rs770302603 Chr.6: 55399464-55399464 on Build GRCh38
rs992285190 Chr.6: 55399336-55399336 on Build GRCh38
rs771722530 Chr.6: 55399195-55399196 on Build GRCh38
rs776608554 Chr.6: 55399360-55399360 on Build GRCh38
rs777016216 Chr.6: 55399216-55399216 on Build GRCh38
rs758604772 Chr.6: 55399524-55399524 on Build GRCh38
rs527905870 Chr.6: 55399238-55399239 on Build GRCh38
rs746108316 Chr.6: 55399513-55399513 on Build GRCh38
rs1014968521 Chr.6: 55399309-55399309 on Build GRCh38
rs779253139 Chr.6: 55399183-55399183 on Build GRCh38
rs372028331 Chr.6: 55399478-55399478 on Build GRCh38
rs373918623 Chr.6: 55399334-55399334 on Build GRCh38
rs751024841 Chr.6: 55399144-55399144 on Build GRCh38
rs747307748 Chr.6: 55399453-55399453 on Build GRCh38
rs917976388 Chr.6: 55399349-55399349 on Build GRCh38
rs749066157 Chr.6: 55399167-55399167 on Build GRCh38
rs765641139 Chr.6: 55399221-55399221 on Build GRCh38
rs752680001 Chr.6: 55399158-55399158 on Build GRCh38
rs1041125911 Chr.6: 55399230-55399230 on Build GRCh38
rs145926895 Chr.6: 55399429-55399429 on Build GRCh38
rs759255450 Chr.6: 55399232-55399232 on Build GRCh38
rs1023975981 Chr.6: 55399390-55399390 on Build GRCh38
rs757085738 Chr.6: 55399264-55399264 on Build GRCh38
rs137898068 Chr.6: 55399225-55399225 on Build GRCh38
rs560037887 Chr.6: 55399415-55399415 on Build GRCh38
rs780116452 Chr.6: 55399274-55399274 on Build GRCh38
rs977531154 Chr.6: 55399430-55399430 on Build GRCh38
rs544648274 Chr.6: 55399165-55399165 on Build GRCh38
rs142349780 Chr.6: 55399409-55399409 on Build GRCh38
rs759118506 Chr.6: 55399337-55399337 on Build GRCh38
rs545487951 Chr.6: 55399471-55399471 on Build GRCh38
rs778917843 Chr.6: 55399267-55399267 on Build GRCh38
rs760228908 Chr.6: 55399256-55399256 on Build GRCh38
rs146209371 Chr.6: 55399190-55399190 on Build GRCh38
rs780724271 Chr.6: 55399147-55399147 on Build GRCh38
rs767045551 Chr.6: 55399241-55399241 on Build GRCh38
rs901174989 Chr.6: 55399250-55399250 on Build GRCh38

View Full Product Details