Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154332213-154332712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B2
Entrez Gene ID
57198
Gene Name
ATPase phospholipid transporting 8B2
Gene Aliases
ATPID
UniGene
Hs.435700
Gene Chromosome Location
Chr.1: 154325541-154351307 on Build GRCh38
Transcripts
NM_001005855.1 NM_020452.3 XM_005245355.2 XM_005245356.3 XM_017001871.1 XM_017001872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5453299 c.588C>T p.D196D Chr.1: 154332696-154332696 on Build GRCh38
5453300 c.645C>T p.D215D Chr.1: 154332696-154332696 on Build GRCh38
5159892 c.646A>G p.T216A Chr.1: 154332655-154332655 on Build GRCh38
7147405 c.688G>A p.G230S Chr.1: 154332697-154332697 on Build GRCh38
6345266 c.661G>T p.D221Y Chr.1: 154332670-154332670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948940476 Chr.1: 154332353-154332353 on Build GRCh38
rs907179295 Chr.1: 154332383-154332383 on Build GRCh38
rs756497833 Chr.1: 154332648-154332648 on Build GRCh38
rs981632475 Chr.1: 154332638-154332638 on Build GRCh38
rs765867648 Chr.1: 154332594-154332594 on Build GRCh38
rs371131370 Chr.1: 154332666-154332666 on Build GRCh38
rs945947984 Chr.1: 154332604-154332604 on Build GRCh38
rs78350897 Chr.1: 154332592-154332592 on Build GRCh38
rs990317788 Chr.1: 154332333-154332333 on Build GRCh38
rs977519876 Chr.1: 154332573-154332573 on Build GRCh38
rs977174971 Chr.1: 154332300-154332300 on Build GRCh38
rs948909566 Chr.1: 154332609-154332609 on Build GRCh38
rs1209264 Chr.1: 154332534-154332534 on Build GRCh38
rs571624192 Chr.1: 154332471-154332471 on Build GRCh38
rs141051422 Chr.1: 154332645-154332645 on Build GRCh38
rs572550130 Chr.1: 154332590-154332590 on Build GRCh38
rs114989837 Chr.1: 154332414-154332414 on Build GRCh38
rs781069879 Chr.1: 154332669-154332669 on Build GRCh38
rs756174163 Chr.1: 154332598-154332599 on Build GRCh38
rs965712365 Chr.1: 154332559-154332560 on Build GRCh38
rs151081718 Chr.1: 154332541-154332541 on Build GRCh38
rs745904972 Chr.1: 154332665-154332665 on Build GRCh38
rs957420256 Chr.1: 154332557-154332557 on Build GRCh38
rs375088244 Chr.1: 154332490-154332490 on Build GRCh38
rs576546996 Chr.1: 154332682-154332682 on Build GRCh38
rs951633445 Chr.1: 154332465-154332465 on Build GRCh38
rs768000646 Chr.1: 154332637-154332637 on Build GRCh38
rs566909866 Chr.1: 154332302-154332302 on Build GRCh38
rs1045740983 Chr.1: 154332248-154332248 on Build GRCh38
rs778919135 Chr.1: 154332664-154332664 on Build GRCh38
rs538301210 Chr.1: 154332516-154332516 on Build GRCh38
rs542935459 Chr.1: 154332281-154332281 on Build GRCh38
rs916157805 Chr.1: 154332340-154332340 on Build GRCh38
rs751065189 Chr.1: 154332605-154332605 on Build GRCh38
rs1001587701 Chr.1: 154332310-154332310 on Build GRCh38
rs759970744 Chr.1: 154332626-154332626 on Build GRCh38
rs554034699 Chr.1: 154332574-154332574 on Build GRCh38
rs1055920798 Chr.1: 154332356-154332356 on Build GRCh38
rs55723479 Chr.1: 154332457-154332457 on Build GRCh38
rs186009420 Chr.1: 154332455-154332455 on Build GRCh38
rs1007443378 Chr.1: 154332544-154332544 on Build GRCh38
rs574954079 Chr.1: 154332536-154332536 on Build GRCh38
rs754154734 Chr.1: 154332653-154332653 on Build GRCh38
rs777137387 Chr.1: 154332315-154332315 on Build GRCh38
rs937363337 Chr.1: 154332656-154332656 on Build GRCh38
rs772807167 Chr.1: 154332591-154332591 on Build GRCh38
rs940031408 Chr.1: 154332387-154332387 on Build GRCh38
rs995711324 Chr.1: 154332456-154332456 on Build GRCh38
rs534178471 Chr.1: 154332396-154332396 on Build GRCh38
rs965691017 Chr.1: 154332528-154332528 on Build GRCh38
rs778259766 Chr.1: 154332652-154332652 on Build GRCh38
rs757497332 Chr.1: 154332658-154332658 on Build GRCh38
rs936165723 Chr.1: 154332326-154332326 on Build GRCh38
rs1026300641 Chr.1: 154332464-154332464 on Build GRCh38
rs1018891279 Chr.1: 154332545-154332545 on Build GRCh38
rs745314468 Chr.1: 154332517-154332517 on Build GRCh38
rs1045687634 Chr.1: 154332618-154332618 on Build GRCh38

View Full Product Details