Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155237976-155238471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA
Entrez Gene ID
2629
Gene Name
glucosylceramidase beta
Gene Aliases
GBA1, GCB, GLUC
UniGene
Hs.282997
Gene Chromosome Location
Chr.1: 155234448-155244862 on Build GRCh38
Transcripts
NM_000157.3 NM_001005741.2 NM_001005742.2 NM_001171811.1 NM_001171812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422315 c.661C>T p.P221S Chr.1: 155238234-155238234 on Build GRCh38
896825 c.688G>A p.V230M Chr.1: 155238207-155238207 on Build GRCh38
4404256 c.674A>C p.K225T Chr.1: 155238221-155238221 on Build GRCh38
4503027 c.629C>T p.P210L Chr.1: 155238266-155238266 on Build GRCh38
3474896 c.649C>T p.P217S Chr.1: 155238246-155238246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183540501 Chr.1: 155238057-155238057 on Build GRCh38
rs772098596 Chr.1: 155238155-155238155 on Build GRCh38
rs188760929 Chr.1: 155238264-155238264 on Build GRCh38
rs398123534 Chr.1: 155238174-155238174 on Build GRCh38
rs76717906 Chr.1: 155238244-155238244 on Build GRCh38
rs364897 Chr.1: 155238215-155238215 on Build GRCh38
rs529870563 Chr.1: 155238075-155238075 on Build GRCh38
rs145066479 Chr.1: 155238055-155238055 on Build GRCh38
rs544083269 Chr.1: 155238238-155238238 on Build GRCh38
rs78659905 Chr.1: 155238256-155238256 on Build GRCh38
rs77916306 Chr.1: 155238301-155238301 on Build GRCh38
rs566671462 Chr.1: 155238376-155238376 on Build GRCh38
rs76727497 Chr.1: 155238232-155238232 on Build GRCh38
rs1009850780 Chr.1: 155238291-155238291 on Build GRCh38
rs1034162688 Chr.1: 155238434-155238434 on Build GRCh38
rs381427 Chr.1: 155238206-155238206 on Build GRCh38
rs527333751 Chr.1: 155238375-155238375 on Build GRCh38
rs867929413 Chr.1: 155238242-155238242 on Build GRCh38
rs372785813 Chr.1: 155238187-155238187 on Build GRCh38
rs772419342 Chr.1: 155238305-155238305 on Build GRCh38
rs77933015 Chr.1: 155238296-155238296 on Build GRCh38
rs398123532 Chr.1: 155238270-155238270 on Build GRCh38
rs77451368 Chr.1: 155238173-155238173 on Build GRCh38
rs398123531 Chr.1: 155238290-155238290 on Build GRCh38
rs76026102 Chr.1: 155238164-155238164 on Build GRCh38
rs369678230 Chr.1: 155238087-155238087 on Build GRCh38
rs79945741 Chr.1: 155238139-155238139 on Build GRCh38
rs550305278 Chr.1: 155238039-155238039 on Build GRCh38
rs1064644 Chr.1: 155238192-155238192 on Build GRCh38
rs1001787113 Chr.1: 155238428-155238428 on Build GRCh38
rs1009903585 Chr.1: 155238031-155238031 on Build GRCh38
rs747992229 Chr.1: 155238086-155238086 on Build GRCh38
rs74486098 Chr.1: 155238184-155238184 on Build GRCh38
rs774609294 Chr.1: 155238272-155238272 on Build GRCh38
rs866075757 Chr.1: 155238234-155238234 on Build GRCh38
rs774539868 Chr.1: 155238193-155238193 on Build GRCh38
rs753425476 Chr.1: 155238097-155238097 on Build GRCh38
rs539805773 Chr.1: 155238088-155238088 on Build GRCh38
rs756942598 Chr.1: 155238122-155238122 on Build GRCh38
rs375731497 Chr.1: 155238199-155238199 on Build GRCh38
rs374591570 Chr.1: 155238258-155238258 on Build GRCh38
rs556008401 Chr.1: 155238259-155238259 on Build GRCh38
rs80205046 Chr.1: 155238233-155238233 on Build GRCh38
rs2974923 Chr.1: 155238392-155238392 on Build GRCh38
rs786200979 Chr.1: 155238214-155238215 on Build GRCh38
rs749416070 Chr.1: 155238269-155238269 on Build GRCh38
rs381737 Chr.1: 155238141-155238141 on Build GRCh38
rs535592331 Chr.1: 155238052-155238052 on Build GRCh38
rs75370695 Chr.1: 155238208-155238208 on Build GRCh38
rs74462743 Chr.1: 155238194-155238194 on Build GRCh38
rs61748906 Chr.1: 155238228-155238228 on Build GRCh38
rs76500263 Chr.1: 155238293-155238293 on Build GRCh38
rs75636769 Chr.1: 155238209-155238209 on Build GRCh38
rs121908300 Chr.1: 155238144-155238144 on Build GRCh38
rs78911246 Chr.1: 155238212-155238212 on Build GRCh38
rs409652 Chr.1: 155238174-155238174 on Build GRCh38
rs76682322 Chr.1: 155238223-155238223 on Build GRCh38
rs777319607 Chr.1: 155238094-155238094 on Build GRCh38
rs201615998 Chr.1: 155238265-155238265 on Build GRCh38
rs904267933 Chr.1: 155238390-155238390 on Build GRCh38
rs764609479 Chr.1: 155238352-155238352 on Build GRCh38
rs560175843 Chr.1: 155238344-155238344 on Build GRCh38
rs373631650 Chr.1: 155238102-155238102 on Build GRCh38
rs376613535 Chr.1: 155238175-155238175 on Build GRCh38
rs76158190 Chr.1: 155238190-155238190 on Build GRCh38
rs381418 Chr.1: 155238214-155238214 on Build GRCh38
rs767914182 Chr.1: 155238202-155238202 on Build GRCh38
rs398123533 Chr.1: 155238261-155238261 on Build GRCh38
rs1051872176 Chr.1: 155238218-155238218 on Build GRCh38
rs80222298 Chr.1: 155238302-155238302 on Build GRCh38
rs773409311 Chr.1: 155238186-155238186 on Build GRCh38
rs558529804 Chr.1: 155238092-155238092 on Build GRCh38
rs749714463 Chr.1: 155238299-155238300 on Build GRCh38
rs1034708011 Chr.1: 155238061-155238061 on Build GRCh38

View Full Product Details