Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155239572-155240066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA
Entrez Gene ID
2629
Gene Name
glucosylceramidase beta
Gene Aliases
GBA1, GCB, GLUC
UniGene
Hs.282997
Gene Chromosome Location
Chr.1: 155234448-155244862 on Build GRCh38
Transcripts
NM_000157.3 NM_001005741.2 NM_001005742.2 NM_001171811.1 NM_001171812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6359123 c.226T>C p.F76L Chr.1: 155239967-155239967 on Build GRCh38
3360370 c.308G>C p.G103A Chr.1: 155239762-155239762 on Build GRCh38
3802096 c.158T>G p.V53G Chr.1: 155240035-155240035 on Build GRCh38
4962341 c.145G>A p.G49S Chr.1: 155240048-155240048 on Build GRCh38
6669659 c.144C>T p.F48F Chr.1: 155240049-155240049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145888253 Chr.1: 155240008-155240008 on Build GRCh38
rs776232497 Chr.1: 155239814-155239823 on Build GRCh38
rs878853314 Chr.1: 155239655-155239655 on Build GRCh38
rs1141814 Chr.1: 155239934-155239934 on Build GRCh38
rs121908312 Chr.1: 155239716-155239716 on Build GRCh38
rs368786234 Chr.1: 155239962-155239962 on Build GRCh38
rs749867889 Chr.1: 155239775-155239775 on Build GRCh38
rs79175920 Chr.1: 155239683-155239683 on Build GRCh38
rs745398454 Chr.1: 155239910-155239910 on Build GRCh38
rs188289486 Chr.1: 155239837-155239837 on Build GRCh38
rs757834551 Chr.1: 155239987-155239987 on Build GRCh38
rs752857428 Chr.1: 155239960-155239960 on Build GRCh38
rs78669556 Chr.1: 155239932-155239932 on Build GRCh38
rs878853316 Chr.1: 155239663-155239663 on Build GRCh38
rs753816998 Chr.1: 155239699-155239699 on Build GRCh38
rs758447515 Chr.1: 155239633-155239633 on Build GRCh38
rs916865472 Chr.1: 155239735-155239735 on Build GRCh38
rs878853318 Chr.1: 155240026-155240026 on Build GRCh38
rs747971975 Chr.1: 155240020-155240020 on Build GRCh38
rs757848523 Chr.1: 155239894-155239894 on Build GRCh38
rs372156961 Chr.1: 155239791-155239791 on Build GRCh38
rs781563451 Chr.1: 155239758-155239758 on Build GRCh38
rs779258874 Chr.1: 155239970-155239970 on Build GRCh38
rs121908302 Chr.1: 155240033-155240033 on Build GRCh38
rs757409399 Chr.1: 155239848-155239848 on Build GRCh38
rs377315750 Chr.1: 155239803-155239803 on Build GRCh38
rs75954905 Chr.1: 155239965-155239965 on Build GRCh38
rs773007510 Chr.1: 155240029-155240029 on Build GRCh38
rs779619231 Chr.1: 155239740-155239740 on Build GRCh38
rs769900428 Chr.1: 155239974-155239974 on Build GRCh38
rs770218957 Chr.1: 155239616-155239616 on Build GRCh38
rs760358958 Chr.1: 155239838-155239838 on Build GRCh38
rs897478056 Chr.1: 155239863-155239863 on Build GRCh38
rs770561256 Chr.1: 155239788-155239788 on Build GRCh38
rs371592589 Chr.1: 155239972-155239972 on Build GRCh38
rs761806383 Chr.1: 155239853-155239853 on Build GRCh38
rs748485792 Chr.1: 155239762-155239762 on Build GRCh38
rs780169533 Chr.1: 155239746-155239746 on Build GRCh38
rs775652188 Chr.1: 155240045-155240045 on Build GRCh38
rs746684178 Chr.1: 155239778-155239778 on Build GRCh38
rs369792423 Chr.1: 155239858-155239858 on Build GRCh38
rs1141820 Chr.1: 155239897-155239897 on Build GRCh38
rs141061530 Chr.1: 155239990-155239990 on Build GRCh38
rs761621516 Chr.1: 155239969-155239971 on Build GRCh38
rs539166948 Chr.1: 155239615-155239615 on Build GRCh38
rs111857613 Chr.1: 155239773-155239773 on Build GRCh38
rs755161012 Chr.1: 155239752-155239752 on Build GRCh38
rs375471032 Chr.1: 155239854-155239854 on Build GRCh38
rs373363286 Chr.1: 155239969-155239969 on Build GRCh38
rs1141818 Chr.1: 155239898-155239898 on Build GRCh38
rs763972468 Chr.1: 155239687-155239687 on Build GRCh38
rs74953658 Chr.1: 155240004-155240004 on Build GRCh38
rs756264143 Chr.1: 155240037-155240037 on Build GRCh38
rs77829017 Chr.1: 155239939-155239939 on Build GRCh38
rs76337315 Chr.1: 155239953-155239953 on Build GRCh38
rs756540913 Chr.1: 155239767-155239767 on Build GRCh38
rs368145008 Chr.1: 155239748-155239748 on Build GRCh38
rs151093421 Chr.1: 155239977-155239977 on Build GRCh38
rs1141815 Chr.1: 155239918-155239918 on Build GRCh38
rs758455177 Chr.1: 155239700-155239700 on Build GRCh38
rs753011212 Chr.1: 155239872-155239872 on Build GRCh38
rs767272691 Chr.1: 155239658-155239658 on Build GRCh38
rs794727708 Chr.1: 155239639-155239639 on Build GRCh38
rs767693527 Chr.1: 155239956-155239956 on Build GRCh38
rs746737219 Chr.1: 155239929-155239929 on Build GRCh38
rs770614424 Chr.1: 155239643-155239643 on Build GRCh38
rs767999479 Chr.1: 155239789-155239790 on Build GRCh38
rs752444566 Chr.1: 155239680-155239680 on Build GRCh38
rs750712570 Chr.1: 155239989-155239989 on Build GRCh38
rs1141821 Chr.1: 155239893-155239893 on Build GRCh38
rs1050455670 Chr.1: 155239914-155239914 on Build GRCh38
rs763010874 Chr.1: 155240031-155240031 on Build GRCh38
rs1038197220 Chr.1: 155239862-155239862 on Build GRCh38
rs1141810 Chr.1: 155239950-155239950 on Build GRCh38
rs146774384 Chr.1: 155239961-155239961 on Build GRCh38
rs765182795 Chr.1: 155239945-155239945 on Build GRCh38
rs750990483 Chr.1: 155239978-155239978 on Build GRCh38
rs759174705 Chr.1: 155239657-155239657 on Build GRCh38
rs1033293531 Chr.1: 155239609-155239609 on Build GRCh38
rs771643561 Chr.1: 155239806-155239806 on Build GRCh38
rs144173415 Chr.1: 155239936-155239936 on Build GRCh38
rs774985960 Chr.1: 155239926-155239926 on Build GRCh38
rs778349378 Chr.1: 155239890-155239890 on Build GRCh38
rs867024654 Chr.1: 155239709-155239709 on Build GRCh38
rs145773486 Chr.1: 155240025-155240025 on Build GRCh38
rs78769774 Chr.1: 155239933-155239933 on Build GRCh38
rs771654706 Chr.1: 155239923-155239923 on Build GRCh38
rs763930060 Chr.1: 155239843-155239843 on Build GRCh38
rs868211463 Chr.1: 155239705-155239705 on Build GRCh38
rs374003673 Chr.1: 155239648-155239648 on Build GRCh38
rs1141811 Chr.1: 155239948-155239948 on Build GRCh38
rs781476674 Chr.1: 155239598-155239598 on Build GRCh38
rs1141816 Chr.1: 155239914-155239914 on Build GRCh38
rs772806479 Chr.1: 155239623-155239623 on Build GRCh38
rs1141812 Chr.1: 155239946-155239946 on Build GRCh38
rs756420893 Chr.1: 155239875-155239875 on Build GRCh38

View Full Product Details