Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155240162-155240706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA
Entrez Gene ID
2629
Gene Name
glucosylceramidase beta
Gene Aliases
GBA1, GCB, GLUC
UniGene
Hs.282997
Gene Chromosome Location
Chr.1: 155234448-155244862 on Build GRCh38
Transcripts
NM_000157.3 NM_001005741.2 NM_001005742.2 NM_001171811.1 NM_001171812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669662 c.109G>A p.A37T Chr.1: 155240636-155240636 on Build GRCh38
896827 c.88C>A p.L30I Chr.1: 155240657-155240657 on Build GRCh38
6121482 c.90A>T p.L30L Chr.1: 155240655-155240655 on Build GRCh38
6121481 c.75C>G p.L25L Chr.1: 155240670-155240670 on Build GRCh38
1747794 c.45G>C p.L15F Chr.1: 155240700-155240700 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs546949267 Chr.1: 155240321-155240321 on Build GRCh38
rs777383151 Chr.1: 155240637-155240637 on Build GRCh38
rs763323321 Chr.1: 155240600-155240600 on Build GRCh38
rs541342886 Chr.1: 155240554-155240554 on Build GRCh38
rs1059731 Chr.1: 155240664-155240664 on Build GRCh38
rs1028070045 Chr.1: 155240429-155240429 on Build GRCh38
rs1041670981 Chr.1: 155240248-155240248 on Build GRCh38
rs903292670 Chr.1: 155240256-155240256 on Build GRCh38
rs771771655 Chr.1: 155240276-155240276 on Build GRCh38
rs573523478 Chr.1: 155240483-155240483 on Build GRCh38
rs748587897 Chr.1: 155240229-155240229 on Build GRCh38
rs907441251 Chr.1: 155240369-155240369 on Build GRCh38
rs912491062 Chr.1: 155240465-155240465 on Build GRCh38
rs377526831 Chr.1: 155240588-155240588 on Build GRCh38
rs776856496 Chr.1: 155240647-155240647 on Build GRCh38
rs1054989688 Chr.1: 155240525-155240525 on Build GRCh38
rs201330214 Chr.1: 155240663-155240663 on Build GRCh38
rs945269864 Chr.1: 155240466-155240466 on Build GRCh38
rs779522491 Chr.1: 155240215-155240215 on Build GRCh38
rs772779630 Chr.1: 155240292-155240292 on Build GRCh38
rs1013084985 Chr.1: 155240291-155240291 on Build GRCh38
rs146532106 Chr.1: 155240394-155240394 on Build GRCh38
rs142348200 Chr.1: 155240336-155240336 on Build GRCh38
rs867447625 Chr.1: 155240245-155240245 on Build GRCh38
rs766943717 Chr.1: 155240610-155240610 on Build GRCh38
rs748322959 Chr.1: 155240658-155240658 on Build GRCh38
rs745860677 Chr.1: 155240644-155240644 on Build GRCh38
rs912461257 Chr.1: 155240202-155240202 on Build GRCh38
rs557469991 Chr.1: 155240371-155240371 on Build GRCh38
rs752104007 Chr.1: 155240615-155240615 on Build GRCh38
rs954035160 Chr.1: 155240439-155240439 on Build GRCh38
rs755265447 Chr.1: 155240184-155240184 on Build GRCh38
rs1000200049 Chr.1: 155240258-155240258 on Build GRCh38
rs755800314 Chr.1: 155240634-155240634 on Build GRCh38
rs104886460 Chr.1: 155240629-155240629 on Build GRCh38
rs936010622 Chr.1: 155240519-155240519 on Build GRCh38
rs894671364 Chr.1: 155240283-155240283 on Build GRCh38
rs894543050 Chr.1: 155240544-155240544 on Build GRCh38
rs767077683 Chr.1: 155240619-155240619 on Build GRCh38
rs397518433 Chr.1: 155240673-155240673 on Build GRCh38
rs1003895526 Chr.1: 155240378-155240378 on Build GRCh38
rs768854161 Chr.1: 155240645-155240645 on Build GRCh38
rs559366562 Chr.1: 155240575-155240575 on Build GRCh38
rs986636304 Chr.1: 155240455-155240456 on Build GRCh38
rs182016990 Chr.1: 155240471-155240471 on Build GRCh38
rs757041827 Chr.1: 155240641-155240641 on Build GRCh38
rs962460364 Chr.1: 155240379-155240379 on Build GRCh38
rs139190127 Chr.1: 155240461-155240461 on Build GRCh38
rs80356760 Chr.1: 155240651-155240652 on Build GRCh38
rs372948636 Chr.1: 155240186-155240186 on Build GRCh38
rs369199473 Chr.1: 155240596-155240596 on Build GRCh38
rs549359970 Chr.1: 155240188-155240188 on Build GRCh38
rs948929646 Chr.1: 155240558-155240558 on Build GRCh38
rs571534886 Chr.1: 155240324-155240324 on Build GRCh38
rs759337640 Chr.1: 155240388-155240388 on Build GRCh38
rs924582879 Chr.1: 155240477-155240477 on Build GRCh38
rs387906315 Chr.1: 155240660-155240661 on Build GRCh38
rs148001886 Chr.1: 155240640-155240640 on Build GRCh38
rs151028758 Chr.1: 155240239-155240239 on Build GRCh38
rs567511270 Chr.1: 155240196-155240196 on Build GRCh38

View Full Product Details