Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136259030-136259536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCLAF1
Entrez Gene ID
9774
Gene Name
BCL2 associated transcription factor 1
Gene Aliases
BTF, bK211L9.1
UniGene
Hs.486542
Gene Chromosome Location
Chr.6: 136256863-136290645 on Build GRCh38
Transcripts
NM_001077440.1 NM_001077441.1 NM_001301038.1 NM_014739.2 XM_005267237.3 XM_017011529.1 XM_017011530.1 XM_017011531.1 XM_017011532.1

SNP Detail

SNP ID Chromosome Location
rs145325273 Chr.6: 136259110-136259110 on Build GRCh38
rs764642141 Chr.6: 136259460-136259460 on Build GRCh38
rs953744225 Chr.6: 136259404-136259404 on Build GRCh38
rs532058609 Chr.6: 136259472-136259472 on Build GRCh38
rs1024240305 Chr.6: 136259301-136259301 on Build GRCh38
rs573341219 Chr.6: 136259159-136259159 on Build GRCh38
rs894507602 Chr.6: 136259286-136259286 on Build GRCh38
rs920960235 Chr.6: 136259166-136259166 on Build GRCh38
rs574103614 Chr.6: 136259088-136259088 on Build GRCh38
rs932331705 Chr.6: 136259172-136259172 on Build GRCh38
rs892330681 Chr.6: 136259061-136259061 on Build GRCh38
rs531268648 Chr.6: 136259216-136259216 on Build GRCh38
rs558017385 Chr.6: 136259150-136259150 on Build GRCh38
rs565314548 Chr.6: 136259209-136259209 on Build GRCh38
rs1009423029 Chr.6: 136259088-136259088 on Build GRCh38
rs1007190719 Chr.6: 136259164-136259164 on Build GRCh38
rs995007493 Chr.6: 136259236-136259236 on Build GRCh38
rs924402677 Chr.6: 136259137-136259137 on Build GRCh38
rs944890043 Chr.6: 136259225-136259225 on Build GRCh38
rs957217123 Chr.6: 136259377-136259377 on Build GRCh38
rs944922461 Chr.6: 136259430-136259430 on Build GRCh38
rs1050658352 Chr.6: 136259206-136259206 on Build GRCh38
rs907127117 Chr.6: 136259498-136259498 on Build GRCh38
rs573169513 Chr.6: 136259227-136259227 on Build GRCh38
rs769067391 Chr.6: 136259233-136259233 on Build GRCh38
rs990343746 Chr.6: 136259390-136259390 on Build GRCh38
rs542996905 Chr.6: 136259091-136259091 on Build GRCh38
rs957308297 Chr.6: 136259151-136259152 on Build GRCh38
rs1017688755 Chr.6: 136259184-136259184 on Build GRCh38
rs951548475 Chr.6: 136259302-136259302 on Build GRCh38
rs375765429 Chr.6: 136259334-136259335 on Build GRCh38
rs977222773 Chr.6: 136259120-136259120 on Build GRCh38
rs761005274 Chr.6: 136259283-136259283 on Build GRCh38
rs986402232 Chr.6: 136259417-136259418 on Build GRCh38
rs983029438 Chr.6: 136259311-136259311 on Build GRCh38
rs971247095 Chr.6: 136259064-136259064 on Build GRCh38
rs529642546 Chr.6: 136259341-136259341 on Build GRCh38
rs181501628 Chr.6: 136259350-136259350 on Build GRCh38
rs187066970 Chr.6: 136259285-136259285 on Build GRCh38
rs147257641 Chr.6: 136259162-136259162 on Build GRCh38
rs776830532 Chr.6: 136259451-136259451 on Build GRCh38
rs1036497231 Chr.6: 136259437-136259437 on Build GRCh38
rs574510374 Chr.6: 136259420-136259420 on Build GRCh38
rs1036467760 Chr.6: 136259232-136259233 on Build GRCh38
rs995296739 Chr.6: 136259282-136259282 on Build GRCh38
rs191880824 Chr.6: 136259322-136259322 on Build GRCh38
rs775864102 Chr.6: 136259269-136259269 on Build GRCh38
rs540843600 Chr.6: 136259475-136259475 on Build GRCh38
rs866505605 Chr.6: 136259243-136259243 on Build GRCh38
rs574079541 Chr.6: 136259084-136259084 on Build GRCh38
rs867458503 Chr.6: 136259450-136259450 on Build GRCh38
rs1031885501 Chr.6: 136259357-136259357 on Build GRCh38
rs1045723740 Chr.6: 136259479-136259479 on Build GRCh38
rs552383502 Chr.6: 136259493-136259493 on Build GRCh38
rs980773881 Chr.6: 136259394-136259394 on Build GRCh38
rs1012908831 Chr.6: 136259300-136259300 on Build GRCh38
rs989837487 Chr.6: 136259156-136259156 on Build GRCh38
rs1049636096 Chr.6: 136259455-136259455 on Build GRCh38

View Full Product Details