Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 168059004-168059496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD1
Entrez Gene ID
79981
Gene Name
FERM domain containing 1
Gene Aliases
bA164L23.1
UniGene
Hs.266746
Gene Chromosome Location
Chr.6: 168055745-168093007 on Build GRCh38
Transcripts
NM_001122841.2 NM_024919.4 NR_110312.1 XM_011536137.1 XM_011536138.1 XM_011536140.1 XM_011536141.1 XM_011536142.2 XM_011536143.1 XM_011536145.1 XM_011536146.2 XM_017011317.1 XM_017011318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3662287 c.1348C>T p.R450C Chr.6: 168059183-168059183 on Build GRCh38
5806010 c.1389C>A p.S463R Chr.6: 168059142-168059142 on Build GRCh38
1442268 c.1349G>A p.R450H Chr.6: 168059182-168059182 on Build GRCh38
5734385 c.1366C>T p.Q456* Chr.6: 168059165-168059165 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200379574 Chr.6: 168059196-168059196 on Build GRCh38
rs963061221 Chr.6: 168059254-168059254 on Build GRCh38
rs561606153 Chr.6: 168059473-168059473 on Build GRCh38
rs142200860 Chr.6: 168059290-168059290 on Build GRCh38
rs761047750 Chr.6: 168059100-168059100 on Build GRCh38
rs932050953 Chr.6: 168059391-168059391 on Build GRCh38
rs766046528 Chr.6: 168059141-168059141 on Build GRCh38
rs922060205 Chr.6: 168059380-168059380 on Build GRCh38
rs947398581 Chr.6: 168059371-168059371 on Build GRCh38
rs1052063279 Chr.6: 168059392-168059392 on Build GRCh38
rs905087009 Chr.6: 168059289-168059289 on Build GRCh38
rs200893521 Chr.6: 168059108-168059108 on Build GRCh38
rs542036816 Chr.6: 168059312-168059312 on Build GRCh38
rs1051445550 Chr.6: 168059094-168059094 on Build GRCh38
rs1015227819 Chr.6: 168059253-168059253 on Build GRCh38
rs1043031417 Chr.6: 168059375-168059375 on Build GRCh38
rs773090242 Chr.6: 168059174-168059174 on Build GRCh38
rs980699061 Chr.6: 168059328-168059328 on Build GRCh38
rs771248336 Chr.6: 168059104-168059104 on Build GRCh38
rs1023285438 Chr.6: 168059433-168059433 on Build GRCh38
rs759276821 Chr.6: 168059192-168059192 on Build GRCh38
rs772067246 Chr.6: 168059173-168059173 on Build GRCh38
rs145347617 Chr.6: 168059132-168059132 on Build GRCh38
rs574950112 Chr.6: 168059285-168059285 on Build GRCh38
rs546135901 Chr.6: 168059402-168059402 on Build GRCh38
rs540295406 Chr.6: 168059444-168059444 on Build GRCh38
rs994542686 Chr.6: 168059426-168059426 on Build GRCh38
rs753435160 Chr.6: 168059142-168059142 on Build GRCh38
rs376385156 Chr.6: 168059182-168059182 on Build GRCh38
rs113303915 Chr.6: 168059109-168059110 on Build GRCh38
rs563691458 Chr.6: 168059403-168059403 on Build GRCh38
rs575523123 Chr.6: 168059356-168059356 on Build GRCh38
rs767322463 Chr.6: 168059138-168059138 on Build GRCh38
rs372913124 Chr.6: 168059110-168059112 on Build GRCh38
rs890309368 Chr.6: 168059400-168059400 on Build GRCh38
rs975576440 Chr.6: 168059374-168059374 on Build GRCh38
rs781577220 Chr.6: 168059161-168059161 on Build GRCh38
rs550778293 Chr.6: 168059207-168059207 on Build GRCh38
rs1052601269 Chr.6: 168059442-168059442 on Build GRCh38
rs757823860 Chr.6: 168059217-168059217 on Build GRCh38
rs748185252 Chr.6: 168059087-168059087 on Build GRCh38
rs753129706 Chr.6: 168059200-168059200 on Build GRCh38
rs900161708 Chr.6: 168059049-168059049 on Build GRCh38
rs370946458 Chr.6: 168059111-168059111 on Build GRCh38
rs765935791 Chr.6: 168059187-168059187 on Build GRCh38
rs913446767 Chr.6: 168059370-168059370 on Build GRCh38
rs767111189 Chr.6: 168059107-168059107 on Build GRCh38
rs758994513 Chr.6: 168059149-168059149 on Build GRCh38
rs371020738 Chr.6: 168059233-168059233 on Build GRCh38
rs772136885 Chr.6: 168059089-168059089 on Build GRCh38
rs1548349 Chr.6: 168059165-168059165 on Build GRCh38
rs969154075 Chr.6: 168059443-168059443 on Build GRCh38
rs147612406 Chr.6: 168059234-168059234 on Build GRCh38
rs1465182 Chr.6: 168059110-168059110 on Build GRCh38
rs75143837 Chr.6: 168059404-168059404 on Build GRCh38
rs931754768 Chr.6: 168059059-168059059 on Build GRCh38
rs778378097 Chr.6: 168059155-168059155 on Build GRCh38
rs530790376 Chr.6: 168059027-168059027 on Build GRCh38
rs557168636 Chr.6: 168059355-168059355 on Build GRCh38
rs955589038 Chr.6: 168059399-168059399 on Build GRCh38
rs749619950 Chr.6: 168059075-168059075 on Build GRCh38
rs535078988 Chr.6: 168059273-168059273 on Build GRCh38
rs928804022 Chr.6: 168059330-168059330 on Build GRCh38
rs144660790 Chr.6: 168059190-168059191 on Build GRCh38
rs911652180 Chr.6: 168059228-168059229 on Build GRCh38
rs12203150 Chr.6: 168059409-168059409 on Build GRCh38
rs189465441 Chr.6: 168059044-168059044 on Build GRCh38
rs758650150 Chr.6: 168059206-168059206 on Build GRCh38
rs747870184 Chr.6: 168059172-168059172 on Build GRCh38
rs768440241 Chr.6: 168059129-168059129 on Build GRCh38
rs780894379 Chr.6: 168059076-168059076 on Build GRCh38
rs375451629 Chr.6: 168059106-168059106 on Build GRCh38
rs570859046 Chr.6: 168059262-168059262 on Build GRCh38
rs374502267 Chr.6: 168059139-168059139 on Build GRCh38
rs781489100 Chr.6: 168059219-168059219 on Build GRCh38
rs774241490 Chr.6: 168059166-168059166 on Build GRCh38
rs776954452 Chr.6: 168059121-168059121 on Build GRCh38
rs749098533 Chr.6: 168059122-168059122 on Build GRCh38
rs368907480 Chr.6: 168059114-168059114 on Build GRCh38
rs750437477 Chr.6: 168059221-168059221 on Build GRCh38
rs574947269 Chr.6: 168059042-168059042 on Build GRCh38
rs779424084 Chr.6: 168059085-168059085 on Build GRCh38
rs2269680 Chr.6: 168059112-168059112 on Build GRCh38
rs761874710 Chr.6: 168059133-168059133 on Build GRCh38
rs569155779 Chr.6: 168059231-168059231 on Build GRCh38
rs747969036 Chr.6: 168059222-168059222 on Build GRCh38
rs552660720 Chr.6: 168059145-168059145 on Build GRCh38
rs553209573 Chr.6: 168059279-168059279 on Build GRCh38
rs762788427 Chr.6: 168059105-168059105 on Build GRCh38
rs773154730 Chr.6: 168059095-168059095 on Build GRCh38
rs951476805 Chr.6: 168059320-168059320 on Build GRCh38
rs757340818 Chr.6: 168059157-168059157 on Build GRCh38
rs776119446 Chr.6: 168059191-168059191 on Build GRCh38
rs760124489 Chr.6: 168059140-168059140 on Build GRCh38
rs754218892 Chr.6: 168059109-168059109 on Build GRCh38

View Full Product Details