Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161548205-161548713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGR3A
Entrez Gene ID
2214
Gene Name
Fc fragment of IgG receptor IIIa
Gene Aliases
CD16, CD16A, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20
UniGene
Hs.372679
Gene Chromosome Location
Chr.1: 161541759-161550623 on Build GRCh38
Transcripts
NM_001127593.1 NM_001127595.1 NM_001127596.1 XM_011509293.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4532731 c.190G>C p.V64L Chr.1: 161548658-161548658 on Build GRCh38
676635 c.187G>A p.V63M Chr.1: 161548661-161548661 on Build GRCh38
676637 c.226A>T p.K76* Chr.1: 161548622-161548622 on Build GRCh38
3477292 c.253G>A p.G85R Chr.1: 161548595-161548595 on Build GRCh38
4594001 c.305T>G p.L102R Chr.1: 161548543-161548543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757822838 Chr.1: 161548373-161548373 on Build GRCh38
rs10127939 Chr.1: 161548543-161548543 on Build GRCh38
rs772478882 Chr.1: 161548599-161548599 on Build GRCh38
rs777018978 Chr.1: 161548646-161548646 on Build GRCh38
rs771778975 Chr.1: 161548462-161548462 on Build GRCh38
rs762177505 Chr.1: 161548426-161548426 on Build GRCh38
rs565792189 Chr.1: 161548625-161548625 on Build GRCh38
rs760031681 Chr.1: 161548549-161548549 on Build GRCh38
rs550971926 Chr.1: 161548662-161548662 on Build GRCh38
rs745497558 Chr.1: 161548460-161548460 on Build GRCh38
rs199840551 Chr.1: 161548494-161548494 on Build GRCh38
rs760235231 Chr.1: 161548471-161548471 on Build GRCh38
rs772451890 Chr.1: 161548552-161548552 on Build GRCh38
rs142959418 Chr.1: 161548628-161548628 on Build GRCh38
rs769796708 Chr.1: 161548401-161548401 on Build GRCh38
rs757234780 Chr.1: 161548501-161548501 on Build GRCh38
rs374159614 Chr.1: 161548438-161548438 on Build GRCh38
rs372866264 Chr.1: 161548681-161548681 on Build GRCh38
rs148181339 Chr.1: 161548424-161548424 on Build GRCh38
rs765603242 Chr.1: 161548473-161548473 on Build GRCh38
rs748001163 Chr.1: 161548400-161548400 on Build GRCh38
rs10127857 Chr.1: 161548647-161548647 on Build GRCh38
rs200225944 Chr.1: 161548369-161548369 on Build GRCh38
rs758196872 Chr.1: 161548539-161548539 on Build GRCh38
rs747025029 Chr.1: 161548389-161548389 on Build GRCh38
rs771557708 Chr.1: 161548508-161548508 on Build GRCh38
rs762943796 Chr.1: 161548661-161548661 on Build GRCh38
rs77144485 Chr.1: 161548546-161548546 on Build GRCh38
rs770709280 Chr.1: 161548457-161548457 on Build GRCh38
rs534629496 Chr.1: 161548252-161548252 on Build GRCh38
rs370021323 Chr.1: 161548387-161548387 on Build GRCh38
rs17853190 Chr.1: 161548497-161548497 on Build GRCh38
rs751824906 Chr.1: 161548440-161548440 on Build GRCh38
rs772846548 Chr.1: 161548402-161548402 on Build GRCh38
rs747338915 Chr.1: 161548602-161548602 on Build GRCh38
rs143657896 Chr.1: 161548619-161548619 on Build GRCh38
rs752370305 Chr.1: 161548629-161548629 on Build GRCh38
rs770430447 Chr.1: 161548563-161548567 on Build GRCh38
rs775115768 Chr.1: 161548425-161548425 on Build GRCh38
rs755119939 Chr.1: 161548489-161548489 on Build GRCh38
rs368021338 Chr.1: 161548532-161548532 on Build GRCh38
rs150808747 Chr.1: 161548509-161548509 on Build GRCh38
rs776818304 Chr.1: 161548609-161548609 on Build GRCh38
rs765356562 Chr.1: 161548567-161548567 on Build GRCh38
rs145700363 Chr.1: 161548592-161548592 on Build GRCh38
rs753847696 Chr.1: 161548564-161548564 on Build GRCh38
rs781395883 Chr.1: 161548399-161548399 on Build GRCh38
rs749283416 Chr.1: 161548447-161548447 on Build GRCh38
rs755288067 Chr.1: 161548442-161548442 on Build GRCh38
rs143870625 Chr.1: 161548525-161548525 on Build GRCh38
rs921169747 Chr.1: 161548250-161548250 on Build GRCh38
rs752014792 Chr.1: 161548478-161548478 on Build GRCh38
rs746365321 Chr.1: 161548597-161548597 on Build GRCh38
rs775885096 Chr.1: 161548600-161548600 on Build GRCh38
rs755092182 Chr.1: 161548390-161548390 on Build GRCh38
rs763528543 Chr.1: 161548513-161548513 on Build GRCh38
rs766171155 Chr.1: 161548575-161548575 on Build GRCh38
rs763618100 Chr.1: 161548476-161548476 on Build GRCh38
rs760848344 Chr.1: 161548528-161548528 on Build GRCh38
rs764331513 Chr.1: 161548561-161548561 on Build GRCh38
rs561419661 Chr.1: 161548615-161548615 on Build GRCh38
rs534373347 Chr.1: 161548659-161548659 on Build GRCh38
rs754633167 Chr.1: 161548586-161548586 on Build GRCh38
rs536649865 Chr.1: 161548690-161548690 on Build GRCh38
rs747670473 Chr.1: 161548545-161548545 on Build GRCh38
rs114535887 Chr.1: 161548524-161548524 on Build GRCh38
rs781008829 Chr.1: 161548588-161548588 on Build GRCh38
rs750126518 Chr.1: 161548537-161548537 on Build GRCh38
rs749089205 Chr.1: 161548407-161548407 on Build GRCh38
rs754297382 Chr.1: 161548372-161548372 on Build GRCh38
rs750685224 Chr.1: 161548428-161548428 on Build GRCh38
rs551336717 Chr.1: 161548548-161548548 on Build GRCh38
rs758652985 Chr.1: 161548385-161548385 on Build GRCh38
rs370926518 Chr.1: 161548540-161548540 on Build GRCh38
rs139622799 Chr.1: 161548443-161548443 on Build GRCh38
rs773814856 Chr.1: 161548577-161548577 on Build GRCh38
rs771583484 Chr.1: 161548516-161548516 on Build GRCh38
rs557303793 Chr.1: 161548422-161548422 on Build GRCh38
rs773981918 Chr.1: 161548421-161548421 on Build GRCh38
rs747872317 Chr.1: 161548589-161548589 on Build GRCh38
rs774155063 Chr.1: 161548664-161548664 on Build GRCh38
rs751329449 Chr.1: 161548577-161548577 on Build GRCh38
rs573948357 Chr.1: 161548265-161548265 on Build GRCh38
rs147185130 Chr.1: 161548620-161548620 on Build GRCh38
rs759589336 Chr.1: 161548671-161548671 on Build GRCh38
rs769001959 Chr.1: 161548644-161548644 on Build GRCh38
rs752665263 Chr.1: 161548493-161548493 on Build GRCh38
rs4656315 Chr.1: 161548382-161548382 on Build GRCh38
rs767401028 Chr.1: 161548675-161548675 on Build GRCh38
rs780796380 Chr.1: 161548544-161548544 on Build GRCh38
rs780437274 Chr.1: 161548640-161548640 on Build GRCh38
rs753330143 Chr.1: 161548691-161548691 on Build GRCh38
rs758304652 Chr.1: 161548573-161548573 on Build GRCh38
rs766816229 Chr.1: 161548477-161548477 on Build GRCh38
rs570953466 Chr.1: 161548687-161548687 on Build GRCh38
rs774794148 Chr.1: 161548522-161548522 on Build GRCh38
rs767787896 Chr.1: 161548490-161548490 on Build GRCh38
rs774822098 Chr.1: 161548468-161548468 on Build GRCh38
rs748181680 Chr.1: 161548500-161548500 on Build GRCh38
rs765266636 Chr.1: 161548618-161548618 on Build GRCh38
rs773839536 Chr.1: 161548475-161548475 on Build GRCh38
rs145420690 Chr.1: 161548445-161548445 on Build GRCh38
rs750369174 Chr.1: 161548571-161548571 on Build GRCh38
rs779125158 Chr.1: 161548375-161548375 on Build GRCh38
rs770515092 Chr.1: 161548414-161548414 on Build GRCh38
rs428888 Chr.1: 161548496-161548496 on Build GRCh38
rs747571784 Chr.1: 161548641-161548641 on Build GRCh38
rs145557772 Chr.1: 161548444-161548444 on Build GRCh38
rs752564507 Chr.1: 161548684-161548684 on Build GRCh38
rs142977184 Chr.1: 161548511-161548511 on Build GRCh38
rs764137201 Chr.1: 161548531-161548531 on Build GRCh38
rs770023781 Chr.1: 161548656-161548656 on Build GRCh38
rs142322800 Chr.1: 161548502-161548502 on Build GRCh38
rs758874296 Chr.1: 161548638-161548638 on Build GRCh38
rs447536 Chr.1: 161548626-161548626 on Build GRCh38
rs931972996 Chr.1: 161548268-161548268 on Build GRCh38
rs781481196 Chr.1: 161548456-161548457 on Build GRCh38
rs777467556 Chr.1: 161548595-161548595 on Build GRCh38
rs52820103 Chr.1: 161548507-161548507 on Build GRCh38
rs760902695 Chr.1: 161548556-161548556 on Build GRCh38
rs142368299 Chr.1: 161548434-161548434 on Build GRCh38
rs200727785 Chr.1: 161548496-161548496 on Build GRCh38
rs775769047 Chr.1: 161548555-161548555 on Build GRCh38

View Full Product Details