Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 23908263-23908762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf58
Entrez Gene ID
254158
Gene Name
chromosome X open reading frame 58
Gene Aliases
-
UniGene
Hs.351265
Gene Chromosome Location
Chr.X: 23907932-23939520 on Build GRCh38
Transcripts
NM_001169574.1 NM_152761.2 XM_011545473.2

SNP Detail

SNP ID Chromosome Location
rs914388540 Chr.X: 23908452-23908452 on Build GRCh38
rs576616204 Chr.X: 23908617-23908617 on Build GRCh38
rs955763257 Chr.X: 23908352-23908352 on Build GRCh38
rs911225840 Chr.X: 23908515-23908515 on Build GRCh38
rs947305761 Chr.X: 23908460-23908460 on Build GRCh38
rs931013436 Chr.X: 23908402-23908402 on Build GRCh38
rs1021558722 Chr.X: 23908664-23908664 on Build GRCh38
rs149309695 Chr.X: 23908535-23908535 on Build GRCh38
rs764338941 Chr.X: 23908338-23908338 on Build GRCh38
rs1012914535 Chr.X: 23908719-23908719 on Build GRCh38
rs1049049243 Chr.X: 23908659-23908659 on Build GRCh38
rs1010453776 Chr.X: 23908654-23908654 on Build GRCh38
rs376475677 Chr.X: 23908699-23908699 on Build GRCh38
rs930037963 Chr.X: 23908641-23908641 on Build GRCh38
rs1049823888 Chr.X: 23908477-23908480 on Build GRCh38
rs544331863 Chr.X: 23908677-23908677 on Build GRCh38
rs989076312 Chr.X: 23908425-23908425 on Build GRCh38
rs969672176 Chr.X: 23908292-23908292 on Build GRCh38
rs111929870 Chr.X: 23908531-23908531 on Build GRCh38
rs762216283 Chr.X: 23908524-23908524 on Build GRCh38
rs185296375 Chr.X: 23908702-23908702 on Build GRCh38
rs766519035 Chr.X: 23908344-23908344 on Build GRCh38
rs762993640 Chr.X: 23908302-23908302 on Build GRCh38
rs1037273484 Chr.X: 23908367-23908367 on Build GRCh38
rs755184203 Chr.X: 23908590-23908590 on Build GRCh38
rs898715756 Chr.X: 23908400-23908404 on Build GRCh38
rs192863887 Chr.X: 23908477-23908477 on Build GRCh38

View Full Product Details