Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47639209-47639711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK1
Entrez Gene ID
2002
Gene Name
ELK1, ETS transcription factor
Gene Aliases
-
UniGene
Hs.181128
Gene Chromosome Location
Chr.X: 47635520-47650604 on Build GRCh38
Transcripts
NM_001114123.2 NM_001257168.1 NM_005229.4 XM_017029339.1

SNP Detail

SNP ID Chromosome Location
rs1022543767 Chr.X: 47639366-47639366 on Build GRCh38
rs755017144 Chr.X: 47639343-47639343 on Build GRCh38
rs747137558 Chr.X: 47639328-47639328 on Build GRCh38
rs760124689 Chr.X: 47639471-47639471 on Build GRCh38
rs765036702 Chr.X: 47639355-47639355 on Build GRCh38
rs776958066 Chr.X: 47639353-47639353 on Build GRCh38
rs948087959 Chr.X: 47639677-47639677 on Build GRCh38
rs759146491 Chr.X: 47639342-47639343 on Build GRCh38
rs887882504 Chr.X: 47639630-47639630 on Build GRCh38
rs976022761 Chr.X: 47639557-47639557 on Build GRCh38
rs188557403 Chr.X: 47639548-47639548 on Build GRCh38
rs988964146 Chr.X: 47639479-47639479 on Build GRCh38
rs1023982461 Chr.X: 47639357-47639357 on Build GRCh38
rs755745130 Chr.X: 47639276-47639276 on Build GRCh38
rs1049131544 Chr.X: 47639626-47639626 on Build GRCh38
rs970172045 Chr.X: 47639394-47639394 on Build GRCh38
rs2235183 Chr.X: 47639416-47639416 on Build GRCh38
rs921959211 Chr.X: 47639564-47639564 on Build GRCh38
rs376731476 Chr.X: 47639261-47639261 on Build GRCh38
rs1019145661 Chr.X: 47639437-47639437 on Build GRCh38
rs1033568088 Chr.X: 47639418-47639418 on Build GRCh38
rs775183854 Chr.X: 47639363-47639363 on Build GRCh38
rs773124002 Chr.X: 47639294-47639294 on Build GRCh38
rs769998944 Chr.X: 47639370-47639370 on Build GRCh38
rs953049530 Chr.X: 47639403-47639403 on Build GRCh38
rs763088656 Chr.X: 47639306-47639306 on Build GRCh38
rs776338870 Chr.X: 47639339-47639340 on Build GRCh38
rs113165175 Chr.X: 47639673-47639673 on Build GRCh38
rs781289398 Chr.X: 47639346-47639346 on Build GRCh38
rs765458087 Chr.X: 47639524-47639524 on Build GRCh38
rs759095742 Chr.X: 47639255-47639255 on Build GRCh38
rs368756202 Chr.X: 47639250-47639250 on Build GRCh38
rs142284754 Chr.X: 47639315-47639315 on Build GRCh38
rs767319405 Chr.X: 47639260-47639260 on Build GRCh38
rs766352376 Chr.X: 47639230-47639230 on Build GRCh38
rs979893181 Chr.X: 47639404-47639404 on Build GRCh38
rs771050532 Chr.X: 47639318-47639318 on Build GRCh38
rs752325820 Chr.X: 47639275-47639275 on Build GRCh38
rs932029388 Chr.X: 47639620-47639620 on Build GRCh38
rs965830716 Chr.X: 47639464-47639464 on Build GRCh38

View Full Product Details