Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 183626922-183627433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPC5
Entrez Gene ID
10092
Gene Name
actin related protein 2/3 complex subunit 5
Gene Aliases
ARC16, dJ127C7.3, p16-Arc
UniGene
Hs.518609
Gene Chromosome Location
Chr.1: 183626193-183635941 on Build GRCh38
Transcripts
NM_001270439.1 NM_005717.3

SNP Detail

SNP ID Chromosome Location
rs1052257875 Chr.1: 183627036-183627036 on Build GRCh38
rs901278345 Chr.1: 183626948-183626948 on Build GRCh38
rs779015384 Chr.1: 183627156-183627156 on Build GRCh38
rs112750148 Chr.1: 183627231-183627231 on Build GRCh38
rs11755 Chr.1: 183627001-183627001 on Build GRCh38
rs962261729 Chr.1: 183627216-183627216 on Build GRCh38
rs186236579 Chr.1: 183627389-183627389 on Build GRCh38
rs959542106 Chr.1: 183627295-183627295 on Build GRCh38
rs947386886 Chr.1: 183626988-183626989 on Build GRCh38
rs372019571 Chr.1: 183627382-183627382 on Build GRCh38
rs997243094 Chr.1: 183627083-183627083 on Build GRCh38
rs556518063 Chr.1: 183627101-183627101 on Build GRCh38
rs1043639610 Chr.1: 183626989-183626989 on Build GRCh38
rs1035823002 Chr.1: 183627293-183627293 on Build GRCh38
rs915878726 Chr.1: 183626947-183626947 on Build GRCh38
rs891684552 Chr.1: 183627037-183627037 on Build GRCh38
rs765760695 Chr.1: 183626960-183626960 on Build GRCh38
rs972785342 Chr.1: 183627357-183627357 on Build GRCh38
rs547502762 Chr.1: 183627024-183627024 on Build GRCh38
rs34219130 Chr.1: 183627002-183627003 on Build GRCh38
rs766685508 Chr.1: 183627397-183627397 on Build GRCh38
rs971205680 Chr.1: 183627213-183627213 on Build GRCh38
rs1049623280 Chr.1: 183626970-183626970 on Build GRCh38
rs1009929696 Chr.1: 183627066-183627066 on Build GRCh38
rs1015913093 Chr.1: 183627203-183627203 on Build GRCh38
rs566037658 Chr.1: 183627033-183627033 on Build GRCh38
rs915998402 Chr.1: 183627360-183627360 on Build GRCh38
rs1024378528 Chr.1: 183627119-183627119 on Build GRCh38
rs866283698 Chr.1: 183627321-183627321 on Build GRCh38
rs552013019 Chr.1: 183627338-183627338 on Build GRCh38
rs889620932 Chr.1: 183626994-183626994 on Build GRCh38
rs940246244 Chr.1: 183627339-183627339 on Build GRCh38
rs1032936711 Chr.1: 183627026-183627026 on Build GRCh38
rs1005824830 Chr.1: 183627135-183627135 on Build GRCh38
rs984226410 Chr.1: 183627306-183627306 on Build GRCh38
rs953052200 Chr.1: 183627110-183627110 on Build GRCh38
rs934054238 Chr.1: 183626962-183626962 on Build GRCh38
rs569256514 Chr.1: 183627222-183627222 on Build GRCh38
rs750940893 Chr.1: 183627000-183627000 on Build GRCh38

View Full Product Details