Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203217848-203218353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIT1
Entrez Gene ID
1118
Gene Name
chitinase 1
Gene Aliases
CHI3, CHIT, CHITD
UniGene
Hs.201688
Gene Chromosome Location
Chr.1: 203216079-203229732 on Build GRCh38
Transcripts
NM_001256125.1 NM_001270509.1 NM_003465.2 NR_045784.1 NR_045785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3360579 c.1044G>C p.K348N Chr.1: 203217851-203217851 on Build GRCh38
1201055 c.1037A>G p.Y346C Chr.1: 203217858-203217858 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748129910 Chr.1: 203217912-203217912 on Build GRCh38
rs757044414 Chr.1: 203217898-203217898 on Build GRCh38
rs1011899120 Chr.1: 203218058-203218058 on Build GRCh38
rs375923575 Chr.1: 203217879-203217879 on Build GRCh38
rs748924397 Chr.1: 203218092-203218093 on Build GRCh38
rs544680955 Chr.1: 203218095-203218095 on Build GRCh38
rs947397804 Chr.1: 203218144-203218144 on Build GRCh38
rs765618204 Chr.1: 203218012-203218012 on Build GRCh38
rs752734289 Chr.1: 203217898-203217898 on Build GRCh38
rs199856053 Chr.1: 203218212-203218212 on Build GRCh38
rs929848177 Chr.1: 203218280-203218280 on Build GRCh38
rs142622254 Chr.1: 203218219-203218220 on Build GRCh38
rs777185350 Chr.1: 203217961-203217961 on Build GRCh38
rs547786138 Chr.1: 203218202-203218202 on Build GRCh38
rs568039684 Chr.1: 203218239-203218239 on Build GRCh38
rs542268412 Chr.1: 203217959-203217959 on Build GRCh38
rs1001685921 Chr.1: 203218025-203218025 on Build GRCh38
rs2297948 Chr.1: 203217958-203217958 on Build GRCh38
rs577366461 Chr.1: 203217885-203217885 on Build GRCh38
rs541338513 Chr.1: 203218040-203218040 on Build GRCh38
rs770505086 Chr.1: 203218099-203218099 on Build GRCh38
rs575661846 Chr.1: 203218082-203218082 on Build GRCh38
rs753266805 Chr.1: 203218016-203218016 on Build GRCh38
rs142759151 Chr.1: 203218213-203218214 on Build GRCh38
rs994298957 Chr.1: 203218264-203218264 on Build GRCh38
rs985439446 Chr.1: 203217892-203217892 on Build GRCh38
rs1035871477 Chr.1: 203218102-203218102 on Build GRCh38
rs751161832 Chr.1: 203218038-203218038 on Build GRCh38
rs756956552 Chr.1: 203218046-203218046 on Build GRCh38
rs532310389 Chr.1: 203218114-203218114 on Build GRCh38
rs755622573 Chr.1: 203217926-203217934 on Build GRCh38
rs757953527 Chr.1: 203217888-203217888 on Build GRCh38
rs764818130 Chr.1: 203217871-203217871 on Build GRCh38
rs965326200 Chr.1: 203218207-203218207 on Build GRCh38
rs1048195362 Chr.1: 203218288-203218288 on Build GRCh38
rs577271761 Chr.1: 203218002-203218002 on Build GRCh38
rs763729427 Chr.1: 203218037-203218037 on Build GRCh38
rs762310177 Chr.1: 203218030-203218030 on Build GRCh38
rs372623018 Chr.1: 203218123-203218123 on Build GRCh38
rs58776333 Chr.1: 203218234-203218235 on Build GRCh38
rs1021742238 Chr.1: 203218163-203218163 on Build GRCh38
rs780072786 Chr.1: 203217905-203217905 on Build GRCh38
rs200696379 Chr.1: 203217894-203217894 on Build GRCh38
rs772208489 Chr.1: 203217918-203217918 on Build GRCh38
rs757760895 Chr.1: 203218253-203218253 on Build GRCh38
rs777181130 Chr.1: 203217986-203217987 on Build GRCh38
rs1025321177 Chr.1: 203218284-203218284 on Build GRCh38
rs745787601 Chr.1: 203217899-203217899 on Build GRCh38
rs956227761 Chr.1: 203218142-203218142 on Build GRCh38
rs984414771 Chr.1: 203218092-203218092 on Build GRCh38
rs552085944 Chr.1: 203218159-203218159 on Build GRCh38
rs769763032 Chr.1: 203217903-203217903 on Build GRCh38
rs773238756 Chr.1: 203217923-203217923 on Build GRCh38
rs767863960 Chr.1: 203218313-203218313 on Build GRCh38
rs968745998 Chr.1: 203217882-203217882 on Build GRCh38
rs977038860 Chr.1: 203218217-203218217 on Build GRCh38
rs755592950 Chr.1: 203217950-203217950 on Build GRCh38
rs41304073 Chr.1: 203217897-203217897 on Build GRCh38
rs985490596 Chr.1: 203218186-203218186 on Build GRCh38
rs550629635 Chr.1: 203218275-203218275 on Build GRCh38
rs1035478742 Chr.1: 203218027-203218027 on Build GRCh38
rs190976402 Chr.1: 203218161-203218161 on Build GRCh38
rs117249305 Chr.1: 203218274-203218274 on Build GRCh38
rs143414130 Chr.1: 203218224-203218225 on Build GRCh38
rs116602841 Chr.1: 203218164-203218164 on Build GRCh38
rs758401351 Chr.1: 203218133-203218133 on Build GRCh38
rs895561035 Chr.1: 203218033-203218033 on Build GRCh38
rs777965042 Chr.1: 203218141-203218141 on Build GRCh38
rs913196885 Chr.1: 203218138-203218138 on Build GRCh38
rs774927713 Chr.1: 203217936-203217936 on Build GRCh38
rs1057287029 Chr.1: 203218227-203218228 on Build GRCh38
rs767273553 Chr.1: 203218080-203218080 on Build GRCh38
rs142943229 Chr.1: 203218096-203218096 on Build GRCh38

View Full Product Details