Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209606228-209606733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK1G
Entrez Gene ID
57172
Gene Name
calcium/calmodulin dependent protein kinase IG
Gene Aliases
CLICK3, CLICKIII, VWS1, dJ272L16.1
UniGene
Hs.199068
Gene Chromosome Location
Chr.1: 209583700-209613939 on Build GRCh38
Transcripts
NM_020439.2 XM_017001866.1 XM_017001867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4668946 c.499C>A p.L167M Chr.1: 209606383-209606383 on Build GRCh38
6840456 c.473C>T p.S158F Chr.1: 209606357-209606357 on Build GRCh38
903284 c.531C>A p.S177S Chr.1: 209606415-209606415 on Build GRCh38
6508581 c.444C>A p.N148K Chr.1: 209606328-209606328 on Build GRCh38
4901492 c.439G>A p.E147K Chr.1: 209606323-209606323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750895420 Chr.1: 209606285-209606285 on Build GRCh38
rs751801643 Chr.1: 209606275-209606275 on Build GRCh38
rs12079523 Chr.1: 209606665-209606665 on Build GRCh38
rs148684307 Chr.1: 209606439-209606439 on Build GRCh38
rs1041426124 Chr.1: 209606586-209606586 on Build GRCh38
rs564514198 Chr.1: 209606530-209606530 on Build GRCh38
rs267598349 Chr.1: 209606400-209606400 on Build GRCh38
rs767522588 Chr.1: 209606366-209606366 on Build GRCh38
rs1017017944 Chr.1: 209606607-209606607 on Build GRCh38
rs777272134 Chr.1: 209606322-209606322 on Build GRCh38
rs764337659 Chr.1: 209606272-209606272 on Build GRCh38
rs775310880 Chr.1: 209606488-209606488 on Build GRCh38
rs1042933785 Chr.1: 209606563-209606563 on Build GRCh38
rs901799454 Chr.1: 209606258-209606258 on Build GRCh38
rs554990333 Chr.1: 209606603-209606603 on Build GRCh38
rs745333577 Chr.1: 209606392-209606392 on Build GRCh38
rs143406129 Chr.1: 209606343-209606343 on Build GRCh38
rs906731818 Chr.1: 209606564-209606564 on Build GRCh38
rs760818427 Chr.1: 209606387-209606387 on Build GRCh38
rs748338518 Chr.1: 209606490-209606490 on Build GRCh38
rs866051094 Chr.1: 209606319-209606319 on Build GRCh38
rs773291208 Chr.1: 209606379-209606379 on Build GRCh38
rs868513059 Chr.1: 209606444-209606444 on Build GRCh38
rs974691050 Chr.1: 209606496-209606496 on Build GRCh38
rs766698214 Chr.1: 209606708-209606708 on Build GRCh38
rs778778827 Chr.1: 209606316-209606316 on Build GRCh38
rs145654288 Chr.1: 209606705-209606705 on Build GRCh38
rs886503897 Chr.1: 209606632-209606632 on Build GRCh38
rs750952802 Chr.1: 209606650-209606650 on Build GRCh38
rs772390239 Chr.1: 209606491-209606491 on Build GRCh38
rs201553423 Chr.1: 209606467-209606467 on Build GRCh38
rs35498535 Chr.1: 209606307-209606307 on Build GRCh38
rs914339904 Chr.1: 209606571-209606571 on Build GRCh38
rs200388906 Chr.1: 209606317-209606317 on Build GRCh38
rs945805665 Chr.1: 209606690-209606690 on Build GRCh38
rs880782 Chr.1: 209606608-209606608 on Build GRCh38
rs935611062 Chr.1: 209606503-209606503 on Build GRCh38
rs149738199 Chr.1: 209606602-209606602 on Build GRCh38
rs77283315 Chr.1: 209606658-209606658 on Build GRCh38
rs551840862 Chr.1: 209606702-209606702 on Build GRCh38
rs537759117 Chr.1: 209606643-209606643 on Build GRCh38
rs796763700 Chr.1: 209606532-209606532 on Build GRCh38
rs767572928 Chr.1: 209606284-209606284 on Build GRCh38
rs994116670 Chr.1: 209606612-209606612 on Build GRCh38
rs1025613414 Chr.1: 209606614-209606614 on Build GRCh38
rs755367866 Chr.1: 209606308-209606308 on Build GRCh38
rs1055344319 Chr.1: 209606590-209606590 on Build GRCh38
rs958893571 Chr.1: 209606656-209606656 on Build GRCh38
rs201992447 Chr.1: 209606462-209606462 on Build GRCh38
rs752297296 Chr.1: 209606436-209606436 on Build GRCh38
rs774446976 Chr.1: 209606351-209606351 on Build GRCh38
rs983665469 Chr.1: 209606403-209606403 on Build GRCh38
rs527593175 Chr.1: 209606686-209606686 on Build GRCh38
rs193260326 Chr.1: 209606704-209606704 on Build GRCh38
rs376936834 Chr.1: 209606293-209606293 on Build GRCh38
rs761856421 Chr.1: 209606282-209606282 on Build GRCh38
rs770903249 Chr.1: 209606329-209606329 on Build GRCh38
rs769577489 Chr.1: 209606473-209606473 on Build GRCh38
rs781164163 Chr.1: 209606466-209606466 on Build GRCh38
rs921592442 Chr.1: 209606497-209606497 on Build GRCh38
rs537943615 Chr.1: 209606259-209606259 on Build GRCh38
rs971597955 Chr.1: 209606636-209606636 on Build GRCh38
rs962736514 Chr.1: 209606363-209606363 on Build GRCh38
rs762023801 Chr.1: 209606364-209606364 on Build GRCh38
rs880783 Chr.1: 209606460-209606460 on Build GRCh38
rs997445080 Chr.1: 209606591-209606591 on Build GRCh38
rs965942550 Chr.1: 209606651-209606651 on Build GRCh38
rs767035299 Chr.1: 209606407-209606407 on Build GRCh38
rs767799311 Chr.1: 209606340-209606340 on Build GRCh38
rs765580376 Chr.1: 209606435-209606435 on Build GRCh38
rs777172542 Chr.1: 209606440-209606440 on Build GRCh38
rs939592066 Chr.1: 209606581-209606581 on Build GRCh38
rs776517584 Chr.1: 209606337-209606337 on Build GRCh38
rs147371769 Chr.1: 209606323-209606323 on Build GRCh38
rs184098780 Chr.1: 209606707-209606707 on Build GRCh38
rs754396153 Chr.1: 209606427-209606427 on Build GRCh38
rs945836331 Chr.1: 209606555-209606555 on Build GRCh38
rs1027660856 Chr.1: 209606331-209606331 on Build GRCh38
rs556793973 Chr.1: 209606305-209606305 on Build GRCh38
rs376907581 Chr.1: 209606548-209606548 on Build GRCh38
rs1054218595 Chr.1: 209606544-209606544 on Build GRCh38
rs755455754 Chr.1: 209606430-209606430 on Build GRCh38

View Full Product Details