Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63490152-63490647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRASLS5
Entrez Gene ID
117245
Gene Name
HRAS like suppressor family member 5
Gene Aliases
HRLP5, HRSL5, RLP1, iNAT
UniGene
Hs.410316
Gene Chromosome Location
Chr.11: 63461404-63491208 on Build GRCh38
Transcripts
NM_001146728.1 NM_001146729.1 NM_054108.3 XM_011544751.2 XM_011544752.2 XM_011544753.1 XM_017017172.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6519198 c.231G>T p.P77P Chr.11: 63490281-63490281 on Build GRCh38
7310222 c.198A>T p.A66A Chr.11: 63490314-63490314 on Build GRCh38
5519049 c.168_169insC p.L57fs*2 Chr.11: 63490343-63490344 on Build GRCh38
6682065 c.232C>A p.P78T Chr.11: 63490280-63490280 on Build GRCh38
1704179 c.185C>T p.S62F Chr.11: 63490327-63490327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201252207 Chr.11: 63490239-63490239 on Build GRCh38
rs34004301 Chr.11: 63490317-63490317 on Build GRCh38
rs778914129 Chr.11: 63490227-63490227 on Build GRCh38
rs568914453 Chr.11: 63490514-63490514 on Build GRCh38
rs896775369 Chr.11: 63490180-63490180 on Build GRCh38
rs749439182 Chr.11: 63490225-63490225 on Build GRCh38
rs150437915 Chr.11: 63490306-63490306 on Build GRCh38
rs114304563 Chr.11: 63490223-63490223 on Build GRCh38
rs903829354 Chr.11: 63490495-63490495 on Build GRCh38
rs185563678 Chr.11: 63490567-63490567 on Build GRCh38
rs747573573 Chr.11: 63490263-63490263 on Build GRCh38
rs762462643 Chr.11: 63490554-63490554 on Build GRCh38
rs867485875 Chr.11: 63490449-63490449 on Build GRCh38
rs997348165 Chr.11: 63490539-63490539 on Build GRCh38
rs142830842 Chr.11: 63490329-63490329 on Build GRCh38
rs1038484087 Chr.11: 63490606-63490606 on Build GRCh38
rs117436678 Chr.11: 63490598-63490598 on Build GRCh38
rs373790972 Chr.11: 63490397-63490397 on Build GRCh38
rs111569913 Chr.11: 63490431-63490431 on Build GRCh38
rs765076212 Chr.11: 63490407-63490407 on Build GRCh38
rs898553821 Chr.11: 63490616-63490616 on Build GRCh38
rs1041911667 Chr.11: 63490473-63490473 on Build GRCh38
rs772502788 Chr.11: 63490242-63490242 on Build GRCh38
rs761603400 Chr.11: 63490375-63490375 on Build GRCh38
rs34908747 Chr.11: 63490348-63490348 on Build GRCh38
rs748242158 Chr.11: 63490238-63490238 on Build GRCh38
rs1030195268 Chr.11: 63490541-63490541 on Build GRCh38
rs768380801 Chr.11: 63490290-63490290 on Build GRCh38
rs758459499 Chr.11: 63490344-63490344 on Build GRCh38
rs1055710316 Chr.11: 63490580-63490580 on Build GRCh38
rs368842108 Chr.11: 63490204-63490204 on Build GRCh38
rs746246342 Chr.11: 63490356-63490356 on Build GRCh38
rs964536267 Chr.11: 63490299-63490299 on Build GRCh38
rs752453390 Chr.11: 63490342-63490343 on Build GRCh38
rs549159971 Chr.11: 63490483-63490483 on Build GRCh38
rs746297293 Chr.11: 63490434-63490434 on Build GRCh38
rs138640590 Chr.11: 63490349-63490349 on Build GRCh38
rs753687897 Chr.11: 63490309-63490309 on Build GRCh38
rs771612086 Chr.11: 63490303-63490303 on Build GRCh38
rs955780393 Chr.11: 63490316-63490316 on Build GRCh38
rs773929456 Chr.11: 63490300-63490300 on Build GRCh38
rs1018124461 Chr.11: 63490271-63490271 on Build GRCh38
rs567323727 Chr.11: 63490443-63490443 on Build GRCh38
rs774131066 Chr.11: 63490373-63490373 on Build GRCh38
rs779112774 Chr.11: 63490325-63490325 on Build GRCh38
rs755258057 Chr.11: 63490226-63490226 on Build GRCh38
rs868334017 Chr.11: 63490341-63490342 on Build GRCh38
rs777098062 Chr.11: 63490281-63490281 on Build GRCh38
rs1056832145 Chr.11: 63490177-63490177 on Build GRCh38
rs945899772 Chr.11: 63490451-63490451 on Build GRCh38
rs1017653513 Chr.11: 63490509-63490509 on Build GRCh38
rs777883276 Chr.11: 63490345-63490345 on Build GRCh38
rs766047379 Chr.11: 63490308-63490308 on Build GRCh38
rs771677418 Chr.11: 63490392-63490392 on Build GRCh38
rs529119022 Chr.11: 63490444-63490444 on Build GRCh38
rs753511605 Chr.11: 63490404-63490404 on Build GRCh38
rs545470378 Chr.11: 63490251-63490251 on Build GRCh38
rs895179920 Chr.11: 63490590-63490590 on Build GRCh38
rs1044131453 Chr.11: 63490399-63490399 on Build GRCh38
rs762476077 Chr.11: 63490282-63490282 on Build GRCh38
rs1001276022 Chr.11: 63490496-63490496 on Build GRCh38
rs895231985 Chr.11: 63490203-63490203 on Build GRCh38
rs565285713 Chr.11: 63490305-63490305 on Build GRCh38
rs752895039 Chr.11: 63490408-63490408 on Build GRCh38
rs759372974 Chr.11: 63490406-63490406 on Build GRCh38
rs747539887 Chr.11: 63490280-63490280 on Build GRCh38
rs767190469 Chr.11: 63490302-63490302 on Build GRCh38
rs1003557350 Chr.11: 63490176-63490176 on Build GRCh38
rs770346695 Chr.11: 63490366-63490366 on Build GRCh38
rs1021174946 Chr.11: 63490494-63490494 on Build GRCh38
rs962896455 Chr.11: 63490276-63490276 on Build GRCh38
rs775022524 Chr.11: 63490553-63490553 on Build GRCh38
rs772879743 Chr.11: 63490394-63490394 on Build GRCh38
rs994355583 Chr.11: 63490622-63490622 on Build GRCh38
rs755844679 Chr.11: 63490216-63490216 on Build GRCh38
rs199684544 Chr.11: 63490237-63490237 on Build GRCh38
rs764939349 Chr.11: 63490234-63490236 on Build GRCh38

View Full Product Details