Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75299457-75300026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
UniGene
Hs.733835
Gene Chromosome Location
Chr.12: 75275979-75390941 on Build GRCh38
Transcripts
NM_001286547.1 NM_001286548.1 NM_001286549.1 NM_032606.3 XM_005269192.1 XM_005269194.3 XM_006719648.2 XM_006719649.2 XM_011538878.1 XM_011538886.2 XM_011538887.2 XM_011538888.1 XM_011538889.2 XM_011538890.1 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020036.1 XM_017020037.1 XM_017020038.1 XM_017020039.1 XM_017020041.1 XM_017020042.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 XM_017020046.1 XM_017020047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2102361 c.197A>G p.D66G Chr.12: 75299855-75299855 on Build GRCh38
2102360 c.893A>G p.D298G Chr.12: 75299855-75299855 on Build GRCh38
7327038 c.184A>C p.K62Q Chr.12: 75299868-75299868 on Build GRCh38
943465 c.911G>A p.R304H Chr.12: 75299837-75299837 on Build GRCh38
3955150 c.908C>T p.S303L Chr.12: 75299840-75299840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11835391 Chr.12: 75299623-75299623 on Build GRCh38
rs763001921 Chr.12: 75299875-75299875 on Build GRCh38
rs556740394 Chr.12: 75299840-75299840 on Build GRCh38
rs531793088 Chr.12: 75299863-75299863 on Build GRCh38
rs1045436795 Chr.12: 75299706-75299706 on Build GRCh38
rs940311383 Chr.12: 75299647-75299647 on Build GRCh38
rs7301532 Chr.12: 75299971-75299971 on Build GRCh38
rs572772192 Chr.12: 75299493-75299493 on Build GRCh38
rs752529603 Chr.12: 75299830-75299830 on Build GRCh38
rs888802605 Chr.12: 75299992-75299992 on Build GRCh38
rs771116692 Chr.12: 75299878-75299878 on Build GRCh38
rs764082293 Chr.12: 75299833-75299833 on Build GRCh38
rs1041509662 Chr.12: 75299781-75299781 on Build GRCh38
rs562989650 Chr.12: 75299838-75299838 on Build GRCh38
rs765142348 Chr.12: 75299920-75299920 on Build GRCh38
rs182679854 Chr.12: 75299491-75299491 on Build GRCh38
rs754678901 Chr.12: 75299952-75299952 on Build GRCh38
rs371673073 Chr.12: 75299855-75299855 on Build GRCh38
rs901610082 Chr.12: 75299978-75299978 on Build GRCh38
rs757308732 Chr.12: 75299835-75299835 on Build GRCh38
rs758053512 Chr.12: 75299844-75299844 on Build GRCh38
rs757852971 Chr.12: 75299931-75299931 on Build GRCh38
rs957876426 Chr.12: 75299596-75299597 on Build GRCh38
rs1053970688 Chr.12: 75299748-75299748 on Build GRCh38
rs138897272 Chr.12: 75299804-75299804 on Build GRCh38
rs142306209 Chr.12: 75299692-75299692 on Build GRCh38
rs1006120818 Chr.12: 75299820-75299820 on Build GRCh38
rs914157166 Chr.12: 75299970-75299970 on Build GRCh38
rs774744790 Chr.12: 75299888-75299888 on Build GRCh38
rs536731071 Chr.12: 75299834-75299834 on Build GRCh38
rs201023122 Chr.12: 75299870-75299870 on Build GRCh38
rs953307700 Chr.12: 75299615-75299615 on Build GRCh38
rs566689942 Chr.12: 75299988-75299988 on Build GRCh38
rs989247263 Chr.12: 75299602-75299602 on Build GRCh38
rs972146013 Chr.12: 75299630-75299630 on Build GRCh38
rs763959666 Chr.12: 75299901-75299901 on Build GRCh38
rs1041577122 Chr.12: 75299973-75299973 on Build GRCh38
rs901582939 Chr.12: 75299790-75299790 on Build GRCh38
rs770243775 Chr.12: 75299866-75299866 on Build GRCh38
rs373936180 Chr.12: 75299900-75299900 on Build GRCh38
rs998900701 Chr.12: 75299496-75299496 on Build GRCh38
rs146189578 Chr.12: 75299699-75299699 on Build GRCh38
rs760497161 Chr.12: 75299889-75299889 on Build GRCh38
rs7486168 Chr.12: 75299520-75299520 on Build GRCh38
rs779584223 Chr.12: 75299845-75299845 on Build GRCh38
rs370892002 Chr.12: 75299913-75299913 on Build GRCh38
rs765978857 Chr.12: 75299934-75299934 on Build GRCh38
rs1054066509 Chr.12: 75299965-75299965 on Build GRCh38
rs138519970 Chr.12: 75299837-75299837 on Build GRCh38
rs551696052 Chr.12: 75299981-75299981 on Build GRCh38
rs781631186 Chr.12: 75299860-75299860 on Build GRCh38
rs753865795 Chr.12: 75299899-75299900 on Build GRCh38
rs747309615 Chr.12: 75299853-75299853 on Build GRCh38
rs11180451 Chr.12: 75299740-75299740 on Build GRCh38
rs781504795 Chr.12: 75299963-75299963 on Build GRCh38
rs796342357 Chr.12: 75299990-75299990 on Build GRCh38
rs760548522 Chr.12: 75299832-75299832 on Build GRCh38
rs926898407 Chr.12: 75299721-75299721 on Build GRCh38
rs61932923 Chr.12: 75299751-75299751 on Build GRCh38
rs749888844 Chr.12: 75299921-75299921 on Build GRCh38
rs369182922 Chr.12: 75299704-75299707 on Build GRCh38
rs149297211 Chr.12: 75299877-75299877 on Build GRCh38
rs1036127740 Chr.12: 75299679-75299679 on Build GRCh38
rs545501362 Chr.12: 75299614-75299614 on Build GRCh38
rs117647160 Chr.12: 75299911-75299911 on Build GRCh38
rs34555729 Chr.12: 75299599-75299600 on Build GRCh38
rs546852948 Chr.12: 75299985-75299985 on Build GRCh38
rs751238454 Chr.12: 75299935-75299935 on Build GRCh38
rs766579082 Chr.12: 75299815-75299815 on Build GRCh38
rs558606793 Chr.12: 75299533-75299533 on Build GRCh38
rs574699088 Chr.12: 75299722-75299722 on Build GRCh38
rs1029050670 Chr.12: 75299799-75299799 on Build GRCh38
rs1009722778 Chr.12: 75299774-75299774 on Build GRCh38
rs532515282 Chr.12: 75299791-75299791 on Build GRCh38
rs909019583 Chr.12: 75299613-75299613 on Build GRCh38
rs1046367722 Chr.12: 75299775-75299775 on Build GRCh38
rs367568156 Chr.12: 75299852-75299852 on Build GRCh38
rs1056015062 Chr.12: 75299986-75299986 on Build GRCh38
rs36046697 Chr.12: 75299981-75299981 on Build GRCh38
rs575286509 Chr.12: 75299668-75299668 on Build GRCh38

View Full Product Details