Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118554899-118555402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT46
Entrez Gene ID
56912
Gene Name
intraflagellar transport 46
Gene Aliases
C11orf2, C11orf60, CFAP32
UniGene
Hs.533738
Gene Chromosome Location
Chr.11: 118544528-118575624 on Build GRCh38
Transcripts
NM_001168618.1 NM_020153.3 XM_011542905.2 XM_011542906.2 XM_017018017.1 XM_017018018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6436009 c.370C>T p.P124S Chr.11: 118555291-118555291 on Build GRCh38
6067579 c.507G>C p.K169N Chr.11: 118554990-118554990 on Build GRCh38
466409 c.483C>A p.V161V Chr.11: 118555014-118555014 on Build GRCh38
6014690 c.414-5C>T p.? Chr.11: 118555088-118555088 on Build GRCh38
923615 c.459C>T p.F153F Chr.11: 118555038-118555038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112341118 Chr.11: 118555163-118555163 on Build GRCh38
rs760516828 Chr.11: 118555372-118555372 on Build GRCh38
rs77812853 Chr.11: 118555347-118555347 on Build GRCh38
rs782112434 Chr.11: 118555079-118555079 on Build GRCh38
rs782745091 Chr.11: 118555086-118555086 on Build GRCh38
rs782604953 Chr.11: 118555221-118555221 on Build GRCh38
rs377719348 Chr.11: 118555145-118555146 on Build GRCh38
rs782457822 Chr.11: 118555228-118555228 on Build GRCh38
rs782212290 Chr.11: 118555284-118555284 on Build GRCh38
rs782494445 Chr.11: 118555207-118555207 on Build GRCh38
rs781857636 Chr.11: 118555205-118555205 on Build GRCh38
rs782283703 Chr.11: 118555071-118555071 on Build GRCh38
rs782782834 Chr.11: 118555202-118555202 on Build GRCh38
rs189609518 Chr.11: 118554939-118554939 on Build GRCh38
rs782609288 Chr.11: 118555013-118555013 on Build GRCh38
rs782750393 Chr.11: 118555326-118555326 on Build GRCh38
rs143192297 Chr.11: 118555317-118555317 on Build GRCh38
rs782089193 Chr.11: 118554944-118554944 on Build GRCh38
rs781828935 Chr.11: 118555327-118555327 on Build GRCh38
rs781995934 Chr.11: 118555077-118555077 on Build GRCh38
rs1043102929 Chr.11: 118555311-118555311 on Build GRCh38
rs782228695 Chr.11: 118555211-118555212 on Build GRCh38
rs139391241 Chr.11: 118555251-118555251 on Build GRCh38
rs1041354477 Chr.11: 118555271-118555271 on Build GRCh38
rs782143718 Chr.11: 118554976-118554976 on Build GRCh38
rs782344309 Chr.11: 118555059-118555059 on Build GRCh38
rs375847066 Chr.11: 118555290-118555290 on Build GRCh38
rs782707333 Chr.11: 118555342-118555342 on Build GRCh38
rs368173666 Chr.11: 118555036-118555036 on Build GRCh38
rs782775929 Chr.11: 118554985-118554985 on Build GRCh38
rs782529471 Chr.11: 118555133-118555133 on Build GRCh38
rs782464871 Chr.11: 118555353-118555353 on Build GRCh38
rs150117746 Chr.11: 118555005-118555005 on Build GRCh38
rs782652017 Chr.11: 118555233-118555233 on Build GRCh38
rs555586147 Chr.11: 118555142-118555142 on Build GRCh38
rs782463624 Chr.11: 118555335-118555335 on Build GRCh38
rs200129173 Chr.11: 118555042-118555042 on Build GRCh38
rs782808904 Chr.11: 118555306-118555306 on Build GRCh38
rs998727846 Chr.11: 118555031-118555031 on Build GRCh38
rs781961185 Chr.11: 118555281-118555281 on Build GRCh38
rs782220462 Chr.11: 118555348-118555348 on Build GRCh38
rs148823925 Chr.11: 118555049-118555049 on Build GRCh38
rs781935786 Chr.11: 118555131-118555131 on Build GRCh38
rs782323249 Chr.11: 118555286-118555286 on Build GRCh38
rs781917504 Chr.11: 118555195-118555195 on Build GRCh38
rs944278383 Chr.11: 118555248-118555248 on Build GRCh38
rs546673640 Chr.11: 118555184-118555184 on Build GRCh38
rs377244915 Chr.11: 118554963-118554963 on Build GRCh38
rs781905648 Chr.11: 118555124-118555124 on Build GRCh38
rs904286287 Chr.11: 118555341-118555341 on Build GRCh38
rs781804734 Chr.11: 118555014-118555014 on Build GRCh38
rs781880943 Chr.11: 118555345-118555345 on Build GRCh38
rs573166702 Chr.11: 118555378-118555378 on Build GRCh38
rs782477133 Chr.11: 118555010-118555010 on Build GRCh38
rs143391860 Chr.11: 118555075-118555075 on Build GRCh38
rs376221796 Chr.11: 118555360-118555360 on Build GRCh38
rs782689893 Chr.11: 118555123-118555123 on Build GRCh38
rs782285232 Chr.11: 118555034-118555034 on Build GRCh38
rs557195802 Chr.11: 118555287-118555287 on Build GRCh38
rs375629023 Chr.11: 118555188-118555188 on Build GRCh38
rs782199189 Chr.11: 118555225-118555225 on Build GRCh38
rs145438119 Chr.11: 118555043-118555043 on Build GRCh38
rs781946028 Chr.11: 118555064-118555064 on Build GRCh38
rs908669819 Chr.11: 118555223-118555223 on Build GRCh38
rs782682536 Chr.11: 118555019-118555019 on Build GRCh38
rs566106571 Chr.11: 118554981-118554981 on Build GRCh38
rs554934951 Chr.11: 118555366-118555366 on Build GRCh38
rs782065735 Chr.11: 118555110-118555110 on Build GRCh38
rs782369335 Chr.11: 118554947-118554947 on Build GRCh38
rs781945654 Chr.11: 118555094-118555094 on Build GRCh38
rs782584112 Chr.11: 118555356-118555356 on Build GRCh38
rs372794442 Chr.11: 118555255-118555255 on Build GRCh38

View Full Product Details