Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 120337181-120337679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF12
Entrez Gene ID
23365
Gene Name
Rho guanine nucleotide exchange factor 12
Gene Aliases
LARG, PRO2792
UniGene
Hs.24598
Gene Chromosome Location
Chr.11: 120336276-120489936 on Build GRCh38
Transcripts
NM_001198665.1 NM_015313.2 XM_006718805.3 XM_011542720.2 XM_017017420.1 XM_017017421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4880226 c.1A>G p.M1V Chr.11: 120337244-120337244 on Build GRCh38
3666864 c.22A>G p.I8V Chr.11: 120337265-120337265 on Build GRCh38
6330628 c.5G>A p.S2N Chr.11: 120337248-120337248 on Build GRCh38
4943867 c.19A>G p.T7A Chr.11: 120337262-120337262 on Build GRCh38
923933 c.25A>T p.T9S Chr.11: 120337268-120337268 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201108097 Chr.11: 120337240-120337240 on Build GRCh38
rs760705048 Chr.11: 120337310-120337310 on Build GRCh38
rs749687348 Chr.11: 120337260-120337260 on Build GRCh38
rs775586284 Chr.11: 120337301-120337301 on Build GRCh38
rs779942827 Chr.11: 120337654-120337654 on Build GRCh38
rs1010790938 Chr.11: 120337634-120337634 on Build GRCh38
rs368974955 Chr.11: 120337550-120337550 on Build GRCh38
rs4936515 Chr.11: 120337476-120337476 on Build GRCh38
rs889528241 Chr.11: 120337618-120337618 on Build GRCh38
rs774639794 Chr.11: 120337270-120337270 on Build GRCh38
rs766885844 Chr.11: 120337205-120337205 on Build GRCh38
rs763718529 Chr.11: 120337374-120337374 on Build GRCh38
rs942454463 Chr.11: 120337621-120337621 on Build GRCh38
rs900848315 Chr.11: 120337641-120337641 on Build GRCh38
rs754652014 Chr.11: 120337207-120337207 on Build GRCh38
rs377292584 Chr.11: 120337418-120337418 on Build GRCh38
rs771144910 Chr.11: 120337265-120337265 on Build GRCh38
rs900503629 Chr.11: 120337518-120337518 on Build GRCh38
rs527814572 Chr.11: 120337629-120337629 on Build GRCh38
rs750390778 Chr.11: 120337502-120337502 on Build GRCh38
rs1020721180 Chr.11: 120337643-120337643 on Build GRCh38
rs978622419 Chr.11: 120337414-120337414 on Build GRCh38
rs759744899 Chr.11: 120337271-120337271 on Build GRCh38
rs185401242 Chr.11: 120337510-120337510 on Build GRCh38
rs1036634106 Chr.11: 120337506-120337506 on Build GRCh38
rs752013308 Chr.11: 120337235-120337235 on Build GRCh38
rs979067885 Chr.11: 120337306-120337306 on Build GRCh38
rs968564094 Chr.11: 120337407-120337407 on Build GRCh38
rs201366154 Chr.11: 120337294-120337297 on Build GRCh38
rs920868164 Chr.11: 120337546-120337546 on Build GRCh38
rs917265245 Chr.11: 120337483-120337483 on Build GRCh38
rs780902732 Chr.11: 120337231-120337231 on Build GRCh38
rs551267432 Chr.11: 120337296-120337296 on Build GRCh38
rs563181274 Chr.11: 120337415-120337415 on Build GRCh38
rs1010342479 Chr.11: 120337648-120337648 on Build GRCh38
rs924921842 Chr.11: 120337378-120337378 on Build GRCh38
rs773819188 Chr.11: 120337241-120337241 on Build GRCh38
rs1006503708 Chr.11: 120337216-120337216 on Build GRCh38
rs370844009 Chr.11: 120337324-120337324 on Build GRCh38
rs1037697202 Chr.11: 120337517-120337517 on Build GRCh38
rs4938799 Chr.11: 120337548-120337548 on Build GRCh38
rs199551576 Chr.11: 120337298-120337301 on Build GRCh38
rs992788014 Chr.11: 120337482-120337482 on Build GRCh38
rs756727657 Chr.11: 120337448-120337448 on Build GRCh38
rs940911673 Chr.11: 120337494-120337494 on Build GRCh38
rs949200401 Chr.11: 120337463-120337463 on Build GRCh38
rs927356310 Chr.11: 120337461-120337461 on Build GRCh38
rs767843124 Chr.11: 120337239-120337239 on Build GRCh38
rs753461038 Chr.11: 120337402-120337402 on Build GRCh38
rs996425104 Chr.11: 120337539-120337539 on Build GRCh38

View Full Product Details