Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75281859-75282401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
UniGene
Hs.733835
Gene Chromosome Location
Chr.12: 75275979-75390941 on Build GRCh38
Transcripts
NM_001286547.1 NM_001286548.1 NM_001286549.1 NM_032606.3 XM_005269192.1 XM_005269194.3 XM_006719648.2 XM_006719649.2 XM_011538878.1 XM_011538886.2 XM_011538887.2 XM_011538888.1 XM_011538889.2 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020036.1 XM_017020037.1 XM_017020038.1 XM_017020039.1 XM_017020040.1 XM_017020041.1 XM_017020042.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 XM_017020046.1 XM_017020047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4798275 c.973G>A p.G325S Chr.12: 75282251-75282251 on Build GRCh38
943451 c.1647G>T p.K549N Chr.12: 75282273-75282273 on Build GRCh38
7359472 c.1664T>C p.I555T Chr.12: 75282256-75282256 on Build GRCh38
6462951 c.900C>A p.N300K Chr.12: 75282324-75282324 on Build GRCh38
6462949 c.901A>C p.K301Q Chr.12: 75282323-75282323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550338080 Chr.12: 75281901-75281901 on Build GRCh38
rs762492521 Chr.12: 75282229-75282229 on Build GRCh38
rs764688065 Chr.12: 75282224-75282224 on Build GRCh38
rs781146949 Chr.12: 75282336-75282336 on Build GRCh38
rs766691480 Chr.12: 75282190-75282190 on Build GRCh38
rs775011447 Chr.12: 75281883-75281883 on Build GRCh38
rs898122920 Chr.12: 75281952-75281952 on Build GRCh38
rs889280754 Chr.12: 75282185-75282185 on Build GRCh38
rs923553452 Chr.12: 75282131-75282131 on Build GRCh38
rs981297233 Chr.12: 75281978-75281978 on Build GRCh38
rs748060593 Chr.12: 75282341-75282341 on Build GRCh38
rs768564295 Chr.12: 75282202-75282202 on Build GRCh38
rs1050590626 Chr.12: 75282148-75282148 on Build GRCh38
rs558378225 Chr.12: 75282305-75282305 on Build GRCh38
rs1024963027 Chr.12: 75281935-75281935 on Build GRCh38
rs374467580 Chr.12: 75282117-75282117 on Build GRCh38
rs1049654346 Chr.12: 75282163-75282163 on Build GRCh38
rs774851586 Chr.12: 75281914-75281914 on Build GRCh38
rs767013925 Chr.12: 75282295-75282295 on Build GRCh38
rs751969624 Chr.12: 75282240-75282240 on Build GRCh38
rs548729955 Chr.12: 75282081-75282081 on Build GRCh38
rs185799511 Chr.12: 75282050-75282050 on Build GRCh38
rs370678711 Chr.12: 75282241-75282241 on Build GRCh38
rs753174404 Chr.12: 75282259-75282259 on Build GRCh38
rs990015598 Chr.12: 75282138-75282138 on Build GRCh38
rs932584651 Chr.12: 75282162-75282162 on Build GRCh38
rs1034192667 Chr.12: 75282278-75282278 on Build GRCh38
rs761324475 Chr.12: 75282208-75282208 on Build GRCh38
rs768069926 Chr.12: 75282256-75282256 on Build GRCh38
rs1025540309 Chr.12: 75281967-75281967 on Build GRCh38
rs776451194 Chr.12: 75282203-75282203 on Build GRCh38
rs772566795 Chr.12: 75282349-75282349 on Build GRCh38
rs148159328 Chr.12: 75282265-75282265 on Build GRCh38
rs766176571 Chr.12: 75282238-75282238 on Build GRCh38
rs750609568 Chr.12: 75282091-75282091 on Build GRCh38
rs375729636 Chr.12: 75282261-75282261 on Build GRCh38
rs777691317 Chr.12: 75282263-75282263 on Build GRCh38
rs890834174 Chr.12: 75282168-75282168 on Build GRCh38
rs971293173 Chr.12: 75281976-75281976 on Build GRCh38
rs367743007 Chr.12: 75282033-75282033 on Build GRCh38
rs370297566 Chr.12: 75282360-75282360 on Build GRCh38
rs762240298 Chr.12: 75281888-75281888 on Build GRCh38
rs757463074 Chr.12: 75282299-75282299 on Build GRCh38
rs777513432 Chr.12: 75282360-75282361 on Build GRCh38
rs772874381 Chr.12: 75282227-75282227 on Build GRCh38
rs199705076 Chr.12: 75282301-75282301 on Build GRCh38
rs559999432 Chr.12: 75281916-75281916 on Build GRCh38
rs959078700 Chr.12: 75282019-75282019 on Build GRCh38
rs767254087 Chr.12: 75281988-75281988 on Build GRCh38
rs147118041 Chr.12: 75281956-75281956 on Build GRCh38
rs768447568 Chr.12: 75282325-75282325 on Build GRCh38
rs933554363 Chr.12: 75282140-75282140 on Build GRCh38
rs535399911 Chr.12: 75282070-75282070 on Build GRCh38
rs977656826 Chr.12: 75282090-75282090 on Build GRCh38
rs573116604 Chr.12: 75281927-75281927 on Build GRCh38
rs946044033 Chr.12: 75282073-75282073 on Build GRCh38
rs549624222 Chr.12: 75281993-75281993 on Build GRCh38
rs181964896 Chr.12: 75281894-75281894 on Build GRCh38
rs371057527 Chr.12: 75281929-75281929 on Build GRCh38
rs769821016 Chr.12: 75282347-75282347 on Build GRCh38

View Full Product Details