Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75289500-75290000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
UniGene
Hs.733835
Gene Chromosome Location
Chr.12: 75275979-75390941 on Build GRCh38
Transcripts
NM_001286547.1 NM_001286548.1 NM_001286549.1 NM_032606.3 XM_005269192.1 XM_005269194.3 XM_006719648.2 XM_006719649.2 XM_011538878.1 XM_011538886.2 XM_011538887.2 XM_011538888.1 XM_011538889.2 XM_011538890.1 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020036.1 XM_017020037.1 XM_017020038.1 XM_017020039.1 XM_017020040.1 XM_017020041.1 XM_017020042.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 XM_017020046.1 XM_017020047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6476672 c.619C>A p.L207I Chr.12: 75289758-75289758 on Build GRCh38
7304688 c.1402G>A p.A468T Chr.12: 75289671-75289671 on Build GRCh38
943462 c.613G>T p.E205* Chr.12: 75289764-75289764 on Build GRCh38
943460 c.636T>C p.V212V Chr.12: 75289741-75289741 on Build GRCh38
4917170 c.1313A>G p.K438R Chr.12: 75289760-75289760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763698424 Chr.12: 75289635-75289635 on Build GRCh38
rs775857438 Chr.12: 75289796-75289796 on Build GRCh38
rs748456589 Chr.12: 75289754-75289754 on Build GRCh38
rs999652144 Chr.12: 75289573-75289573 on Build GRCh38
rs776951767 Chr.12: 75289651-75289651 on Build GRCh38
rs375986205 Chr.12: 75289577-75289577 on Build GRCh38
rs111709227 Chr.12: 75289817-75289817 on Build GRCh38
rs144198119 Chr.12: 75289740-75289740 on Build GRCh38
rs149880273 Chr.12: 75289836-75289836 on Build GRCh38
rs778213391 Chr.12: 75289772-75289772 on Build GRCh38
rs749779254 Chr.12: 75289777-75289777 on Build GRCh38
rs757255362 Chr.12: 75289620-75289620 on Build GRCh38
rs199654216 Chr.12: 75289739-75289739 on Build GRCh38
rs918342023 Chr.12: 75289701-75289701 on Build GRCh38
rs759771107 Chr.12: 75289705-75289705 on Build GRCh38
rs765776044 Chr.12: 75289589-75289589 on Build GRCh38
rs139225720 Chr.12: 75289677-75289677 on Build GRCh38
rs753668971 Chr.12: 75289723-75289723 on Build GRCh38
rs766296728 Chr.12: 75289822-75289822 on Build GRCh38
rs978036351 Chr.12: 75289569-75289569 on Build GRCh38
rs113629658 Chr.12: 75289766-75289766 on Build GRCh38
rs543030476 Chr.12: 75289944-75289944 on Build GRCh38
rs746116728 Chr.12: 75289585-75289587 on Build GRCh38
rs889618892 Chr.12: 75289942-75289942 on Build GRCh38
rs571969398 Chr.12: 75289834-75289834 on Build GRCh38
rs367762993 Chr.12: 75289683-75289683 on Build GRCh38
rs771063948 Chr.12: 75289691-75289691 on Build GRCh38
rs201840767 Chr.12: 75289676-75289676 on Build GRCh38
rs763926727 Chr.12: 75289609-75289609 on Build GRCh38
rs573037660 Chr.12: 75289965-75289965 on Build GRCh38
rs765143265 Chr.12: 75289797-75289797 on Build GRCh38
rs146549801 Chr.12: 75289753-75289753 on Build GRCh38
rs779048611 Chr.12: 75289630-75289630 on Build GRCh38
rs777078192 Chr.12: 75289571-75289571 on Build GRCh38
rs753724103 Chr.12: 75289612-75289612 on Build GRCh38
rs367774244 Chr.12: 75289725-75289725 on Build GRCh38
rs548767303 Chr.12: 75289717-75289717 on Build GRCh38
rs955013496 Chr.12: 75289595-75289595 on Build GRCh38
rs781742906 Chr.12: 75289833-75289833 on Build GRCh38
rs182933190 Chr.12: 75289547-75289547 on Build GRCh38
rs779529108 Chr.12: 75289633-75289633 on Build GRCh38
rs1019529464 Chr.12: 75289949-75289949 on Build GRCh38
rs894549165 Chr.12: 75289540-75289540 on Build GRCh38
rs765196232 Chr.12: 75289727-75289727 on Build GRCh38
rs112805358 Chr.12: 75289632-75289632 on Build GRCh38
rs370428667 Chr.12: 75289647-75289647 on Build GRCh38
rs553028978 Chr.12: 75289946-75289946 on Build GRCh38
rs755815175 Chr.12: 75289601-75289601 on Build GRCh38
rs746700405 Chr.12: 75289637-75289637 on Build GRCh38
rs1043311070 Chr.12: 75289933-75289933 on Build GRCh38
rs772146664 Chr.12: 75289789-75289790 on Build GRCh38
rs762632179 Chr.12: 75289597-75289597 on Build GRCh38
rs191110069 Chr.12: 75289803-75289803 on Build GRCh38
rs754658078 Chr.12: 75289742-75289742 on Build GRCh38
rs763153348 Chr.12: 75289596-75289596 on Build GRCh38
rs183422645 Chr.12: 75289858-75289858 on Build GRCh38
rs771382042 Chr.12: 75289783-75289783 on Build GRCh38
rs916734248 Chr.12: 75289752-75289752 on Build GRCh38
rs573920293 Chr.12: 75289877-75289877 on Build GRCh38
rs774483081 Chr.12: 75289785-75289785 on Build GRCh38
rs555876843 Chr.12: 75289853-75289853 on Build GRCh38
rs967609809 Chr.12: 75289563-75289563 on Build GRCh38
rs772360094 Chr.12: 75289794-75289794 on Build GRCh38
rs1021570877 Chr.12: 75289558-75289558 on Build GRCh38
rs773276589 Chr.12: 75289592-75289592 on Build GRCh38
rs762921051 Chr.12: 75289812-75289812 on Build GRCh38
rs751069184 Chr.12: 75289827-75289827 on Build GRCh38
rs555686849 Chr.12: 75289894-75289894 on Build GRCh38
rs750402070 Chr.12: 75289732-75289732 on Build GRCh38
rs759592120 Chr.12: 75289790-75289790 on Build GRCh38
rs748579510 Chr.12: 75289653-75289653 on Build GRCh38
rs540817996 Chr.12: 75289966-75289966 on Build GRCh38
rs539039943 Chr.12: 75289829-75289829 on Build GRCh38
rs774342958 Chr.12: 75289696-75289696 on Build GRCh38

View Full Product Details