Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 76344354-76344866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS10
Entrez Gene ID
79738
Gene Name
Bardet-Biedl syndrome 10
Gene Aliases
C12orf58
UniGene
Hs.96322
Gene Chromosome Location
Chr.12: 76344486-76348442 on Build GRCh38
Transcripts
NM_024685.3

SNP Detail

SNP ID Chromosome Location
rs985831711 Chr.12: 76344673-76344673 on Build GRCh38
rs3087711 Chr.12: 76344762-76344762 on Build GRCh38
rs565888621 Chr.12: 76344701-76344701 on Build GRCh38
rs938686534 Chr.12: 76344800-76344800 on Build GRCh38
rs886049841 Chr.12: 76344668-76344668 on Build GRCh38
rs1041290249 Chr.12: 76344451-76344452 on Build GRCh38
rs894532627 Chr.12: 76344827-76344827 on Build GRCh38
rs1055821608 Chr.12: 76344801-76344801 on Build GRCh38
rs910327467 Chr.12: 76344676-76344676 on Build GRCh38
rs922929416 Chr.12: 76344649-76344649 on Build GRCh38
rs890037203 Chr.12: 76344413-76344413 on Build GRCh38
rs886049840 Chr.12: 76344623-76344624 on Build GRCh38
rs1047238153 Chr.12: 76344842-76344842 on Build GRCh38
rs569852661 Chr.12: 76344659-76344659 on Build GRCh38
rs987981156 Chr.12: 76344668-76344668 on Build GRCh38
rs1050174142 Chr.12: 76344391-76344391 on Build GRCh38
rs554886819 Chr.12: 76344452-76344452 on Build GRCh38
rs555388982 Chr.12: 76344687-76344687 on Build GRCh38
rs886049839 Chr.12: 76344609-76344609 on Build GRCh38
rs993443025 Chr.12: 76344485-76344485 on Build GRCh38
rs1007061047 Chr.12: 76344438-76344438 on Build GRCh38
rs375193000 Chr.12: 76344395-76344395 on Build GRCh38
rs973617163 Chr.12: 76344723-76344723 on Build GRCh38
rs182439491 Chr.12: 76344563-76344563 on Build GRCh38
rs1037564922 Chr.12: 76344462-76344462 on Build GRCh38
rs886049842 Chr.12: 76344673-76344673 on Build GRCh38
rs368516705 Chr.12: 76344519-76344519 on Build GRCh38
rs897714079 Chr.12: 76344463-76344463 on Build GRCh38
rs115516881 Chr.12: 76344565-76344565 on Build GRCh38
rs1043013282 Chr.12: 76344407-76344407 on Build GRCh38
rs977702326 Chr.12: 76344637-76344637 on Build GRCh38
rs886049838 Chr.12: 76344569-76344572 on Build GRCh38
rs901410812 Chr.12: 76344825-76344825 on Build GRCh38
rs1011726926 Chr.12: 76344600-76344600 on Build GRCh38
rs191208685 Chr.12: 76344484-76344484 on Build GRCh38
rs1011674525 Chr.12: 76344834-76344834 on Build GRCh38
rs907239849 Chr.12: 76344507-76344507 on Build GRCh38
rs930148037 Chr.12: 76344832-76344832 on Build GRCh38
rs571032291 Chr.12: 76344488-76344488 on Build GRCh38
rs576987597 Chr.12: 76344840-76344840 on Build GRCh38
rs941803629 Chr.12: 76344717-76344717 on Build GRCh38
rs1005240030 Chr.12: 76344538-76344538 on Build GRCh38
rs1021806409 Chr.12: 76344616-76344616 on Build GRCh38
rs907185176 Chr.12: 76344790-76344790 on Build GRCh38
rs886049837 Chr.12: 76344538-76344538 on Build GRCh38
rs534103908 Chr.12: 76344727-76344727 on Build GRCh38
rs1002950898 Chr.12: 76344527-76344527 on Build GRCh38
rs941898528 Chr.12: 76344453-76344453 on Build GRCh38
rs967997882 Chr.12: 76344618-76344618 on Build GRCh38
rs528033396 Chr.12: 76344474-76344474 on Build GRCh38
rs954414018 Chr.12: 76344668-76344668 on Build GRCh38
rs956772858 Chr.12: 76344667-76344667 on Build GRCh38
rs535547043 Chr.12: 76344680-76344680 on Build GRCh38
rs753528860 Chr.12: 76344815-76344815 on Build GRCh38
rs942712425 Chr.12: 76344788-76344788 on Build GRCh38
rs994327877 Chr.12: 76344496-76344496 on Build GRCh38
rs919105577 Chr.12: 76344726-76344726 on Build GRCh38

View Full Product Details