Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83017560-83018106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD1
Entrez Gene ID
53339
Gene Name
BTB domain containing 1
Gene Aliases
C15orf1, NS5ATP8
UniGene
Hs.459149
Gene Chromosome Location
Chr.15: 83016423-83067434 on Build GRCh38
Transcripts
NM_001011885.1 NM_025238.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7413002 c.1412A>G p.E471G Chr.15: 83018104-83018104 on Build GRCh38
3504616 c.1421A>G p.Q474R Chr.15: 83018095-83018095 on Build GRCh38
1478407 c.1420C>A p.Q474K Chr.15: 83018096-83018096 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769264663 Chr.15: 83018027-83018027 on Build GRCh38
rs745529203 Chr.15: 83018018-83018018 on Build GRCh38
rs553935846 Chr.15: 83017977-83017977 on Build GRCh38
rs887682312 Chr.15: 83017786-83017786 on Build GRCh38
rs567456564 Chr.15: 83017591-83017591 on Build GRCh38
rs1040544365 Chr.15: 83017962-83017962 on Build GRCh38
rs1029128991 Chr.15: 83017657-83017657 on Build GRCh38
rs772989747 Chr.15: 83018016-83018016 on Build GRCh38
rs529688914 Chr.15: 83017615-83017615 on Build GRCh38
rs189317063 Chr.15: 83017858-83017858 on Build GRCh38
rs373426191 Chr.15: 83018051-83018051 on Build GRCh38
rs201126475 Chr.15: 83018050-83018050 on Build GRCh38
rs897553569 Chr.15: 83017993-83017993 on Build GRCh38
rs577225234 Chr.15: 83017794-83017794 on Build GRCh38
rs542866416 Chr.15: 83017751-83017751 on Build GRCh38
rs1040090068 Chr.15: 83017703-83017703 on Build GRCh38
rs995925484 Chr.15: 83017640-83017640 on Build GRCh38
rs1055817764 Chr.15: 83017839-83017839 on Build GRCh38
rs897498508 Chr.15: 83017728-83017728 on Build GRCh38
rs944457383 Chr.15: 83017676-83017676 on Build GRCh38
rs775222265 Chr.15: 83018032-83018032 on Build GRCh38
rs767575976 Chr.15: 83018072-83018072 on Build GRCh38
rs763181393 Chr.15: 83018033-83018033 on Build GRCh38
rs769042541 Chr.15: 83018045-83018045 on Build GRCh38
rs971212466 Chr.15: 83017830-83017830 on Build GRCh38
rs184980550 Chr.15: 83017774-83017774 on Build GRCh38
rs112228893 Chr.15: 83017948-83017948 on Build GRCh38
rs990245676 Chr.15: 83017688-83017688 on Build GRCh38
rs985671461 Chr.15: 83017946-83017946 on Build GRCh38
rs57498938 Chr.15: 83017674-83017674 on Build GRCh38
rs932887290 Chr.15: 83017932-83017932 on Build GRCh38
rs937311727 Chr.15: 83017791-83017791 on Build GRCh38
rs750538880 Chr.15: 83018075-83018075 on Build GRCh38
rs914862063 Chr.15: 83017908-83017908 on Build GRCh38
rs756094447 Chr.15: 83018056-83018058 on Build GRCh38
rs1048917350 Chr.15: 83017898-83017898 on Build GRCh38
rs17158127 Chr.15: 83018007-83018007 on Build GRCh38
rs571280050 Chr.15: 83017943-83017943 on Build GRCh38
rs537876949 Chr.15: 83017812-83017812 on Build GRCh38
rs981685007 Chr.15: 83017770-83017770 on Build GRCh38
rs931421883 Chr.15: 83017895-83017895 on Build GRCh38
rs913438631 Chr.15: 83017706-83017706 on Build GRCh38
rs912731751 Chr.15: 83017947-83017947 on Build GRCh38
rs990139174 Chr.15: 83017908-83017909 on Build GRCh38
rs967603152 Chr.15: 83017738-83017738 on Build GRCh38
rs1028658990 Chr.15: 83018064-83018064 on Build GRCh38
rs1012736734 Chr.15: 83017805-83017805 on Build GRCh38
rs956260060 Chr.15: 83017890-83017890 on Build GRCh38
rs529808008 Chr.15: 83017605-83017605 on Build GRCh38
rs760881146 Chr.15: 83017951-83017953 on Build GRCh38
rs1023215773 Chr.15: 83017813-83017813 on Build GRCh38
rs76064823 Chr.15: 83017667-83017667 on Build GRCh38

View Full Product Details