Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83018627-83019059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD1
Entrez Gene ID
53339
Gene Name
BTB domain containing 1
Gene Aliases
C15orf1, NS5ATP8
UniGene
Hs.459149
Gene Chromosome Location
Chr.15: 83016423-83067434 on Build GRCh38
Transcripts
NM_001011885.1 NM_025238.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3936878 c.1146C>G p.I382M Chr.15: 83018851-83018851 on Build GRCh38
4936240 c.1196G>A p.S399N Chr.15: 83018801-83018801 on Build GRCh38
4634026 c.1207A>T p.T403S Chr.15: 83018790-83018790 on Build GRCh38
1374995 c.1268A>G p.Y423C Chr.15: 83018729-83018729 on Build GRCh38
3711861 c.1201G>C p.D401H Chr.15: 83018796-83018796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947031621 Chr.15: 83018990-83018990 on Build GRCh38
rs149040830 Chr.15: 83018749-83018749 on Build GRCh38
rs770901616 Chr.15: 83018778-83018778 on Build GRCh38
rs779667774 Chr.15: 83018702-83018702 on Build GRCh38
rs59417219 Chr.15: 83018908-83018908 on Build GRCh38
rs751331989 Chr.15: 83018678-83018678 on Build GRCh38
rs1020313594 Chr.15: 83018692-83018692 on Build GRCh38
rs931875554 Chr.15: 83018933-83018933 on Build GRCh38
rs758664495 Chr.15: 83018866-83018866 on Build GRCh38
rs982792346 Chr.15: 83018902-83018902 on Build GRCh38
rs778257108 Chr.15: 83018867-83018867 on Build GRCh38
rs889859495 Chr.15: 83019006-83019011 on Build GRCh38
rs754502749 Chr.15: 83018837-83018837 on Build GRCh38
rs755549700 Chr.15: 83018697-83018697 on Build GRCh38
rs772514339 Chr.15: 83018748-83018748 on Build GRCh38
rs776915819 Chr.15: 83018799-83018799 on Build GRCh38
rs112017737 Chr.15: 83018800-83018800 on Build GRCh38
rs954046530 Chr.15: 83018684-83018684 on Build GRCh38
rs893069584 Chr.15: 83018978-83018978 on Build GRCh38
rs199578481 Chr.15: 83018690-83018690 on Build GRCh38
rs750306692 Chr.15: 83018819-83018819 on Build GRCh38
rs747597658 Chr.15: 83018769-83018769 on Build GRCh38
rs753281574 Chr.15: 83018828-83018828 on Build GRCh38
rs763976259 Chr.15: 83018673-83018673 on Build GRCh38
rs1044061991 Chr.15: 83018940-83018940 on Build GRCh38
rs760726780 Chr.15: 83018823-83018823 on Build GRCh38
rs759894123 Chr.15: 83018664-83018664 on Build GRCh38
rs763663190 Chr.15: 83018771-83018771 on Build GRCh38
rs778493119 Chr.15: 83018845-83018845 on Build GRCh38
rs1053411517 Chr.15: 83018968-83018968 on Build GRCh38
rs990653451 Chr.15: 83018997-83018997 on Build GRCh38
rs1011392406 Chr.15: 83018984-83018991 on Build GRCh38
rs765806228 Chr.15: 83018825-83018825 on Build GRCh38
rs767387329 Chr.15: 83018816-83018816 on Build GRCh38
rs908602944 Chr.15: 83018907-83018907 on Build GRCh38
rs747465729 Chr.15: 83018895-83018895 on Build GRCh38
rs754697311 Chr.15: 83018703-83018703 on Build GRCh38
rs771401246 Chr.15: 83018897-83018897 on Build GRCh38
rs778544522 Chr.15: 83018716-83018716 on Build GRCh38
rs559624330 Chr.15: 83018809-83018809 on Build GRCh38
rs908941116 Chr.15: 83018921-83018921 on Build GRCh38
rs1027381034 Chr.15: 83018779-83018779 on Build GRCh38
rs369493678 Chr.15: 83018849-83018849 on Build GRCh38
rs376123965 Chr.15: 83018746-83018746 on Build GRCh38
rs576678528 Chr.15: 83018995-83018995 on Build GRCh38
rs960574484 Chr.15: 83018989-83018989 on Build GRCh38
rs767308168 Chr.15: 83018694-83018694 on Build GRCh38
rs976284902 Chr.15: 83018910-83018910 on Build GRCh38
rs750407971 Chr.15: 83018696-83018696 on Build GRCh38
rs199871904 Chr.15: 83018963-83018966 on Build GRCh38
rs928584875 Chr.15: 83018942-83018942 on Build GRCh38
rs1050394787 Chr.15: 83018970-83018970 on Build GRCh38
rs1043188184 Chr.15: 83018991-83018991 on Build GRCh38
rs761660948 Chr.15: 83018810-83018810 on Build GRCh38
rs147308967 Chr.15: 83019006-83019007 on Build GRCh38
rs923171011 Chr.15: 83018911-83018911 on Build GRCh38
rs11557551 Chr.15: 83018732-83018732 on Build GRCh38
rs747190044 Chr.15: 83018718-83018733 on Build GRCh38
rs780156164 Chr.15: 83018693-83018694 on Build GRCh38
rs973350600 Chr.15: 83018761-83018761 on Build GRCh38
rs948419681 Chr.15: 83018937-83018937 on Build GRCh38
rs765497456 Chr.15: 83018808-83018808 on Build GRCh38

View Full Product Details