Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39329369-39329780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMFG
Entrez Gene ID
9535
Gene Name
glia maturation factor gamma
Gene Aliases
GMF-GAMMA
UniGene
Hs.5210
Gene Chromosome Location
Chr.19: 39328357-39336086 on Build GRCh38
Transcripts
NM_001301008.1 NM_004877.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1731042 c.253C>A p.P85T Chr.19: 39329574-39329574 on Build GRCh38
250965 c.283+1G>C p.? Chr.19: 39329543-39329543 on Build GRCh38
1630918 c.242G>C p.R81P Chr.19: 39329585-39329585 on Build GRCh38
3533685 c.204C>T p.F68F Chr.19: 39329623-39329623 on Build GRCh38
148618 c.225C>T p.Y75Y Chr.19: 39329602-39329602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761084892 Chr.19: 39329575-39329575 on Build GRCh38
rs752963982 Chr.19: 39329658-39329658 on Build GRCh38
rs115230602 Chr.19: 39329593-39329593 on Build GRCh38
rs542194581 Chr.19: 39329392-39329392 on Build GRCh38
rs560035206 Chr.19: 39329680-39329680 on Build GRCh38
rs779631043 Chr.19: 39329675-39329675 on Build GRCh38
rs1038778798 Chr.19: 39329489-39329489 on Build GRCh38
rs914066819 Chr.19: 39329410-39329410 on Build GRCh38
rs1039098701 Chr.19: 39329549-39329549 on Build GRCh38
rs563848194 Chr.19: 39329473-39329473 on Build GRCh38
rs866359378 Chr.19: 39329400-39329400 on Build GRCh38
rs771921863 Chr.19: 39329501-39329501 on Build GRCh38
rs778824916 Chr.19: 39329497-39329497 on Build GRCh38
rs369802035 Chr.19: 39329513-39329513 on Build GRCh38
rs370207146 Chr.19: 39329623-39329623 on Build GRCh38
rs750946002 Chr.19: 39329487-39329488 on Build GRCh38
rs143496592 Chr.19: 39329581-39329581 on Build GRCh38
rs759883574 Chr.19: 39329651-39329651 on Build GRCh38
rs568232274 Chr.19: 39329631-39329631 on Build GRCh38
rs564231096 Chr.19: 39329579-39329579 on Build GRCh38
rs751590062 Chr.19: 39329662-39329662 on Build GRCh38
rs62119672 Chr.19: 39329396-39329396 on Build GRCh38
rs990106930 Chr.19: 39329687-39329687 on Build GRCh38
rs766705633 Chr.19: 39329576-39329576 on Build GRCh38
rs181597528 Chr.19: 39329668-39329668 on Build GRCh38
rs530473219 Chr.19: 39329431-39329431 on Build GRCh38
rs762476934 Chr.19: 39329557-39329557 on Build GRCh38
rs746808384 Chr.19: 39329506-39329506 on Build GRCh38
rs899104087 Chr.19: 39329552-39329552 on Build GRCh38
rs373751976 Chr.19: 39329679-39329680 on Build GRCh38
rs1053997603 Chr.19: 39329397-39329397 on Build GRCh38
rs757294166 Chr.19: 39329665-39329665 on Build GRCh38
rs769388612 Chr.19: 39329535-39329535 on Build GRCh38
rs375323802 Chr.19: 39329505-39329505 on Build GRCh38
rs559049849 Chr.19: 39329733-39329733 on Build GRCh38
rs945524937 Chr.19: 39329734-39329734 on Build GRCh38
rs1051421391 Chr.19: 39329484-39329484 on Build GRCh38
rs779145008 Chr.19: 39329585-39329585 on Build GRCh38
rs758468716 Chr.19: 39329660-39329660 on Build GRCh38
rs753890668 Chr.19: 39329640-39329640 on Build GRCh38
rs201400008 Chr.19: 39329645-39329645 on Build GRCh38
rs745672660 Chr.19: 39329669-39329669 on Build GRCh38
rs767923581 Chr.19: 39329560-39329560 on Build GRCh38
rs775150813 Chr.19: 39329614-39329614 on Build GRCh38
rs750925189 Chr.19: 39329574-39329574 on Build GRCh38
rs1001574921 Chr.19: 39329421-39329421 on Build GRCh38
rs200517926 Chr.19: 39329653-39329653 on Build GRCh38
rs111976645 Chr.19: 39329438-39329438 on Build GRCh38
rs777980724 Chr.19: 39329661-39329661 on Build GRCh38
rs770900962 Chr.19: 39329600-39329600 on Build GRCh38
rs759244014 Chr.19: 39329527-39329527 on Build GRCh38
rs761138010 Chr.19: 39329632-39329632 on Build GRCh38
rs898613539 Chr.19: 39329511-39329511 on Build GRCh38
rs769354620 Chr.19: 39329613-39329613 on Build GRCh38
rs945138945 Chr.19: 39329481-39329481 on Build GRCh38
rs537482567 Chr.19: 39329698-39329698 on Build GRCh38
rs1007198850 Chr.19: 39329548-39329548 on Build GRCh38
rs1051473982 Chr.19: 39329738-39329738 on Build GRCh38
rs551849740 Chr.19: 39329462-39329462 on Build GRCh38
rs926615654 Chr.19: 39329649-39329649 on Build GRCh38
rs766828327 Chr.19: 39329638-39329638 on Build GRCh38
rs75976955 Chr.19: 39329602-39329602 on Build GRCh38
rs755183533 Chr.19: 39329584-39329584 on Build GRCh38
rs748216740 Chr.19: 39329499-39329499 on Build GRCh38
rs372922067 Chr.19: 39329541-39329541 on Build GRCh38
rs752728731 Chr.19: 39329586-39329586 on Build GRCh38
rs777642468 Chr.19: 39329592-39329592 on Build GRCh38

View Full Product Details