Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29608513-29609026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABBR1
Entrez Gene ID
2550
Gene Name
gamma-aminobutyric acid type B receptor subunit 1
Gene Aliases
GABABR1, GABBR1-3, GB1, GPRC3A
UniGene
Hs.167017
Gene Chromosome Location
Chr.6: 29602228-29633135 on Build GRCh38
Transcripts
NM_001319053.1 NM_001470.3 NM_021903.2 NM_021904.3 XM_005248982.2 XM_006715047.3 XM_011514453.2 XM_011514455.1 XM_017010676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4941663 c.1980T>C p.P660P Chr.6: 29608613-29608613 on Build GRCh38
6668428 c.1597G>A p.D533N Chr.6: 29608645-29608645 on Build GRCh38
3861289 c.1867C>A p.Q623K Chr.6: 29608726-29608726 on Build GRCh38
184857 c.1983C>T p.F661F Chr.6: 29608610-29608610 on Build GRCh38
6106415 c.1632C>T p.F544F Chr.6: 29608610-29608610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899652820 Chr.6: 29608947-29608947 on Build GRCh38
rs764151332 Chr.6: 29608701-29608701 on Build GRCh38
rs1015647192 Chr.6: 29608730-29608730 on Build GRCh38
rs1041255022 Chr.6: 29608903-29608903 on Build GRCh38
rs758422439 Chr.6: 29608588-29608588 on Build GRCh38
rs754756242 Chr.6: 29608752-29608752 on Build GRCh38
rs774307185 Chr.6: 29608622-29608622 on Build GRCh38
rs765811910 Chr.6: 29608567-29608567 on Build GRCh38
rs755369306 Chr.6: 29608773-29608774 on Build GRCh38
rs758908025 Chr.6: 29608699-29608699 on Build GRCh38
rs577865940 Chr.6: 29608890-29608890 on Build GRCh38
rs370044776 Chr.6: 29608784-29608784 on Build GRCh38
rs757129884 Chr.6: 29608973-29608973 on Build GRCh38
rs777840382 Chr.6: 29608595-29608595 on Build GRCh38
rs926779474 Chr.6: 29608794-29608794 on Build GRCh38
rs773355069 Chr.6: 29608924-29608924 on Build GRCh38
rs952697403 Chr.6: 29608690-29608690 on Build GRCh38
rs753174125 Chr.6: 29608574-29608574 on Build GRCh38
rs932596094 Chr.6: 29608952-29608952 on Build GRCh38
rs535450853 Chr.6: 29608774-29608774 on Build GRCh38
rs2076489 Chr.6: 29608660-29608660 on Build GRCh38
rs750453732 Chr.6: 29608697-29608697 on Build GRCh38
rs902907696 Chr.6: 29608569-29608569 on Build GRCh38
rs528175691 Chr.6: 29608555-29608555 on Build GRCh38
rs573856731 Chr.6: 29608953-29608953 on Build GRCh38
rs570026645 Chr.6: 29608666-29608666 on Build GRCh38
rs1047757131 Chr.6: 29608946-29608946 on Build GRCh38
rs750646029 Chr.6: 29608750-29608750 on Build GRCh38
rs772853503 Chr.6: 29608654-29608654 on Build GRCh38
rs113339079 Chr.6: 29608814-29608814 on Build GRCh38
rs760417835 Chr.6: 29608557-29608557 on Build GRCh38
rs938221156 Chr.6: 29608541-29608541 on Build GRCh38
rs777680256 Chr.6: 29608930-29608930 on Build GRCh38
rs3025635 Chr.6: 29608548-29608548 on Build GRCh38
rs754295052 Chr.6: 29608559-29608559 on Build GRCh38
rs558331306 Chr.6: 29608816-29608816 on Build GRCh38
rs144688306 Chr.6: 29608630-29608630 on Build GRCh38
rs751692575 Chr.6: 29608734-29608734 on Build GRCh38
rs113020133 Chr.6: 29608814-29608815 on Build GRCh38
rs757538966 Chr.6: 29608610-29608610 on Build GRCh38
rs29263 Chr.6: 29608813-29608813 on Build GRCh38
rs763157649 Chr.6: 29608777-29608777 on Build GRCh38
rs375396008 Chr.6: 29608578-29608578 on Build GRCh38
rs111603715 Chr.6: 29608810-29608811 on Build GRCh38
rs17218739 Chr.6: 29608563-29608563 on Build GRCh38
rs977051164 Chr.6: 29608851-29608851 on Build GRCh38
rs924387964 Chr.6: 29608900-29608900 on Build GRCh38
rs1023218099 Chr.6: 29608960-29608960 on Build GRCh38
rs386698389 Chr.6: 29608813-29608815 on Build GRCh38
rs543651774 Chr.6: 29608904-29608904 on Build GRCh38
rs779705251 Chr.6: 29608615-29608615 on Build GRCh38
rs76632246 Chr.6: 29608655-29608655 on Build GRCh38
rs185645531 Chr.6: 29608965-29608965 on Build GRCh38
rs765540037 Chr.6: 29608679-29608679 on Build GRCh38
rs757452736 Chr.6: 29608737-29608737 on Build GRCh38
rs143545412 Chr.6: 29608558-29608558 on Build GRCh38
rs29230 Chr.6: 29608616-29608616 on Build GRCh38
rs781520426 Chr.6: 29608748-29608748 on Build GRCh38
rs111899366 Chr.6: 29608790-29608790 on Build GRCh38
rs138809665 Chr.6: 29608609-29608609 on Build GRCh38
rs146033694 Chr.6: 29608910-29608910 on Build GRCh38
rs368884104 Chr.6: 29608553-29608553 on Build GRCh38
rs778587866 Chr.6: 29608758-29608758 on Build GRCh38
rs771651154 Chr.6: 29608646-29608646 on Build GRCh38
rs888151010 Chr.6: 29608949-29608949 on Build GRCh38
rs372019251 Chr.6: 29608552-29608552 on Build GRCh38
rs555708553 Chr.6: 29608812-29608812 on Build GRCh38
rs29262 Chr.6: 29608815-29608815 on Build GRCh38

View Full Product Details