Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137879064-137879611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L2
Entrez Gene ID
64764
Gene Name
cAMP responsive element binding protein 3 like 2
Gene Aliases
BBF2H7
UniGene
Hs.490273
Gene Chromosome Location
Chr.7: 137874979-138002101 on Build GRCh38
Transcripts
NM_001318246.1 NM_194071.3 XM_005250541.4 XM_017012540.1 XM_017012541.1

SNP Detail

SNP ID Chromosome Location
rs768898825 Chr.7: 137879226-137879226 on Build GRCh38
rs749600344 Chr.7: 137879204-137879204 on Build GRCh38
rs559381642 Chr.7: 137879179-137879180 on Build GRCh38
rs927111709 Chr.7: 137879476-137879476 on Build GRCh38
rs77988292 Chr.7: 137879429-137879429 on Build GRCh38
rs757006314 Chr.7: 137879169-137879169 on Build GRCh38
rs532139233 Chr.7: 137879168-137879169 on Build GRCh38
rs940675256 Chr.7: 137879122-137879122 on Build GRCh38
rs908122847 Chr.7: 137879346-137879346 on Build GRCh38
rs891420794 Chr.7: 137879229-137879229 on Build GRCh38
rs569747282 Chr.7: 137879326-137879326 on Build GRCh38
rs975134401 Chr.7: 137879228-137879228 on Build GRCh38
rs1021276281 Chr.7: 137879270-137879270 on Build GRCh38
rs569765755 Chr.7: 137879493-137879493 on Build GRCh38
rs917278393 Chr.7: 137879420-137879420 on Build GRCh38
rs549806191 Chr.7: 137879467-137879467 on Build GRCh38
rs576460127 Chr.7: 137879370-137879370 on Build GRCh38
rs199843739 Chr.7: 137879179-137879179 on Build GRCh38
rs371123233 Chr.7: 137879179-137879179 on Build GRCh38
rs564395497 Chr.7: 137879456-137879456 on Build GRCh38
rs1045569739 Chr.7: 137879474-137879474 on Build GRCh38
rs769075208 Chr.7: 137879296-137879296 on Build GRCh38
rs955063221 Chr.7: 137879524-137879524 on Build GRCh38
rs201156794 Chr.7: 137879178-137879179 on Build GRCh38
rs962323788 Chr.7: 137879382-137879382 on Build GRCh38
rs775815959 Chr.7: 137879142-137879142 on Build GRCh38
rs564972163 Chr.7: 137879418-137879418 on Build GRCh38
rs534260231 Chr.7: 137879176-137879176 on Build GRCh38
rs926449544 Chr.7: 137879343-137879343 on Build GRCh38
rs755101178 Chr.7: 137879285-137879285 on Build GRCh38
rs903439221 Chr.7: 137879448-137879448 on Build GRCh38
rs895799462 Chr.7: 137879107-137879107 on Build GRCh38
rs542631193 Chr.7: 137879311-137879311 on Build GRCh38
rs527566036 Chr.7: 137879421-137879421 on Build GRCh38
rs962618809 Chr.7: 137879318-137879318 on Build GRCh38
rs562599361 Chr.7: 137879361-137879361 on Build GRCh38
rs1037745642 Chr.7: 137879123-137879123 on Build GRCh38
rs547501145 Chr.7: 137879116-137879116 on Build GRCh38
rs547836757 Chr.7: 137879208-137879208 on Build GRCh38
rs1035576371 Chr.7: 137879210-137879210 on Build GRCh38
rs372097757 Chr.7: 137879180-137879181 on Build GRCh38
rs1010134665 Chr.7: 137879576-137879576 on Build GRCh38
rs1055788750 Chr.7: 137879413-137879413 on Build GRCh38
rs891705407 Chr.7: 137879550-137879550 on Build GRCh38
rs1002764536 Chr.7: 137879180-137879180 on Build GRCh38
rs578190505 Chr.7: 137879137-137879137 on Build GRCh38
rs561187565 Chr.7: 137879440-137879440 on Build GRCh38
rs777132283 Chr.7: 137879317-137879317 on Build GRCh38
rs1029409320 Chr.7: 137879517-137879517 on Build GRCh38
rs529678747 Chr.7: 137879466-137879466 on Build GRCh38
rs115510765 Chr.7: 137879248-137879248 on Build GRCh38
rs755717824 Chr.7: 137879157-137879157 on Build GRCh38
rs1004288753 Chr.7: 137879543-137879543 on Build GRCh38
rs1009854445 Chr.7: 137879260-137879260 on Build GRCh38
rs373410712 Chr.7: 137879393-137879393 on Build GRCh38
rs937446353 Chr.7: 137879359-137879359 on Build GRCh38
rs1000771291 Chr.7: 137879189-137879189 on Build GRCh38
rs955011251 Chr.7: 137879310-137879310 on Build GRCh38
rs996625618 Chr.7: 137879498-137879498 on Build GRCh38
rs758919104 Chr.7: 137879512-137879512 on Build GRCh38
rs376927156 Chr.7: 137879522-137879522 on Build GRCh38
rs903386660 Chr.7: 137879178-137879178 on Build GRCh38
rs148937028 Chr.7: 137879334-137879334 on Build GRCh38
rs780413554 Chr.7: 137879186-137879186 on Build GRCh38

View Full Product Details