Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 61299599-61300102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLVS1
Entrez Gene ID
157807
Gene Name
clavesin 1
Gene Aliases
CRALBPL, RLBP1L1
UniGene
Hs.591874
Gene Chromosome Location
Chr.8: 61057158-61501645 on Build GRCh38
Transcripts
NM_173519.2 XM_017013141.1 XM_017013142.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1729946 c.175A>T p.M59L Chr.8: 61300002-61300002 on Build GRCh38
6113613 c.183C>G p.I61M Chr.8: 61300010-61300010 on Build GRCh38
4151538 c.209G>A p.R70H Chr.8: 61300036-61300036 on Build GRCh38
5358810 c.212C>T p.T71I Chr.8: 61300039-61300039 on Build GRCh38
4904782 c.136C>T p.P46S Chr.8: 61299963-61299963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002273135 Chr.8: 61299766-61299766 on Build GRCh38
rs765680966 Chr.8: 61299926-61299926 on Build GRCh38
rs375556372 Chr.8: 61299991-61299991 on Build GRCh38
rs187032551 Chr.8: 61299943-61299943 on Build GRCh38
rs766850570 Chr.8: 61300036-61300036 on Build GRCh38
rs1052571834 Chr.8: 61299685-61299685 on Build GRCh38
rs866957503 Chr.8: 61300076-61300076 on Build GRCh38
rs373908809 Chr.8: 61299942-61299942 on Build GRCh38
rs370764198 Chr.8: 61299931-61299931 on Build GRCh38
rs759006982 Chr.8: 61299930-61299930 on Build GRCh38
rs201184358 Chr.8: 61299797-61299797 on Build GRCh38
rs769664828 Chr.8: 61299870-61299870 on Build GRCh38
rs764407581 Chr.8: 61299780-61299780 on Build GRCh38
rs761065879 Chr.8: 61299759-61299759 on Build GRCh38
rs776829272 Chr.8: 61299982-61299982 on Build GRCh38
rs749033608 Chr.8: 61299965-61299965 on Build GRCh38
rs766720972 Chr.8: 61300023-61300023 on Build GRCh38
rs114990777 Chr.8: 61299767-61299767 on Build GRCh38
rs777295607 Chr.8: 61300080-61300080 on Build GRCh38
rs548652090 Chr.8: 61299683-61299683 on Build GRCh38
rs202247798 Chr.8: 61299849-61299849 on Build GRCh38
rs779670056 Chr.8: 61299832-61299832 on Build GRCh38
rs751609341 Chr.8: 61299864-61299871 on Build GRCh38
rs762182128 Chr.8: 61300002-61300002 on Build GRCh38
rs763236302 Chr.8: 61300018-61300018 on Build GRCh38
rs866356412 Chr.8: 61299865-61299865 on Build GRCh38
rs755864413 Chr.8: 61300069-61300069 on Build GRCh38
rs368485692 Chr.8: 61300035-61300035 on Build GRCh38
rs750795149 Chr.8: 61299878-61299878 on Build GRCh38
rs981496938 Chr.8: 61299962-61299962 on Build GRCh38
rs953445501 Chr.8: 61299951-61299951 on Build GRCh38
rs146100118 Chr.8: 61299944-61299944 on Build GRCh38
rs374267023 Chr.8: 61299818-61299818 on Build GRCh38
rs766968154 Chr.8: 61299925-61299925 on Build GRCh38
rs774940994 Chr.8: 61299629-61299629 on Build GRCh38
rs757717709 Chr.8: 61299800-61299800 on Build GRCh38
rs780840731 Chr.8: 61299852-61299852 on Build GRCh38
rs867676093 Chr.8: 61299841-61299841 on Build GRCh38
rs928298993 Chr.8: 61299967-61299967 on Build GRCh38
rs765369429 Chr.8: 61300013-61300013 on Build GRCh38
rs762094349 Chr.8: 61299918-61299918 on Build GRCh38
rs758807314 Chr.8: 61299824-61299824 on Build GRCh38
rs912154891 Chr.8: 61299998-61299998 on Build GRCh38
rs912128306 Chr.8: 61299643-61299643 on Build GRCh38
rs1030443283 Chr.8: 61299762-61299762 on Build GRCh38
rs754046598 Chr.8: 61299791-61299791 on Build GRCh38
rs914410607 Chr.8: 61300056-61300056 on Build GRCh38
rs377639699 Chr.8: 61299891-61299891 on Build GRCh38
rs538889155 Chr.8: 61299936-61299936 on Build GRCh38
rs756476226 Chr.8: 61300015-61300015 on Build GRCh38
rs267601962 Chr.8: 61300055-61300055 on Build GRCh38
rs747753203 Chr.8: 61299854-61299854 on Build GRCh38
rs370102890 Chr.8: 61299974-61299974 on Build GRCh38
rs752095864 Chr.8: 61300024-61300024 on Build GRCh38
rs535901986 Chr.8: 61299969-61299969 on Build GRCh38
rs71525448 Chr.8: 61299770-61299770 on Build GRCh38
rs765478333 Chr.8: 61299801-61299801 on Build GRCh38
rs986531054 Chr.8: 61299989-61299989 on Build GRCh38
rs754414771 Chr.8: 61299840-61299840 on Build GRCh38
rs1043481920 Chr.8: 61299754-61299754 on Build GRCh38
rs904880179 Chr.8: 61299758-61299758 on Build GRCh38
rs1010631363 Chr.8: 61299746-61299746 on Build GRCh38
rs28564572 Chr.8: 61299805-61299805 on Build GRCh38
rs374411661 Chr.8: 61300053-61300053 on Build GRCh38
rs764221332 Chr.8: 61299905-61299905 on Build GRCh38
rs770693777 Chr.8: 61299966-61299966 on Build GRCh38
rs763716458 Chr.8: 61299734-61299734 on Build GRCh38
rs770762643 Chr.8: 61299879-61299879 on Build GRCh38
rs182742681 Chr.8: 61299845-61299845 on Build GRCh38
rs746514753 Chr.8: 61299839-61299839 on Build GRCh38
rs750738056 Chr.8: 61299928-61299928 on Build GRCh38
rs777024134 Chr.8: 61299917-61299917 on Build GRCh38
rs973490347 Chr.8: 61299921-61299921 on Build GRCh38
rs768896897 Chr.8: 61299981-61299981 on Build GRCh38
rs1041617475 Chr.8: 61299705-61299705 on Build GRCh38
rs377032110 Chr.8: 61299946-61299946 on Build GRCh38
rs116925923 Chr.8: 61299717-61299717 on Build GRCh38
rs773413383 Chr.8: 61300017-61300017 on Build GRCh38

View Full Product Details