Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 21910434-21910955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK2
Entrez Gene ID
9046
Gene Name
docking protein 2
Gene Aliases
p56DOK, p56dok-2
UniGene
Hs.71215
Gene Chromosome Location
Chr.8: 21908873-21913717 on Build GRCh38
Transcripts
NM_001317800.1 NM_003974.3 NM_201349.2 XM_005273680.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
226192 c.577G>A p.D193N Chr.8: 21910714-21910714 on Build GRCh38
5663869 c.586T>A p.Y196N Chr.8: 21910705-21910705 on Build GRCh38
5725239 c.448G>A p.E150K Chr.8: 21910843-21910843 on Build GRCh38
4548443 c.447G>A p.K149K Chr.8: 21910844-21910844 on Build GRCh38
6495210 c.434-2A>G p.? Chr.8: 21910859-21910859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761651238 Chr.8: 21910724-21910724 on Build GRCh38
rs534191632 Chr.8: 21910881-21910881 on Build GRCh38
rs199722634 Chr.8: 21910702-21910702 on Build GRCh38
rs766322178 Chr.8: 21910741-21910741 on Build GRCh38
rs550676966 Chr.8: 21910706-21910706 on Build GRCh38
rs142797319 Chr.8: 21910870-21910873 on Build GRCh38
rs754293546 Chr.8: 21910868-21910868 on Build GRCh38
rs201260853 Chr.8: 21910883-21910883 on Build GRCh38
rs776341045 Chr.8: 21910718-21910718 on Build GRCh38
rs533626602 Chr.8: 21910871-21910871 on Build GRCh38
rs546586223 Chr.8: 21910690-21910690 on Build GRCh38
rs747799833 Chr.8: 21910816-21910816 on Build GRCh38
rs965226141 Chr.8: 21910483-21910483 on Build GRCh38
rs371052882 Chr.8: 21910788-21910788 on Build GRCh38
rs749142572 Chr.8: 21910900-21910900 on Build GRCh38
rs780508425 Chr.8: 21910795-21910795 on Build GRCh38
rs1001569538 Chr.8: 21910470-21910470 on Build GRCh38
rs777403513 Chr.8: 21910890-21910890 on Build GRCh38
rs989563939 Chr.8: 21910503-21910503 on Build GRCh38
rs149955790 Chr.8: 21910736-21910736 on Build GRCh38
rs577282664 Chr.8: 21910475-21910475 on Build GRCh38
rs771236641 Chr.8: 21910736-21910736 on Build GRCh38
rs767640407 Chr.8: 21910856-21910856 on Build GRCh38
rs945414596 Chr.8: 21910789-21910789 on Build GRCh38
rs764169515 Chr.8: 21910579-21910579 on Build GRCh38
rs748525720 Chr.8: 21910624-21910624 on Build GRCh38
rs750375145 Chr.8: 21910758-21910758 on Build GRCh38
rs79048610 Chr.8: 21910685-21910685 on Build GRCh38
rs758760513 Chr.8: 21910882-21910882 on Build GRCh38
rs575570215 Chr.8: 21910921-21910921 on Build GRCh38
rs746566350 Chr.8: 21910790-21910790 on Build GRCh38
rs756933437 Chr.8: 21910680-21910680 on Build GRCh38
rs577146088 Chr.8: 21910889-21910889 on Build GRCh38
rs766207394 Chr.8: 21910878-21910879 on Build GRCh38
rs750322402 Chr.8: 21910871-21910873 on Build GRCh38
rs371068266 Chr.8: 21910761-21910761 on Build GRCh38
rs199970520 Chr.8: 21910464-21910464 on Build GRCh38
rs1004338135 Chr.8: 21910898-21910898 on Build GRCh38
rs756338298 Chr.8: 21910775-21910775 on Build GRCh38
rs34417983 Chr.8: 21910638-21910638 on Build GRCh38
rs759334784 Chr.8: 21910852-21910852 on Build GRCh38
rs766447432 Chr.8: 21910681-21910681 on Build GRCh38
rs769930603 Chr.8: 21910849-21910849 on Build GRCh38
rs149146251 Chr.8: 21910569-21910569 on Build GRCh38
rs569510103 Chr.8: 21910715-21910715 on Build GRCh38
rs936518071 Chr.8: 21910497-21910497 on Build GRCh38
rs759659439 Chr.8: 21910771-21910771 on Build GRCh38
rs1019487244 Chr.8: 21910481-21910481 on Build GRCh38
rs549675779 Chr.8: 21910836-21910836 on Build GRCh38
rs578115923 Chr.8: 21910486-21910486 on Build GRCh38
rs779451507 Chr.8: 21910708-21910708 on Build GRCh38
rs1026726697 Chr.8: 21910693-21910693 on Build GRCh38
rs778227189 Chr.8: 21910699-21910699 on Build GRCh38
rs925936873 Chr.8: 21910841-21910841 on Build GRCh38
rs776853541 Chr.8: 21910836-21910848 on Build GRCh38
rs763753741 Chr.8: 21910649-21910649 on Build GRCh38
rs575081316 Chr.8: 21910578-21910578 on Build GRCh38
rs775421537 Chr.8: 21910864-21910866 on Build GRCh38
rs985391641 Chr.8: 21910657-21910657 on Build GRCh38
rs996498855 Chr.8: 21910913-21910913 on Build GRCh38
rs773043465 Chr.8: 21910822-21910822 on Build GRCh38
rs769392516 Chr.8: 21910818-21910818 on Build GRCh38
rs769587613 Chr.8: 21910731-21910731 on Build GRCh38
rs771722273 Chr.8: 21910636-21910636 on Build GRCh38
rs759689010 Chr.8: 21910845-21910845 on Build GRCh38
rs1140295 Chr.8: 21910837-21910837 on Build GRCh38
rs768196912 Chr.8: 21910792-21910792 on Build GRCh38
rs1027173902 Chr.8: 21910457-21910457 on Build GRCh38
rs979335869 Chr.8: 21910813-21910813 on Build GRCh38
rs549851531 Chr.8: 21910530-21910530 on Build GRCh38
rs958236474 Chr.8: 21910927-21910927 on Build GRCh38
rs764387277 Chr.8: 21910867-21910867 on Build GRCh38
rs774335789 Chr.8: 21910849-21910849 on Build GRCh38
rs746506119 Chr.8: 21910714-21910714 on Build GRCh38
rs886615847 Chr.8: 21910873-21910873 on Build GRCh38
rs558636070 Chr.8: 21910474-21910474 on Build GRCh38
rs953387403 Chr.8: 21910793-21910793 on Build GRCh38
rs776337770 Chr.8: 21910651-21910651 on Build GRCh38
rs761637146 Chr.8: 21910661-21910661 on Build GRCh38
rs542553428 Chr.8: 21910582-21910582 on Build GRCh38
rs757691082 Chr.8: 21910869-21910869 on Build GRCh38
rs149080191 Chr.8: 21910859-21910859 on Build GRCh38
rs767837190 Chr.8: 21910516-21910516 on Build GRCh38
rs542580793 Chr.8: 21910924-21910924 on Build GRCh38
rs374967168 Chr.8: 21910625-21910625 on Build GRCh38
rs1019339086 Chr.8: 21910905-21910905 on Build GRCh38
rs774999392 Chr.8: 21910640-21910640 on Build GRCh38
rs752787121 Chr.8: 21910857-21910857 on Build GRCh38
rs376016280 Chr.8: 21910886-21910886 on Build GRCh38
rs762707505 Chr.8: 21910824-21910824 on Build GRCh38
rs899449022 Chr.8: 21910858-21910858 on Build GRCh38
rs1055371058 Chr.8: 21910850-21910850 on Build GRCh38
rs756172563 Chr.8: 21910806-21910806 on Build GRCh38
rs377500664 Chr.8: 21910773-21910773 on Build GRCh38
rs376128734 Chr.8: 21910660-21910660 on Build GRCh38
rs931919114 Chr.8: 21910632-21910632 on Build GRCh38
rs544349080 Chr.8: 21910494-21910494 on Build GRCh38
rs760556468 Chr.8: 21910871-21910877 on Build GRCh38
rs982200824 Chr.8: 21910791-21910791 on Build GRCh38
rs752857479 Chr.8: 21910644-21910644 on Build GRCh38
rs372929533 Chr.8: 21910735-21910735 on Build GRCh38
rs1049372467 Chr.8: 21910902-21910902 on Build GRCh38
rs143602195 Chr.8: 21910909-21910909 on Build GRCh38
rs770844622 Chr.8: 21910828-21910828 on Build GRCh38
rs951065725 Chr.8: 21910466-21910466 on Build GRCh38
rs569587738 Chr.8: 21910774-21910774 on Build GRCh38
rs754205816 Chr.8: 21910787-21910787 on Build GRCh38
rs773852020 Chr.8: 21910629-21910629 on Build GRCh38
rs145022595 Chr.8: 21910757-21910757 on Build GRCh38
rs765221775 Chr.8: 21910666-21910666 on Build GRCh38
rs966298305 Chr.8: 21910523-21910523 on Build GRCh38
rs200755956 Chr.8: 21910692-21910692 on Build GRCh38
rs761864096 Chr.8: 21910868-21910868 on Build GRCh38
rs751632704 Chr.8: 21910750-21910750 on Build GRCh38
rs113214277 Chr.8: 21910689-21910689 on Build GRCh38
rs778314744 Chr.8: 21910785-21910785 on Build GRCh38

View Full Product Details