Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92618371-92618881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPPK
Entrez Gene ID
64768
Gene Name
inositol-pentakisphosphate 2-kinase
Gene Aliases
C9orf12, INSP5K2, IP5K, IPK1, bA476B13.1
UniGene
Hs.459896
Gene Chromosome Location
Chr.9: 92613184-92670265 on Build GRCh38
Transcripts
NM_022755.5 XM_017015041.1 NR_036505.1

Gene Symbol
LOC100128361
Entrez Gene ID
100128361
Gene Name
uncharacterized LOC100128361
Gene Aliases
-
UniGene
Hs.660395
Gene Chromosome Location
Chr.9: 92618050-92620533 on Build GRCh38
Transcripts
NM_022755.5 XM_017015041.1 NR_036505.1

SNP Detail

SNP ID Chromosome Location
rs544631120 Chr.9: 92618450-92618450 on Build GRCh38
rs774354132 Chr.9: 92618835-92618835 on Build GRCh38
rs891631368 Chr.9: 92618515-92618515 on Build GRCh38
rs766877347 Chr.9: 92618521-92618521 on Build GRCh38
rs905620241 Chr.9: 92618809-92618809 on Build GRCh38
rs753859759 Chr.9: 92618535-92618535 on Build GRCh38
rs542520439 Chr.9: 92618519-92618519 on Build GRCh38
rs922057353 Chr.9: 92618617-92618617 on Build GRCh38
rs887644091 Chr.9: 92618558-92618558 on Build GRCh38
rs753037112 Chr.9: 92618490-92618490 on Build GRCh38
rs941875019 Chr.9: 92618747-92618747 on Build GRCh38
rs1039255005 Chr.9: 92618749-92618749 on Build GRCh38
rs931981523 Chr.9: 92618446-92618446 on Build GRCh38
rs532632490 Chr.9: 92618573-92618573 on Build GRCh38
rs952302586 Chr.9: 92618570-92618570 on Build GRCh38
rs902215153 Chr.9: 92618851-92618851 on Build GRCh38
rs187647302 Chr.9: 92618575-92618575 on Build GRCh38
rs781488367 Chr.9: 92618460-92618460 on Build GRCh38
rs757996413 Chr.9: 92618547-92618558 on Build GRCh38
rs769265408 Chr.9: 92618603-92618603 on Build GRCh38
rs543596354 Chr.9: 92618556-92618556 on Build GRCh38
rs780607396 Chr.9: 92618742-92618743 on Build GRCh38
rs537279880 Chr.9: 92618788-92618788 on Build GRCh38
rs957595054 Chr.9: 92618594-92618594 on Build GRCh38
rs779134458 Chr.9: 92618559-92618559 on Build GRCh38
rs3839884 Chr.9: 92618743-92618743 on Build GRCh38
rs981422887 Chr.9: 92618686-92618686 on Build GRCh38
rs953556479 Chr.9: 92618646-92618646 on Build GRCh38
rs1054584649 Chr.9: 92618452-92618452 on Build GRCh38
rs758775107 Chr.9: 92618566-92618566 on Build GRCh38
rs924832401 Chr.9: 92618443-92618443 on Build GRCh38
rs570319503 Chr.9: 92618775-92618775 on Build GRCh38
rs993347874 Chr.9: 92618545-92618545 on Build GRCh38
rs976594901 Chr.9: 92618610-92618610 on Build GRCh38
rs949933733 Chr.9: 92618782-92618782 on Build GRCh38
rs147005093 Chr.9: 92618457-92618457 on Build GRCh38
rs990690365 Chr.9: 92618626-92618626 on Build GRCh38
rs772797476 Chr.9: 92618622-92618622 on Build GRCh38
rs73526054 Chr.9: 92618572-92618572 on Build GRCh38
rs777908768 Chr.9: 92618534-92618534 on Build GRCh38
rs576336638 Chr.9: 92618694-92618694 on Build GRCh38
rs193278744 Chr.9: 92618401-92618401 on Build GRCh38
rs184462941 Chr.9: 92618535-92618535 on Build GRCh38
rs561186895 Chr.9: 92618528-92618528 on Build GRCh38
rs762519861 Chr.9: 92618639-92618639 on Build GRCh38
rs2148537 Chr.9: 92618444-92618444 on Build GRCh38
rs770840558 Chr.9: 92618709-92618709 on Build GRCh38
rs937048038 Chr.9: 92618816-92618816 on Build GRCh38
rs966139798 Chr.9: 92618430-92618430 on Build GRCh38
rs997201050 Chr.9: 92618568-92618568 on Build GRCh38
rs746151975 Chr.9: 92618560-92618560 on Build GRCh38
rs548902833 Chr.9: 92618763-92618763 on Build GRCh38
rs563417405 Chr.9: 92618842-92618843 on Build GRCh38
rs530076492 Chr.9: 92618710-92618710 on Build GRCh38
rs1052943371 Chr.9: 92618477-92618477 on Build GRCh38
rs1045946021 Chr.9: 92618787-92618787 on Build GRCh38
rs772648352 Chr.9: 92618672-92618672 on Build GRCh38
rs747826823 Chr.9: 92618598-92618598 on Build GRCh38
rs571671405 Chr.9: 92618507-92618507 on Build GRCh38
rs77439188 Chr.9: 92618590-92618590 on Build GRCh38
rs935942959 Chr.9: 92618717-92618717 on Build GRCh38
rs921954578 Chr.9: 92618445-92618445 on Build GRCh38
rs948731946 Chr.9: 92618458-92618458 on Build GRCh38
rs905587689 Chr.9: 92618532-92618532 on Build GRCh38
rs997290937 Chr.9: 92618811-92618811 on Build GRCh38
rs1030074141 Chr.9: 92618844-92618844 on Build GRCh38
rs374257924 Chr.9: 92618529-92618529 on Build GRCh38
rs915999387 Chr.9: 92618640-92618640 on Build GRCh38
rs908013440 Chr.9: 92618429-92618429 on Build GRCh38
rs139089248 Chr.9: 92618745-92618759 on Build GRCh38
rs749235651 Chr.9: 92618704-92618704 on Build GRCh38
rs113296921 Chr.9: 92618595-92618595 on Build GRCh38
rs887655080 Chr.9: 92618714-92618714 on Build GRCh38

View Full Product Details