Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110686805-110687319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUSK
Entrez Gene ID
4593
Gene Name
muscle associated receptor tyrosine kinase
Gene Aliases
CMS9, FADS
UniGene
Hs.521653
Gene Chromosome Location
Chr.9: 110668189-110806633 on Build GRCh38
Transcripts
NM_001166280.1 NM_001166281.1 NM_005592.3 XM_005251994.3 XM_005251995.3 XM_005251996.3 XM_017014734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4704148 c.358+2T>C p.? Chr.9: 110687270-110687270 on Build GRCh38
6517329 c.327G>A p.V109V Chr.9: 110687237-110687237 on Build GRCh38
7119278 c.277G>C p.D93H Chr.9: 110687187-110687187 on Build GRCh38
4704149 c.4+2T>C p.? Chr.9: 110687270-110687270 on Build GRCh38
3652940 c.268G>A p.E90K Chr.9: 110687178-110687178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765760347 Chr.9: 110687212-110687212 on Build GRCh38
rs754254692 Chr.9: 110687106-110687106 on Build GRCh38
rs56130155 Chr.9: 110687135-110687135 on Build GRCh38
rs189825814 Chr.9: 110687073-110687073 on Build GRCh38
rs928578941 Chr.9: 110686871-110686871 on Build GRCh38
rs369626158 Chr.9: 110687088-110687088 on Build GRCh38
rs186208818 Chr.9: 110686920-110686920 on Build GRCh38
rs374507956 Chr.9: 110687050-110687050 on Build GRCh38
rs547322569 Chr.9: 110687140-110687140 on Build GRCh38
rs1023377168 Chr.9: 110686966-110686966 on Build GRCh38
rs754539657 Chr.9: 110687227-110687227 on Build GRCh38
rs55786136 Chr.9: 110687230-110687230 on Build GRCh38
rs551423795 Chr.9: 110687218-110687218 on Build GRCh38
rs779767402 Chr.9: 110687134-110687134 on Build GRCh38
rs1021042074 Chr.9: 110687127-110687127 on Build GRCh38
rs777606910 Chr.9: 110687271-110687271 on Build GRCh38
rs139108780 Chr.9: 110687084-110687084 on Build GRCh38
rs879255561 Chr.9: 110687130-110687131 on Build GRCh38
rs71501643 Chr.9: 110687097-110687097 on Build GRCh38
rs776815006 Chr.9: 110687143-110687143 on Build GRCh38
rs926599889 Chr.9: 110687175-110687175 on Build GRCh38
rs1020150091 Chr.9: 110686855-110686855 on Build GRCh38
rs761147453 Chr.9: 110687096-110687096 on Build GRCh38
rs180928221 Chr.9: 110687210-110687210 on Build GRCh38
rs919782247 Chr.9: 110686922-110686922 on Build GRCh38
rs931133158 Chr.9: 110686972-110686972 on Build GRCh38
rs779359576 Chr.9: 110687112-110687112 on Build GRCh38
rs1046843126 Chr.9: 110686980-110686980 on Build GRCh38
rs779636395 Chr.9: 110687071-110687071 on Build GRCh38
rs747736305 Chr.9: 110687179-110687179 on Build GRCh38
rs182019182 Chr.9: 110686852-110686852 on Build GRCh38
rs746157382 Chr.9: 110687165-110687165 on Build GRCh38
rs201273265 Chr.9: 110687221-110687221 on Build GRCh38
rs376236950 Chr.9: 110687287-110687287 on Build GRCh38
rs900977379 Chr.9: 110686877-110686877 on Build GRCh38
rs752772407 Chr.9: 110687092-110687092 on Build GRCh38
rs937232227 Chr.9: 110686911-110686911 on Build GRCh38
rs564153883 Chr.9: 110687043-110687043 on Build GRCh38
rs7869057 Chr.9: 110686991-110686991 on Build GRCh38
rs376146245 Chr.9: 110687102-110687102 on Build GRCh38
rs750101214 Chr.9: 110687128-110687128 on Build GRCh38
rs144807641 Chr.9: 110687288-110687288 on Build GRCh38
rs575256000 Chr.9: 110687266-110687266 on Build GRCh38
rs748123050 Chr.9: 110687259-110687259 on Build GRCh38
rs757659128 Chr.9: 110687108-110687108 on Build GRCh38
rs760781844 Chr.9: 110687206-110687206 on Build GRCh38
rs747987063 Chr.9: 110687142-110687142 on Build GRCh38
rs751667058 Chr.9: 110687096-110687097 on Build GRCh38
rs772926677 Chr.9: 110687149-110687149 on Build GRCh38
rs922487042 Chr.9: 110687186-110687186 on Build GRCh38
rs768622483 Chr.9: 110687080-110687080 on Build GRCh38
rs1005175267 Chr.9: 110686948-110686948 on Build GRCh38
rs35142681 Chr.9: 110687209-110687209 on Build GRCh38
rs767592370 Chr.9: 110687089-110687089 on Build GRCh38
rs768514590 Chr.9: 110687138-110687138 on Build GRCh38
rs769436204 Chr.9: 110687087-110687087 on Build GRCh38
rs560732808 Chr.9: 110686872-110686872 on Build GRCh38
rs1041074793 Chr.9: 110687045-110687045 on Build GRCh38
rs772270944 Chr.9: 110687173-110687173 on Build GRCh38
rs775598630 Chr.9: 110687192-110687192 on Build GRCh38
rs751133130 Chr.9: 110687225-110687225 on Build GRCh38
rs56181115 Chr.9: 110687222-110687222 on Build GRCh38
rs758118087 Chr.9: 110687131-110687131 on Build GRCh38
rs568882789 Chr.9: 110686961-110686961 on Build GRCh38

View Full Product Details