Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110767679-110768221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUSK
Entrez Gene ID
4593
Gene Name
muscle associated receptor tyrosine kinase
Gene Aliases
CMS9, FADS
UniGene
Hs.521653
Gene Chromosome Location
Chr.9: 110668189-110806633 on Build GRCh38
Transcripts
NM_001166280.1 NM_001166281.1 NM_005592.3 XM_005251994.3 XM_005251995.3 XM_005251996.3 XM_011518708.2 XM_017014734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6452645 c.1088G>T p.S363I Chr.9: 110767987-110767987 on Build GRCh38
6007526 c.1043C>T p.A348V Chr.9: 110767924-110767924 on Build GRCh38
7130904 c.1104G>C p.V368V Chr.9: 110767985-110767985 on Build GRCh38
3664005 c.1045G>T p.D349Y Chr.9: 110767926-110767926 on Build GRCh38
75495 c.1003G>A p.V335I Chr.9: 110767902-110767902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545091549 Chr.9: 110767751-110767751 on Build GRCh38
rs758777519 Chr.9: 110767962-110767962 on Build GRCh38
rs761698956 Chr.9: 110767888-110767888 on Build GRCh38
rs769749384 Chr.9: 110767912-110767912 on Build GRCh38
rs780437974 Chr.9: 110768132-110768132 on Build GRCh38
rs917141943 Chr.9: 110767943-110767943 on Build GRCh38
rs773285595 Chr.9: 110768027-110768027 on Build GRCh38
rs563391603 Chr.9: 110767877-110767877 on Build GRCh38
rs772388991 Chr.9: 110767868-110767868 on Build GRCh38
rs373175897 Chr.9: 110767913-110767913 on Build GRCh38
rs753955487 Chr.9: 110768098-110768098 on Build GRCh38
rs767542639 Chr.9: 110768101-110768101 on Build GRCh38
rs940003619 Chr.9: 110768140-110768140 on Build GRCh38
rs761980765 Chr.9: 110768103-110768103 on Build GRCh38
rs750990375 Chr.9: 110768124-110768124 on Build GRCh38
rs765009604 Chr.9: 110767748-110767748 on Build GRCh38
rs77508668 Chr.9: 110767708-110767708 on Build GRCh38
rs916924056 Chr.9: 110767714-110767716 on Build GRCh38
rs139945437 Chr.9: 110767961-110767961 on Build GRCh38
rs369023786 Chr.9: 110767851-110767851 on Build GRCh38
rs780616845 Chr.9: 110768004-110768004 on Build GRCh38
rs747717789 Chr.9: 110768018-110768018 on Build GRCh38
rs1038479586 Chr.9: 110767999-110767999 on Build GRCh38
rs201791900 Chr.9: 110767956-110767956 on Build GRCh38
rs760844052 Chr.9: 110768080-110768080 on Build GRCh38
rs770907001 Chr.9: 110768058-110768058 on Build GRCh38
rs757775674 Chr.9: 110767845-110767845 on Build GRCh38
rs376831940 Chr.9: 110767833-110767833 on Build GRCh38
rs780387236 Chr.9: 110767966-110767966 on Build GRCh38
rs372968908 Chr.9: 110768135-110768135 on Build GRCh38
rs758906037 Chr.9: 110768130-110768130 on Build GRCh38
rs543324300 Chr.9: 110767750-110767750 on Build GRCh38
rs934080592 Chr.9: 110767805-110767805 on Build GRCh38
rs1039859283 Chr.9: 110767798-110767800 on Build GRCh38
rs746608909 Chr.9: 110767871-110767871 on Build GRCh38
rs532285449 Chr.9: 110768085-110768085 on Build GRCh38
rs764142979 Chr.9: 110767937-110767937 on Build GRCh38
rs968145791 Chr.9: 110767853-110767853 on Build GRCh38
rs200434321 Chr.9: 110767925-110767925 on Build GRCh38
rs957660061 Chr.9: 110768013-110768013 on Build GRCh38
rs757240600 Chr.9: 110767931-110767932 on Build GRCh38
rs879041045 Chr.9: 110767745-110767745 on Build GRCh38
rs921440607 Chr.9: 110768061-110768061 on Build GRCh38
rs201014623 Chr.9: 110767998-110767998 on Build GRCh38
rs750173541 Chr.9: 110768165-110768165 on Build GRCh38
rs767875673 Chr.9: 110767794-110767794 on Build GRCh38
rs1018424290 Chr.9: 110768142-110768142 on Build GRCh38
rs1007037770 Chr.9: 110767741-110767741 on Build GRCh38
rs113978287 Chr.9: 110767744-110767745 on Build GRCh38
rs936153070 Chr.9: 110767778-110767778 on Build GRCh38
rs76912503 Chr.9: 110767755-110767755 on Build GRCh38
rs387906803 Chr.9: 110767930-110767930 on Build GRCh38
rs775131984 Chr.9: 110767869-110767869 on Build GRCh38
rs749815609 Chr.9: 110767843-110767843 on Build GRCh38
rs760965385 Chr.9: 110767931-110767931 on Build GRCh38
rs373788285 Chr.9: 110767935-110767935 on Build GRCh38
rs530795175 Chr.9: 110767919-110767919 on Build GRCh38
rs571312240 Chr.9: 110768047-110768047 on Build GRCh38
rs768323977 Chr.9: 110768001-110768001 on Build GRCh38
rs903973656 Chr.9: 110768147-110768147 on Build GRCh38
rs746206053 Chr.9: 110767859-110767859 on Build GRCh38
rs1051362388 Chr.9: 110768105-110768105 on Build GRCh38
rs199891500 Chr.9: 110767970-110767970 on Build GRCh38
rs891300911 Chr.9: 110767738-110767738 on Build GRCh38
rs776201930 Chr.9: 110767878-110767878 on Build GRCh38
rs756266862 Chr.9: 110767836-110767836 on Build GRCh38
rs867394741 Chr.9: 110767920-110767920 on Build GRCh38
rs375762569 Chr.9: 110767924-110767924 on Build GRCh38
rs201804790 Chr.9: 110767828-110767828 on Build GRCh38
rs755300514 Chr.9: 110767753-110767753 on Build GRCh38
rs1030997195 Chr.9: 110767904-110767904 on Build GRCh38
rs765175137 Chr.9: 110768119-110768119 on Build GRCh38
rs777849522 Chr.9: 110767841-110767841 on Build GRCh38
rs899901944 Chr.9: 110767798-110767798 on Build GRCh38
rs200064775 Chr.9: 110767960-110767960 on Build GRCh38
rs1038386393 Chr.9: 110768151-110768151 on Build GRCh38
rs975514015 Chr.9: 110768014-110768014 on Build GRCh38
rs753919630 Chr.9: 110767954-110767954 on Build GRCh38
rs774282945 Chr.9: 110768074-110768074 on Build GRCh38

View Full Product Details