Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127457194-127457694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
UniGene
Hs.495188
Gene Chromosome Location
Chr.9: 127451486-127503501 on Build GRCh38
Transcripts
NM_001005373.3 NM_001005374.3 NM_001190723.2 NM_138361.5 XM_006717316.3 XM_017015283.1 XM_017015284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3904216 c.185T>A p.V62D Chr.9: 127457326-127457326 on Build GRCh38
7244067 c.249C>T p.I83I Chr.9: 127457390-127457390 on Build GRCh38
5571719 c.184G>A p.V62I Chr.9: 127457325-127457325 on Build GRCh38
6704599 c.225C>T p.C75C Chr.9: 127457366-127457366 on Build GRCh38
1755937 c.230T>C p.L77P Chr.9: 127457371-127457371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750961361 Chr.9: 127457423-127457423 on Build GRCh38
rs772059949 Chr.9: 127457266-127457266 on Build GRCh38
rs764331245 Chr.9: 127457396-127457396 on Build GRCh38
rs1013134942 Chr.9: 127457283-127457283 on Build GRCh38
rs1035065631 Chr.9: 127457583-127457583 on Build GRCh38
rs149769300 Chr.9: 127457372-127457372 on Build GRCh38
rs373669417 Chr.9: 127457386-127457386 on Build GRCh38
rs768056477 Chr.9: 127457331-127457331 on Build GRCh38
rs1009540300 Chr.9: 127457593-127457593 on Build GRCh38
rs899549693 Chr.9: 127457233-127457233 on Build GRCh38
rs200961616 Chr.9: 127457272-127457272 on Build GRCh38
rs1022289571 Chr.9: 127457604-127457604 on Build GRCh38
rs376815374 Chr.9: 127457262-127457262 on Build GRCh38
rs372890516 Chr.9: 127457290-127457290 on Build GRCh38
rs754499152 Chr.9: 127457430-127457430 on Build GRCh38
rs748775036 Chr.9: 127457330-127457330 on Build GRCh38
rs112435797 Chr.9: 127457443-127457443 on Build GRCh38
rs773271495 Chr.9: 127457271-127457271 on Build GRCh38
rs778338519 Chr.9: 127457435-127457435 on Build GRCh38
rs946830150 Chr.9: 127457223-127457223 on Build GRCh38
rs758774140 Chr.9: 127457297-127457297 on Build GRCh38
rs765686374 Chr.9: 127457403-127457403 on Build GRCh38
rs986516483 Chr.9: 127457668-127457668 on Build GRCh38
rs921465600 Chr.9: 127457647-127457647 on Build GRCh38
rs1035054313 Chr.9: 127457314-127457314 on Build GRCh38
rs942116544 Chr.9: 127457347-127457347 on Build GRCh38
rs376362600 Chr.9: 127457287-127457287 on Build GRCh38
rs531640717 Chr.9: 127457246-127457246 on Build GRCh38
rs1004118951 Chr.9: 127457264-127457264 on Build GRCh38
rs777594391 Chr.9: 127457442-127457442 on Build GRCh38
rs898447143 Chr.9: 127457518-127457518 on Build GRCh38
rs111841854 Chr.9: 127457251-127457251 on Build GRCh38
rs113869335 Chr.9: 127457324-127457324 on Build GRCh38
rs939649761 Chr.9: 127457482-127457482 on Build GRCh38
rs1012818848 Chr.9: 127457526-127457526 on Build GRCh38
rs142333739 Chr.9: 127457444-127457444 on Build GRCh38
rs778625125 Chr.9: 127457333-127457333 on Build GRCh38
rs540894306 Chr.9: 127457505-127457505 on Build GRCh38
rs200731968 Chr.9: 127457441-127457441 on Build GRCh38
rs955533865 Chr.9: 127457667-127457667 on Build GRCh38
rs2243903 Chr.9: 127457464-127457464 on Build GRCh38
rs754120092 Chr.9: 127457280-127457280 on Build GRCh38
rs781680086 Chr.9: 127457329-127457329 on Build GRCh38
rs573856964 Chr.9: 127457574-127457574 on Build GRCh38
rs570688892 Chr.9: 127457325-127457325 on Build GRCh38
rs546497500 Chr.9: 127457366-127457366 on Build GRCh38
rs562544656 Chr.9: 127457224-127457224 on Build GRCh38
rs751872462 Chr.9: 127457401-127457401 on Build GRCh38
rs975687441 Chr.9: 127457630-127457630 on Build GRCh38
rs145947414 Chr.9: 127457527-127457527 on Build GRCh38
rs929873709 Chr.9: 127457587-127457587 on Build GRCh38
rs933740966 Chr.9: 127457234-127457234 on Build GRCh38
rs533231306 Chr.9: 127457339-127457339 on Build GRCh38
rs778392265 Chr.9: 127457567-127457567 on Build GRCh38
rs998198362 Chr.9: 127457397-127457397 on Build GRCh38
rs988397209 Chr.9: 127457380-127457380 on Build GRCh38
rs367844312 Chr.9: 127457409-127457409 on Build GRCh38
rs551352437 Chr.9: 127457288-127457288 on Build GRCh38
rs566623718 Chr.9: 127457375-127457375 on Build GRCh38
rs956712708 Chr.9: 127457588-127457588 on Build GRCh38
rs752926906 Chr.9: 127457291-127457291 on Build GRCh38
rs968726938 Chr.9: 127457614-127457614 on Build GRCh38
rs2243906 Chr.9: 127457390-127457390 on Build GRCh38
rs752028583 Chr.9: 127457298-127457298 on Build GRCh38
rs992734236 Chr.9: 127457230-127457230 on Build GRCh38
rs760581208 Chr.9: 127457370-127457370 on Build GRCh38

View Full Product Details