Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 63696725-63697265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF9
Entrez Gene ID
23229
Gene Name
Cdc42 guanine nucleotide exchange factor 9
Gene Aliases
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2
UniGene
Hs.54697
Gene Chromosome Location
Chr.X: 63634967-63786025 on Build GRCh38
Transcripts
NM_001173479.1 NM_001173480.1 NM_015185.2 XM_005262249.2 XM_005262250.3 XM_005262251.1 XM_005262252.1 XM_011530890.1 XM_011530892.1 XM_017029363.1 XM_017029364.1 XM_017029365.1 XM_017029366.1 XM_017029367.1 XM_017029368.1 XM_017029369.1 XM_017029370.1 XM_017029371.1 XM_017029372.1 XM_017029373.1 XM_017029374.1 XM_017029375.1 XM_017029376.1 XM_017029377.1 XM_017029378.1 XM_017029379.1 XM_017029380.1

SNP Detail

SNP ID Chromosome Location
rs186701260 Chr.X: 63696751-63696751 on Build GRCh38
rs376741993 Chr.X: 63696761-63696761 on Build GRCh38
rs56182751 Chr.X: 63697128-63697128 on Build GRCh38
rs782511320 Chr.X: 63697080-63697080 on Build GRCh38
rs138513536 Chr.X: 63696816-63696816 on Build GRCh38
rs782684706 Chr.X: 63697173-63697173 on Build GRCh38
rs7882107 Chr.X: 63697235-63697235 on Build GRCh38
rs781955613 Chr.X: 63696835-63696835 on Build GRCh38
rs150129110 Chr.X: 63697125-63697125 on Build GRCh38
rs781819993 Chr.X: 63697122-63697122 on Build GRCh38
rs782401617 Chr.X: 63697239-63697239 on Build GRCh38
rs182966851 Chr.X: 63697045-63697045 on Build GRCh38
rs191101828 Chr.X: 63696894-63696894 on Build GRCh38
rs782685817 Chr.X: 63696821-63696821 on Build GRCh38
rs782246639 Chr.X: 63697225-63697225 on Build GRCh38
rs782778780 Chr.X: 63697109-63697109 on Build GRCh38
rs3747335 Chr.X: 63697018-63697018 on Build GRCh38
rs782108335 Chr.X: 63696873-63696873 on Build GRCh38
rs782187939 Chr.X: 63697143-63697143 on Build GRCh38
rs782644562 Chr.X: 63696949-63696949 on Build GRCh38
rs782598395 Chr.X: 63697065-63697065 on Build GRCh38
rs782520132 Chr.X: 63697152-63697152 on Build GRCh38
rs371207820 Chr.X: 63696896-63696896 on Build GRCh38
rs781799757 Chr.X: 63696981-63696981 on Build GRCh38

View Full Product Details