Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71101997-71102493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXO4
Entrez Gene ID
4303
Gene Name
forkhead box O4
Gene Aliases
AFX, AFX1, MLLT7
UniGene
Hs.584654
Gene Chromosome Location
Chr.X: 71096149-71103534 on Build GRCh38
Transcripts
NM_001170931.1 NM_005938.3 XM_011530957.2

SNP Detail

SNP ID Chromosome Location
rs781100300 Chr.X: 71102045-71102045 on Build GRCh38
rs769715798 Chr.X: 71102059-71102059 on Build GRCh38
rs752546328 Chr.X: 71102225-71102225 on Build GRCh38
rs920350032 Chr.X: 71102144-71102144 on Build GRCh38
rs905882899 Chr.X: 71102091-71102091 on Build GRCh38
rs907736873 Chr.X: 71102074-71102074 on Build GRCh38
rs894583099 Chr.X: 71102463-71102464 on Build GRCh38
rs1015438395 Chr.X: 71102273-71102273 on Build GRCh38
rs950348528 Chr.X: 71102073-71102073 on Build GRCh38
rs903397543 Chr.X: 71102433-71102433 on Build GRCh38
rs907191787 Chr.X: 71102471-71102471 on Build GRCh38
rs1001526371 Chr.X: 71102178-71102178 on Build GRCh38
rs961477430 Chr.X: 71102316-71102316 on Build GRCh38
rs761170509 Chr.X: 71102036-71102036 on Build GRCh38
rs985163355 Chr.X: 71102442-71102442 on Build GRCh38
rs746012861 Chr.X: 71102054-71102054 on Build GRCh38
rs765120789 Chr.X: 71102107-71102107 on Build GRCh38
rs758094485 Chr.X: 71102400-71102400 on Build GRCh38
rs758355302 Chr.X: 71102313-71102313 on Build GRCh38
rs945005162 Chr.X: 71102346-71102346 on Build GRCh38
rs751087989 Chr.X: 71102450-71102450 on Build GRCh38
rs1005361364 Chr.X: 71102268-71102268 on Build GRCh38
rs925726791 Chr.X: 71102022-71102022 on Build GRCh38
rs911528649 Chr.X: 71102298-71102298 on Build GRCh38
rs887888380 Chr.X: 71102260-71102260 on Build GRCh38
rs779377088 Chr.X: 71102437-71102437 on Build GRCh38
rs1012989267 Chr.X: 71102464-71102464 on Build GRCh38
rs953935641 Chr.X: 71102436-71102436 on Build GRCh38
rs779351544 Chr.X: 71102047-71102048 on Build GRCh38
rs764953250 Chr.X: 71102167-71102167 on Build GRCh38
rs1053319758 Chr.X: 71102067-71102067 on Build GRCh38
rs769156618 Chr.X: 71102130-71102130 on Build GRCh38
rs931702320 Chr.X: 71102152-71102152 on Build GRCh38
rs201345911 Chr.X: 71102069-71102069 on Build GRCh38
rs750004634 Chr.X: 71102197-71102197 on Build GRCh38
rs998116095 Chr.X: 71102379-71102379 on Build GRCh38
rs754909839 Chr.X: 71102466-71102466 on Build GRCh38

View Full Product Details