Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110697196-110697784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUSK
Entrez Gene ID
4593
Gene Name
muscle associated receptor tyrosine kinase
Gene Aliases
CMS9, FADS
UniGene
Hs.521653
Gene Chromosome Location
Chr.9: 110668189-110806633 on Build GRCh38
Transcripts
NM_001166280.1 NM_001166281.1 NM_005592.3 XM_005251994.3 XM_005251995.3 XM_005251996.3 XM_017014734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4704151 c.581A>C p.N194T Chr.9: 110697419-110697419 on Build GRCh38
6718233 c.538G>A p.V180I Chr.9: 110697376-110697376 on Build GRCh38
4637215 c.157G>T p.E53* Chr.9: 110697349-110697349 on Build GRCh38
5021611 c.183C>T p.N61N Chr.9: 110697375-110697375 on Build GRCh38
4994409 c.496C>T p.R166* Chr.9: 110697334-110697334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761259585 Chr.9: 110697435-110697435 on Build GRCh38
rs750542388 Chr.9: 110697651-110697651 on Build GRCh38
rs1012874690 Chr.9: 110697642-110697643 on Build GRCh38
rs947722830 Chr.9: 110697304-110697304 on Build GRCh38
rs777933853 Chr.9: 110697278-110697278 on Build GRCh38
rs564402810 Chr.9: 110697584-110697584 on Build GRCh38
rs1021776155 Chr.9: 110697737-110697737 on Build GRCh38
rs756442664 Chr.9: 110697455-110697455 on Build GRCh38
rs202045225 Chr.9: 110697338-110697338 on Build GRCh38
rs377403553 Chr.9: 110697495-110697495 on Build GRCh38
rs779839934 Chr.9: 110697371-110697371 on Build GRCh38
rs140983107 Chr.9: 110697535-110697535 on Build GRCh38
rs542900379 Chr.9: 110697404-110697404 on Build GRCh38
rs773036359 Chr.9: 110697442-110697442 on Build GRCh38
rs918672918 Chr.9: 110697583-110697583 on Build GRCh38
rs188095325 Chr.9: 110697727-110697727 on Build GRCh38
rs761192971 Chr.9: 110697665-110697665 on Build GRCh38
rs371267116 Chr.9: 110697436-110697436 on Build GRCh38
rs992320364 Chr.9: 110697233-110697233 on Build GRCh38
rs372148404 Chr.9: 110697303-110697303 on Build GRCh38
rs750334364 Chr.9: 110697343-110697343 on Build GRCh38
rs561525761 Chr.9: 110697431-110697431 on Build GRCh38
rs942289974 Chr.9: 110697379-110697379 on Build GRCh38
rs373304727 Chr.9: 110697395-110697395 on Build GRCh38
rs41279053 Chr.9: 110697488-110697488 on Build GRCh38
rs954202988 Chr.9: 110697750-110697750 on Build GRCh38
rs764387006 Chr.9: 110697460-110697460 on Build GRCh38
rs774830039 Chr.9: 110697283-110697283 on Build GRCh38
rs369254308 Chr.9: 110697387-110697387 on Build GRCh38
rs1049736056 Chr.9: 110697217-110697217 on Build GRCh38
rs770008360 Chr.9: 110697275-110697275 on Build GRCh38
rs370795070 Chr.9: 110697507-110697507 on Build GRCh38
rs969187973 Chr.9: 110697656-110697656 on Build GRCh38
rs968681825 Chr.9: 110697559-110697559 on Build GRCh38
rs1036994108 Chr.9: 110697579-110697579 on Build GRCh38
rs762882154 Chr.9: 110697447-110697447 on Build GRCh38
rs199573004 Chr.9: 110697484-110697484 on Build GRCh38
rs41279051 Chr.9: 110697375-110697375 on Build GRCh38
rs554508414 Chr.9: 110697690-110697690 on Build GRCh38
rs775032591 Chr.9: 110697307-110697307 on Build GRCh38
rs777642014 Chr.9: 110697277-110697277 on Build GRCh38
rs1031306799 Chr.9: 110697701-110697701 on Build GRCh38
rs200789677 Chr.9: 110697477-110697477 on Build GRCh38
rs1021607988 Chr.9: 110697543-110697543 on Build GRCh38
rs529933737 Chr.9: 110697220-110697220 on Build GRCh38
rs747354449 Chr.9: 110697491-110697491 on Build GRCh38
rs746976294 Chr.9: 110697309-110697309 on Build GRCh38
rs765052433 Chr.9: 110697340-110697340 on Build GRCh38
rs746719179 Chr.9: 110697423-110697423 on Build GRCh38
rs772324297 Chr.9: 110697417-110697417 on Build GRCh38
rs762720455 Chr.9: 110697679-110697679 on Build GRCh38
rs760250797 Chr.9: 110697317-110697317 on Build GRCh38
rs753330850 Chr.9: 110697336-110697336 on Build GRCh38
rs747766753 Chr.9: 110697581-110697581 on Build GRCh38
rs912543448 Chr.9: 110697225-110697225 on Build GRCh38
rs897695611 Chr.9: 110697254-110697254 on Build GRCh38
rs1002666517 Chr.9: 110697402-110697402 on Build GRCh38
rs866747138 Chr.9: 110697412-110697412 on Build GRCh38
rs369558015 Chr.9: 110697335-110697335 on Build GRCh38
rs759792090 Chr.9: 110697285-110697285 on Build GRCh38
rs754060972 Chr.9: 110697465-110697465 on Build GRCh38
rs944033254 Chr.9: 110697562-110697562 on Build GRCh38
rs1044335661 Chr.9: 110697658-110697658 on Build GRCh38
rs377353611 Chr.9: 110697420-110697420 on Build GRCh38
rs558248796 Chr.9: 110697741-110697741 on Build GRCh38
rs758149352 Chr.9: 110697351-110697351 on Build GRCh38
rs533666930 Chr.9: 110697736-110697736 on Build GRCh38
rs539930697 Chr.9: 110697239-110697239 on Build GRCh38
rs749359907 Chr.9: 110697391-110697391 on Build GRCh38
rs375179321 Chr.9: 110697308-110697308 on Build GRCh38
rs576493819 Chr.9: 110697762-110697762 on Build GRCh38
rs769683158 Chr.9: 110697494-110697494 on Build GRCh38
rs751393601 Chr.9: 110697454-110697454 on Build GRCh38
rs550911832 Chr.9: 110697610-110697610 on Build GRCh38
rs532568105 Chr.9: 110697591-110697591 on Build GRCh38
rs767878101 Chr.9: 110697337-110697337 on Build GRCh38
rs763656507 Chr.9: 110697334-110697334 on Build GRCh38
rs184382460 Chr.9: 110697282-110697282 on Build GRCh38
rs536542832 Chr.9: 110697678-110697678 on Build GRCh38
rs866681178 Chr.9: 110697366-110697366 on Build GRCh38
rs768273510 Chr.9: 110697427-110697427 on Build GRCh38
rs569058852 Chr.9: 110697614-110697614 on Build GRCh38
rs749878301 Chr.9: 110697280-110697280 on Build GRCh38
rs746340721 Chr.9: 110697408-110697408 on Build GRCh38
rs944163647 Chr.9: 110697236-110697236 on Build GRCh38

View Full Product Details