Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110668575-110669127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUSK
Entrez Gene ID
4593
Gene Name
muscle associated receptor tyrosine kinase
Gene Aliases
CMS9, FADS
UniGene
Hs.521653
Gene Chromosome Location
Chr.9: 110668189-110806633 on Build GRCh38
Transcripts
NM_001166280.1 NM_001166281.1 NM_005592.3 XM_005251994.3 XM_005251995.3 XM_005251996.3 XM_017014734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3652932 c.64G>A p.E22K Chr.9: 110668968-110668968 on Build GRCh38
752076 c.7G>A p.E3K Chr.9: 110668911-110668911 on Build GRCh38
5607505 c.22C>T p.P8S Chr.9: 110668926-110668926 on Build GRCh38
6291602 c.79G>A p.A27T Chr.9: 110668983-110668983 on Build GRCh38
3432769 c.13G>A p.V5I Chr.9: 110668917-110668917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs863223335 Chr.9: 110668944-110668945 on Build GRCh38
rs200783529 Chr.9: 110668985-110668985 on Build GRCh38
rs1004624845 Chr.9: 110668831-110668831 on Build GRCh38
rs767552333 Chr.9: 110669029-110669029 on Build GRCh38
rs189842985 Chr.9: 110668759-110668759 on Build GRCh38
rs138941963 Chr.9: 110668644-110668644 on Build GRCh38
rs368386911 Chr.9: 110669000-110669000 on Build GRCh38
rs41279047 Chr.9: 110668823-110668823 on Build GRCh38
rs762340994 Chr.9: 110668911-110668911 on Build GRCh38
rs894991540 Chr.9: 110668802-110668802 on Build GRCh38
rs775462791 Chr.9: 110669032-110669032 on Build GRCh38
rs781594297 Chr.9: 110668987-110668987 on Build GRCh38
rs990747864 Chr.9: 110668888-110668888 on Build GRCh38
rs146290093 Chr.9: 110668649-110668649 on Build GRCh38
rs116884176 Chr.9: 110668862-110668862 on Build GRCh38
rs142830788 Chr.9: 110669028-110669028 on Build GRCh38
rs752539984 Chr.9: 110668934-110668934 on Build GRCh38
rs747203404 Chr.9: 110668921-110668921 on Build GRCh38
rs913349921 Chr.9: 110668668-110668668 on Build GRCh38
rs139516442 Chr.9: 110668994-110668994 on Build GRCh38
rs746254848 Chr.9: 110668947-110668947 on Build GRCh38
rs754793598 Chr.9: 110668761-110668761 on Build GRCh38
rs886226793 Chr.9: 110668825-110668825 on Build GRCh38
rs761520279 Chr.9: 110668887-110668887 on Build GRCh38
rs182277457 Chr.9: 110668896-110668896 on Build GRCh38
rs965664665 Chr.9: 110668846-110668846 on Build GRCh38
rs765521756 Chr.9: 110668950-110668950 on Build GRCh38
rs149441176 Chr.9: 110668647-110668647 on Build GRCh38
rs776914649 Chr.9: 110668885-110668885 on Build GRCh38
rs538723944 Chr.9: 110668880-110668880 on Build GRCh38
rs1055427410 Chr.9: 110668785-110668785 on Build GRCh38
rs995107188 Chr.9: 110668807-110668807 on Build GRCh38
rs772827480 Chr.9: 110668901-110668901 on Build GRCh38
rs916508850 Chr.9: 110669073-110669073 on Build GRCh38
rs376094463 Chr.9: 110668836-110668836 on Build GRCh38
rs773846871 Chr.9: 110669012-110669012 on Build GRCh38
rs374456998 Chr.9: 110668916-110668916 on Build GRCh38
rs748491244 Chr.9: 110668990-110668990 on Build GRCh38
rs907206161 Chr.9: 110668733-110668733 on Build GRCh38
rs1039983244 Chr.9: 110669096-110669096 on Build GRCh38
rs993634182 Chr.9: 110668675-110668675 on Build GRCh38
rs1055785043 Chr.9: 110669039-110669039 on Build GRCh38
rs990265978 Chr.9: 110668658-110668658 on Build GRCh38
rs765874906 Chr.9: 110668917-110668917 on Build GRCh38
rs185957122 Chr.9: 110669010-110669010 on Build GRCh38
rs577843415 Chr.9: 110668876-110668876 on Build GRCh38
rs888067514 Chr.9: 110668730-110668730 on Build GRCh38
rs771171200 Chr.9: 110668719-110668719 on Build GRCh38
rs769020314 Chr.9: 110668881-110668881 on Build GRCh38
rs897541127 Chr.9: 110668673-110668673 on Build GRCh38
rs748922389 Chr.9: 110669008-110669008 on Build GRCh38
rs757545049 Chr.9: 110669013-110669013 on Build GRCh38
rs747473837 Chr.9: 110668965-110668965 on Build GRCh38
rs745909042 Chr.9: 110668863-110668863 on Build GRCh38
rs1018628051 Chr.9: 110668838-110668838 on Build GRCh38
rs758427621 Chr.9: 110668962-110668962 on Build GRCh38
rs767429379 Chr.9: 110668931-110668931 on Build GRCh38
rs966315457 Chr.9: 110668605-110668605 on Build GRCh38
rs537397996 Chr.9: 110668704-110668704 on Build GRCh38
rs946117039 Chr.9: 110668676-110668676 on Build GRCh38
rs750615817 Chr.9: 110668961-110668961 on Build GRCh38
rs542475895 Chr.9: 110668761-110668762 on Build GRCh38
rs772670014 Chr.9: 110668873-110668873 on Build GRCh38
rs569703798 Chr.9: 110668671-110668671 on Build GRCh38

View Full Product Details