Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6643374-6643972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACRBP
Entrez Gene ID
84519
Gene Name
acrosin binding protein
Gene Aliases
CT23, OY-TES-1, SP32
UniGene
Hs.123239
Gene Chromosome Location
Chr.12: 6638076-6647414 on Build GRCh38
Transcripts
NM_032489.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592486 c.993G>A p.S331S Chr.12: 6643623-6643623 on Build GRCh38
4610939 c.1023delC p.T342fs*39 Chr.12: 6643593-6643593 on Build GRCh38
3704373 c.1070G>A p.G357E Chr.12: 6643546-6643546 on Build GRCh38
2098771 c.1053G>T p.E351D Chr.12: 6643563-6643563 on Build GRCh38
4044372 c.1077G>A p.S359S Chr.12: 6643539-6643539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202038844 Chr.12: 6643515-6643515 on Build GRCh38
rs746360356 Chr.12: 6643571-6643571 on Build GRCh38
rs1025545898 Chr.12: 6643651-6643651 on Build GRCh38
rs754631736 Chr.12: 6643535-6643535 on Build GRCh38
rs924726415 Chr.12: 6643922-6643922 on Build GRCh38
rs537117163 Chr.12: 6643676-6643678 on Build GRCh38
rs558099847 Chr.12: 6643782-6643782 on Build GRCh38
rs975333174 Chr.12: 6643851-6643851 on Build GRCh38
rs986672434 Chr.12: 6643793-6643793 on Build GRCh38
rs912380321 Chr.12: 6643819-6643819 on Build GRCh38
rs758950655 Chr.12: 6643666-6643666 on Build GRCh38
rs748307760 Chr.12: 6643697-6643697 on Build GRCh38
rs1033973984 Chr.12: 6643418-6643418 on Build GRCh38
rs578107271 Chr.12: 6643792-6643792 on Build GRCh38
rs747509075 Chr.12: 6643830-6643830 on Build GRCh38
rs148058585 Chr.12: 6643668-6643668 on Build GRCh38
rs762873728 Chr.12: 6643623-6643623 on Build GRCh38
rs201182033 Chr.12: 6643582-6643582 on Build GRCh38
rs549350360 Chr.12: 6643719-6643719 on Build GRCh38
rs141443837 Chr.12: 6643619-6643619 on Build GRCh38
rs375634366 Chr.12: 6643715-6643715 on Build GRCh38
rs200946482 Chr.12: 6643587-6643587 on Build GRCh38
rs760943448 Chr.12: 6643496-6643496 on Build GRCh38
rs866331954 Chr.12: 6643595-6643595 on Build GRCh38
rs147022968 Chr.12: 6643662-6643662 on Build GRCh38
rs3741923 Chr.12: 6643610-6643610 on Build GRCh38
rs1025838518 Chr.12: 6643774-6643774 on Build GRCh38
rs962469463 Chr.12: 6643440-6643440 on Build GRCh38
rs936618403 Chr.12: 6643608-6643608 on Build GRCh38
rs34652723 Chr.12: 6643572-6643572 on Build GRCh38
rs774114556 Chr.12: 6643627-6643627 on Build GRCh38
rs914013384 Chr.12: 6643946-6643946 on Build GRCh38
rs768800434 Chr.12: 6643500-6643500 on Build GRCh38
rs148544015 Chr.12: 6643907-6643907 on Build GRCh38
rs1041790814 Chr.12: 6643401-6643401 on Build GRCh38
rs779501607 Chr.12: 6643552-6643552 on Build GRCh38
rs918306175 Chr.12: 6643557-6643557 on Build GRCh38
rs780534523 Chr.12: 6643574-6643574 on Build GRCh38
rs763689381 Chr.12: 6643658-6643658 on Build GRCh38
rs192129797 Chr.12: 6643506-6643506 on Build GRCh38
rs770069287 Chr.12: 6643700-6643700 on Build GRCh38
rs1057123504 Chr.12: 6643667-6643667 on Build GRCh38
rs964015659 Chr.12: 6643852-6643852 on Build GRCh38
rs553821136 Chr.12: 6643877-6643877 on Build GRCh38
rs766150389 Chr.12: 6643533-6643533 on Build GRCh38
rs767126238 Chr.12: 6643539-6643539 on Build GRCh38
rs531475457 Chr.12: 6643575-6643575 on Build GRCh38
rs773139542 Chr.12: 6643514-6643514 on Build GRCh38
rs200428818 Chr.12: 6643650-6643650 on Build GRCh38
rs773338294 Chr.12: 6643620-6643620 on Build GRCh38
rs947229315 Chr.12: 6643895-6643895 on Build GRCh38
rs116003603 Chr.12: 6643706-6643706 on Build GRCh38
rs755301869 Chr.12: 6643685-6643685 on Build GRCh38
rs758763792 Chr.12: 6643645-6643645 on Build GRCh38
rs752303255 Chr.12: 6643647-6643647 on Build GRCh38
rs75009607 Chr.12: 6643474-6643474 on Build GRCh38
rs781419975 Chr.12: 6643689-6643689 on Build GRCh38
rs912506881 Chr.12: 6643548-6643548 on Build GRCh38
rs748194272 Chr.12: 6643597-6643597 on Build GRCh38
rs751426530 Chr.12: 6643534-6643534 on Build GRCh38
rs370490182 Chr.12: 6643882-6643882 on Build GRCh38
rs766347409 Chr.12: 6643624-6643624 on Build GRCh38
rs117531360 Chr.12: 6643752-6643752 on Build GRCh38
rs755580617 Chr.12: 6643545-6643545 on Build GRCh38
rs986911732 Chr.12: 6643799-6643799 on Build GRCh38
rs987008013 Chr.12: 6643544-6643544 on Build GRCh38
rs1002854948 Chr.12: 6643469-6643469 on Build GRCh38
rs963525694 Chr.12: 6643724-6643724 on Build GRCh38
rs562110688 Chr.12: 6643919-6643919 on Build GRCh38
rs903197398 Chr.12: 6643468-6643468 on Build GRCh38
rs377344589 Chr.12: 6643495-6643495 on Build GRCh38
rs776750088 Chr.12: 6643502-6643502 on Build GRCh38
rs936670297 Chr.12: 6643883-6643883 on Build GRCh38
rs759275347 Chr.12: 6643635-6643635 on Build GRCh38
rs184177675 Chr.12: 6643716-6643716 on Build GRCh38
rs79806562 Chr.12: 6643820-6643820 on Build GRCh38
rs942631993 Chr.12: 6643609-6643609 on Build GRCh38
rs903251949 Chr.12: 6643659-6643659 on Build GRCh38
rs747357088 Chr.12: 6643669-6643669 on Build GRCh38
rs894758134 Chr.12: 6643672-6643672 on Build GRCh38
rs373641669 Chr.12: 6643541-6643541 on Build GRCh38
rs376767799 Chr.12: 6643513-6643513 on Build GRCh38
rs1013783217 Chr.12: 6643648-6643648 on Build GRCh38
rs569591906 Chr.12: 6643746-6643746 on Build GRCh38

View Full Product Details