Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50137504-50138019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS5
Entrez Gene ID
91012
Gene Name
ceramide synthase 5
Gene Aliases
LASS5, Trh4
UniGene
Hs.270525
Gene Chromosome Location
Chr.12: 50129306-50167533 on Build GRCh38
Transcripts
NM_001281731.1 NM_147190.3 NR_104035.1 XM_005269220.2 XM_006719687.3 XM_011538975.2 XM_017020202.1 XM_017020203.1 XM_017020204.1 XM_017020205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5445629 c.629_631delAAA p.K210delK Chr.12: 50137733-50137735 on Build GRCh38
358698 c.589T>G p.F197V Chr.12: 50137775-50137775 on Build GRCh38
6479655 c.576C>T p.I192I Chr.12: 50137788-50137788 on Build GRCh38
940308 c.632G>T p.R211I Chr.12: 50137732-50137732 on Build GRCh38
4602069 c.593A>G p.Y198C Chr.12: 50137771-50137771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760914801 Chr.12: 50137699-50137699 on Build GRCh38
rs141350586 Chr.12: 50137540-50137540 on Build GRCh38
rs986975719 Chr.12: 50137596-50137596 on Build GRCh38
rs1017860770 Chr.12: 50137981-50137981 on Build GRCh38
rs1010931672 Chr.12: 50137965-50137965 on Build GRCh38
rs955786996 Chr.12: 50137978-50137978 on Build GRCh38
rs1054669989 Chr.12: 50137946-50137946 on Build GRCh38
rs778158310 Chr.12: 50137850-50137850 on Build GRCh38
rs758932233 Chr.12: 50137839-50137839 on Build GRCh38
rs375263430 Chr.12: 50137538-50137538 on Build GRCh38
rs1020514964 Chr.12: 50137976-50137976 on Build GRCh38
rs963165217 Chr.12: 50137621-50137621 on Build GRCh38
rs759894439 Chr.12: 50137826-50137826 on Build GRCh38
rs991598356 Chr.12: 50137656-50137656 on Build GRCh38
rs145132541 Chr.12: 50137614-50137614 on Build GRCh38
rs779197897 Chr.12: 50137720-50137720 on Build GRCh38
rs572861694 Chr.12: 50137654-50137654 on Build GRCh38
rs1020402072 Chr.12: 50137541-50137541 on Build GRCh38
rs770560896 Chr.12: 50137737-50137737 on Build GRCh38
rs973884680 Chr.12: 50137675-50137675 on Build GRCh38
rs1004415277 Chr.12: 50137610-50137610 on Build GRCh38
rs535960520 Chr.12: 50137651-50137651 on Build GRCh38
rs762848453 Chr.12: 50137771-50137771 on Build GRCh38
rs200952043 Chr.12: 50137866-50137866 on Build GRCh38
rs950098139 Chr.12: 50137853-50137853 on Build GRCh38
rs769283988 Chr.12: 50137749-50137749 on Build GRCh38
rs747363608 Chr.12: 50137729-50137729 on Build GRCh38
rs188522300 Chr.12: 50137591-50137591 on Build GRCh38
rs193171969 Chr.12: 50137835-50137835 on Build GRCh38
rs565323292 Chr.12: 50137636-50137636 on Build GRCh38
rs1001581526 Chr.12: 50137903-50137903 on Build GRCh38
rs753242928 Chr.12: 50137721-50137721 on Build GRCh38
rs752862863 Chr.12: 50137838-50137838 on Build GRCh38
rs760048596 Chr.12: 50137729-50137733 on Build GRCh38
rs201248008 Chr.12: 50137740-50137740 on Build GRCh38
rs942423746 Chr.12: 50137618-50137618 on Build GRCh38
rs1045736112 Chr.12: 50137861-50137861 on Build GRCh38
rs761523615 Chr.12: 50137789-50137789 on Build GRCh38
rs373859358 Chr.12: 50137719-50137719 on Build GRCh38
rs745501333 Chr.12: 50137868-50137868 on Build GRCh38
rs752153335 Chr.12: 50137590-50137590 on Build GRCh38
rs775033866 Chr.12: 50137768-50137768 on Build GRCh38
rs370492865 Chr.12: 50137706-50137706 on Build GRCh38
rs766551381 Chr.12: 50137792-50137792 on Build GRCh38
rs963766074 Chr.12: 50137995-50137995 on Build GRCh38
rs377417503 Chr.12: 50137693-50137693 on Build GRCh38
rs778167149 Chr.12: 50137727-50137727 on Build GRCh38
rs879574690 Chr.12: 50137665-50137665 on Build GRCh38
rs774144063 Chr.12: 50137781-50137781 on Build GRCh38
rs561117963 Chr.12: 50137905-50137905 on Build GRCh38
rs745587443 Chr.12: 50137748-50137748 on Build GRCh38
rs1046963827 Chr.12: 50137776-50137776 on Build GRCh38
rs893294982 Chr.12: 50137958-50137958 on Build GRCh38
rs753895191 Chr.12: 50137715-50137715 on Build GRCh38
rs369848938 Chr.12: 50137744-50137744 on Build GRCh38
rs575645059 Chr.12: 50137851-50137851 on Build GRCh38
rs766530055 Chr.12: 50137704-50137704 on Build GRCh38
rs868176004 Chr.12: 50137535-50137535 on Build GRCh38
rs558765035 Chr.12: 50137836-50137836 on Build GRCh38
rs753998784 Chr.12: 50137793-50137793 on Build GRCh38
rs752054845 Chr.12: 50137979-50137979 on Build GRCh38
rs768510162 Chr.12: 50137773-50137773 on Build GRCh38
rs370748035 Chr.12: 50137852-50137852 on Build GRCh38
rs955052966 Chr.12: 50137553-50137553 on Build GRCh38
rs759022271 Chr.12: 50137724-50137724 on Build GRCh38

View Full Product Details