Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4514441-4515003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDIP1
Entrez Gene ID
29965
Gene Name
cell death-inducing p53 target 1
Gene Aliases
C16orf5, CDIP, I1, LITAFL
UniGene
Hs.654653
Gene Chromosome Location
Chr.16: 4510675-4538815 on Build GRCh38
Transcripts
NM_001199054.1 NM_001199055.1 NM_001199056.1 NM_013399.2

SNP Detail

SNP ID Chromosome Location
rs922410703 Chr.16: 4514715-4514715 on Build GRCh38
rs1001759724 Chr.16: 4514553-4514553 on Build GRCh38
rs1050027101 Chr.16: 4514784-4514784 on Build GRCh38
rs558836918 Chr.16: 4514577-4514577 on Build GRCh38
rs888025236 Chr.16: 4514906-4514906 on Build GRCh38
rs115971859 Chr.16: 4514893-4514893 on Build GRCh38
rs962451719 Chr.16: 4514934-4514934 on Build GRCh38
rs754775293 Chr.16: 4514838-4514838 on Build GRCh38
rs922514133 Chr.16: 4514465-4514465 on Build GRCh38
rs1037131597 Chr.16: 4514492-4514492 on Build GRCh38
rs536967230 Chr.16: 4514870-4514870 on Build GRCh38
rs556125618 Chr.16: 4514871-4514871 on Build GRCh38
rs959754432 Chr.16: 4514659-4514659 on Build GRCh38
rs771433591 Chr.16: 4514490-4514491 on Build GRCh38
rs972985932 Chr.16: 4514689-4514689 on Build GRCh38
rs772312410 Chr.16: 4514514-4514514 on Build GRCh38
rs945784537 Chr.16: 4514840-4514840 on Build GRCh38
rs955309055 Chr.16: 4514775-4514775 on Build GRCh38
rs143739825 Chr.16: 4514698-4514698 on Build GRCh38
rs1006634960 Chr.16: 4514915-4514915 on Build GRCh38
rs995583422 Chr.16: 4514837-4514837 on Build GRCh38
rs550120205 Chr.16: 4514490-4514490 on Build GRCh38
rs759889852 Chr.16: 4514512-4514512 on Build GRCh38
rs997586117 Chr.16: 4514946-4514946 on Build GRCh38
rs1009941318 Chr.16: 4514889-4514889 on Build GRCh38
rs912978067 Chr.16: 4514832-4514832 on Build GRCh38
rs534044460 Chr.16: 4514589-4514589 on Build GRCh38
rs912990852 Chr.16: 4514691-4514691 on Build GRCh38
rs182874739 Chr.16: 4514516-4514516 on Build GRCh38
rs907348077 Chr.16: 4514878-4514878 on Build GRCh38
rs767101041 Chr.16: 4514491-4514491 on Build GRCh38
rs907462853 Chr.16: 4514524-4514524 on Build GRCh38
rs952052690 Chr.16: 4514564-4514564 on Build GRCh38
rs962113632 Chr.16: 4514686-4514686 on Build GRCh38
rs912822152 Chr.16: 4514965-4514965 on Build GRCh38
rs537236880 Chr.16: 4514788-4514788 on Build GRCh38
rs985994270 Chr.16: 4514964-4514964 on Build GRCh38
rs935703248 Chr.16: 4514812-4514812 on Build GRCh38
rs545291525 Chr.16: 4514874-4514874 on Build GRCh38
rs1050217776 Chr.16: 4514464-4514464 on Build GRCh38
rs552647079 Chr.16: 4514872-4514873 on Build GRCh38
rs955237670 Chr.16: 4514980-4514980 on Build GRCh38
rs905678062 Chr.16: 4514841-4514841 on Build GRCh38
rs1030353690 Chr.16: 4514959-4514959 on Build GRCh38
rs569929732 Chr.16: 4514495-4514495 on Build GRCh38
rs755834508 Chr.16: 4514602-4514602 on Build GRCh38
rs565859157 Chr.16: 4514583-4514583 on Build GRCh38
rs1001752768 Chr.16: 4514853-4514856 on Build GRCh38
rs968434044 Chr.16: 4514704-4514708 on Build GRCh38
rs529645250 Chr.16: 4514481-4514481 on Build GRCh38
rs386788000 Chr.16: 4514588-4514589 on Build GRCh38
rs933876149 Chr.16: 4514471-4514471 on Build GRCh38
rs925440528 Chr.16: 4514709-4514709 on Build GRCh38
rs944280232 Chr.16: 4514966-4514966 on Build GRCh38
rs574976124 Chr.16: 4514883-4514893 on Build GRCh38
rs897283842 Chr.16: 4514816-4514816 on Build GRCh38
rs1034580563 Chr.16: 4514555-4514555 on Build GRCh38
rs558381069 Chr.16: 4514463-4514463 on Build GRCh38
rs979602131 Chr.16: 4514704-4514704 on Build GRCh38
rs1014892021 Chr.16: 4514919-4514919 on Build GRCh38
rs374909809 Chr.16: 4514888-4514898 on Build GRCh38
rs911125930 Chr.16: 4514793-4514793 on Build GRCh38
rs576354020 Chr.16: 4514873-4514873 on Build GRCh38
rs145947735 Chr.16: 4514813-4514813 on Build GRCh38
rs1010861027 Chr.16: 4514507-4514507 on Build GRCh38
rs954231337 Chr.16: 4514955-4514955 on Build GRCh38
rs867695655 Chr.16: 4514554-4514554 on Build GRCh38
rs553176932 Chr.16: 4514506-4514506 on Build GRCh38
rs889425100 Chr.16: 4514483-4514483 on Build GRCh38
rs1001689201 Chr.16: 4514890-4514890 on Build GRCh38
rs778532708 Chr.16: 4514844-4514844 on Build GRCh38
rs533110480 Chr.16: 4514588-4514588 on Build GRCh38

View Full Product Details