Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15439556-15440154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EAF1
Entrez Gene ID
85403
Gene Name
ELL associated factor 1
Gene Aliases
-
UniGene
Hs.716733
Gene Chromosome Location
Chr.3: 15427557-15442613 on Build GRCh38
Transcripts
NM_033083.6 XM_005265518.1 XM_011534165.1 XM_011534166.1

SNP Detail

SNP ID Chromosome Location
rs988939496 Chr.3: 15440078-15440078 on Build GRCh38
rs901322518 Chr.3: 15439637-15439637 on Build GRCh38
rs1026686974 Chr.3: 15439920-15439920 on Build GRCh38
rs912744841 Chr.3: 15439695-15439695 on Build GRCh38
rs773919220 Chr.3: 15439701-15439702 on Build GRCh38
rs553216885 Chr.3: 15439730-15439730 on Build GRCh38
rs187006178 Chr.3: 15439935-15439935 on Build GRCh38
rs1005625667 Chr.3: 15440015-15440016 on Build GRCh38
rs749427464 Chr.3: 15439686-15439686 on Build GRCh38
rs903760619 Chr.3: 15439807-15439807 on Build GRCh38
rs369047658 Chr.3: 15439784-15439784 on Build GRCh38
rs114263900 Chr.3: 15439814-15439814 on Build GRCh38
rs932766242 Chr.3: 15439941-15439941 on Build GRCh38
rs979911786 Chr.3: 15439812-15439812 on Build GRCh38
rs542089028 Chr.3: 15439775-15439779 on Build GRCh38
rs771267958 Chr.3: 15439747-15439747 on Build GRCh38
rs1024679633 Chr.3: 15439767-15439767 on Build GRCh38
rs574069723 Chr.3: 15440076-15440076 on Build GRCh38
rs1041722807 Chr.3: 15439583-15439583 on Build GRCh38
rs1053101869 Chr.3: 15439745-15439745 on Build GRCh38
rs573975596 Chr.3: 15439673-15439673 on Build GRCh38
rs913284461 Chr.3: 15440095-15440095 on Build GRCh38
rs896339869 Chr.3: 15439880-15439880 on Build GRCh38
rs927308346 Chr.3: 15440109-15440109 on Build GRCh38
rs986142980 Chr.3: 15440024-15440024 on Build GRCh38
rs1054948650 Chr.3: 15439816-15439816 on Build GRCh38
rs961121440 Chr.3: 15439915-15439915 on Build GRCh38
rs1043046254 Chr.3: 15439785-15439785 on Build GRCh38
rs773697376 Chr.3: 15439903-15439903 on Build GRCh38
rs1043034806 Chr.3: 15440099-15440099 on Build GRCh38
rs988409558 Chr.3: 15439868-15439868 on Build GRCh38
rs1005532069 Chr.3: 15439731-15439731 on Build GRCh38
rs932906187 Chr.3: 15439683-15439683 on Build GRCh38
rs997057130 Chr.3: 15439654-15439654 on Build GRCh38
rs115976562 Chr.3: 15440097-15440097 on Build GRCh38
rs961003211 Chr.3: 15439613-15439613 on Build GRCh38
rs888503445 Chr.3: 15439999-15439999 on Build GRCh38
rs1049939293 Chr.3: 15439942-15439942 on Build GRCh38
rs973752950 Chr.3: 15439638-15439638 on Build GRCh38
rs912884408 Chr.3: 15439891-15439891 on Build GRCh38
rs1015329486 Chr.3: 15439902-15439902 on Build GRCh38
rs954319227 Chr.3: 15439991-15439991 on Build GRCh38
rs372574859 Chr.3: 15439917-15439917 on Build GRCh38
rs573020593 Chr.3: 15439738-15439738 on Build GRCh38
rs146693113 Chr.3: 15440071-15440071 on Build GRCh38
rs1013438010 Chr.3: 15439897-15439897 on Build GRCh38

View Full Product Details