Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 71866579-71866927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD17B
Entrez Gene ID
51104
Gene Name
abhydrolase domain containing 17B
Gene Aliases
C9orf77, CGI-67, FAM108B1
UniGene
Hs.380389
Gene Chromosome Location
Chr.9: 71859461-71911245 on Build GRCh38
Transcripts
NM_001025780.2 NM_016014.3 XM_006717134.3 XM_017014787.1 XM_017014788.1 XM_017014789.1 XM_017014790.1 XM_017014791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6590877 c.732C>T p.L244L Chr.9: 71866922-71866922 on Build GRCh38
3657978 c.829T>C p.L277L Chr.9: 71866825-71866825 on Build GRCh38
6115658 c.732C>G p.L244L Chr.9: 71866922-71866922 on Build GRCh38
4914301 c.851A>C p.E284A Chr.9: 71866803-71866803 on Build GRCh38
4866219 c.759T>C p.P253P Chr.9: 71866895-71866895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977585945 Chr.9: 71866643-71866643 on Build GRCh38
rs376028645 Chr.9: 71866895-71866895 on Build GRCh38
rs879701112 Chr.9: 71866766-71866766 on Build GRCh38
rs372004707 Chr.9: 71866894-71866894 on Build GRCh38
rs947537207 Chr.9: 71866619-71866619 on Build GRCh38
rs775797358 Chr.9: 71866845-71866845 on Build GRCh38
rs181805585 Chr.9: 71866710-71866710 on Build GRCh38
rs938422769 Chr.9: 71866697-71866697 on Build GRCh38
rs369334690 Chr.9: 71866902-71866902 on Build GRCh38
rs956815248 Chr.9: 71866637-71866637 on Build GRCh38
rs912643551 Chr.9: 71866715-71866715 on Build GRCh38
rs776895458 Chr.9: 71866892-71866892 on Build GRCh38
rs975563479 Chr.9: 71866722-71866722 on Build GRCh38
rs924360419 Chr.9: 71866678-71866678 on Build GRCh38
rs554947530 Chr.9: 71866860-71866860 on Build GRCh38
rs774243027 Chr.9: 71866829-71866829 on Build GRCh38
rs1031385786 Chr.9: 71866620-71866620 on Build GRCh38
rs558730000 Chr.9: 71866740-71866740 on Build GRCh38
rs770905338 Chr.9: 71866826-71866826 on Build GRCh38
rs760746048 Chr.9: 71866846-71866846 on Build GRCh38
rs565932773 Chr.9: 71866814-71866814 on Build GRCh38
rs755653161 Chr.9: 71866767-71866767 on Build GRCh38
rs200191910 Chr.9: 71866841-71866841 on Build GRCh38
rs777225213 Chr.9: 71866806-71866806 on Build GRCh38
rs538706930 Chr.9: 71866716-71866716 on Build GRCh38

View Full Product Details