Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7414644-7415218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY2
Entrez Gene ID
108
Gene Name
adenylate cyclase 2
Gene Aliases
AC2, HBAC2
UniGene
Hs.481545
Gene Chromosome Location
Chr.5: 7396230-7830081 on Build GRCh38
Transcripts
NM_020546.2 XM_011513942.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1311312 c.362G>T p.G121V Chr.5: 7414724-7414724 on Build GRCh38
5816054 c.342G>T p.W114C Chr.5: 7414704-7414704 on Build GRCh38
6448234 c.330G>A p.S110S Chr.5: 7414692-7414692 on Build GRCh38
7316203 c.386G>C p.G129A Chr.5: 7414748-7414748 on Build GRCh38
342522 c.291C>A p.C97* Chr.5: 7414653-7414653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569765374 Chr.5: 7414925-7414925 on Build GRCh38
rs1055881424 Chr.5: 7414929-7414929 on Build GRCh38
rs372373069 Chr.5: 7414681-7414681 on Build GRCh38
rs577315373 Chr.5: 7415192-7415192 on Build GRCh38
rs972342739 Chr.5: 7415189-7415189 on Build GRCh38
rs200419013 Chr.5: 7414761-7414761 on Build GRCh38
rs894988610 Chr.5: 7414942-7414942 on Build GRCh38
rs912105244 Chr.5: 7414837-7414837 on Build GRCh38
rs558630252 Chr.5: 7414984-7414984 on Build GRCh38
rs138333689 Chr.5: 7415125-7415125 on Build GRCh38
rs538729991 Chr.5: 7414997-7415018 on Build GRCh38
rs532485075 Chr.5: 7414824-7414824 on Build GRCh38
rs1000234646 Chr.5: 7414913-7414913 on Build GRCh38
rs559319924 Chr.5: 7414845-7414845 on Build GRCh38
rs555253265 Chr.5: 7415082-7415082 on Build GRCh38
rs962443398 Chr.5: 7414683-7414683 on Build GRCh38
rs912052171 Chr.5: 7415175-7415175 on Build GRCh38
rs774404794 Chr.5: 7414696-7414696 on Build GRCh38
rs374414946 Chr.5: 7414796-7414796 on Build GRCh38
rs191910435 Chr.5: 7415060-7415060 on Build GRCh38
rs768647838 Chr.5: 7415179-7415184 on Build GRCh38
rs561505729 Chr.5: 7414707-7414707 on Build GRCh38
rs957807550 Chr.5: 7414748-7414748 on Build GRCh38
rs774466655 Chr.5: 7415044-7415045 on Build GRCh38
rs1041687454 Chr.5: 7414891-7414891 on Build GRCh38
rs765899703 Chr.5: 7414747-7414747 on Build GRCh38
rs572061260 Chr.5: 7415055-7415055 on Build GRCh38
rs567128755 Chr.5: 7414904-7414904 on Build GRCh38
rs763949561 Chr.5: 7414759-7414759 on Build GRCh38
rs149121326 Chr.5: 7414752-7414752 on Build GRCh38
rs549624755 Chr.5: 7414917-7414917 on Build GRCh38
rs547251321 Chr.5: 7414898-7414898 on Build GRCh38
rs145083777 Chr.5: 7414713-7414713 on Build GRCh38
rs951314300 Chr.5: 7415120-7415120 on Build GRCh38
rs552569662 Chr.5: 7414828-7414828 on Build GRCh38
rs1428399 Chr.5: 7414989-7414989 on Build GRCh38
rs899643480 Chr.5: 7414956-7414956 on Build GRCh38
rs761828558 Chr.5: 7414776-7414776 on Build GRCh38
rs749765547 Chr.5: 7414692-7414692 on Build GRCh38
rs879010941 Chr.5: 7414750-7414750 on Build GRCh38
rs1012455426 Chr.5: 7414983-7414983 on Build GRCh38
rs112260510 Chr.5: 7414997-7414997 on Build GRCh38
rs907482340 Chr.5: 7415001-7415001 on Build GRCh38
rs1016604837 Chr.5: 7415129-7415129 on Build GRCh38
rs891918242 Chr.5: 7415005-7415005 on Build GRCh38
rs186214107 Chr.5: 7414887-7414887 on Build GRCh38
rs998127932 Chr.5: 7414987-7414987 on Build GRCh38
rs536421111 Chr.5: 7414906-7414906 on Build GRCh38
rs999430695 Chr.5: 7415043-7415044 on Build GRCh38
rs904651004 Chr.5: 7414897-7414897 on Build GRCh38
rs1031854851 Chr.5: 7415003-7415003 on Build GRCh38
rs920578436 Chr.5: 7414890-7414890 on Build GRCh38
rs1036801866 Chr.5: 7414871-7414871 on Build GRCh38
rs143227108 Chr.5: 7414753-7414753 on Build GRCh38
rs753770510 Chr.5: 7415169-7415169 on Build GRCh38
rs538331146 Chr.5: 7414936-7414936 on Build GRCh38
rs765331515 Chr.5: 7414785-7414785 on Build GRCh38
rs906470616 Chr.5: 7415009-7415009 on Build GRCh38
rs757789496 Chr.5: 7414814-7414814 on Build GRCh38
rs755629280 Chr.5: 7415037-7415037 on Build GRCh38
rs6148896 Chr.5: 7414968-7414989 on Build GRCh38
rs752365071 Chr.5: 7414791-7414791 on Build GRCh38
rs1003095165 Chr.5: 7415002-7415002 on Build GRCh38
rs142824732 Chr.5: 7415157-7415157 on Build GRCh38
rs183236897 Chr.5: 7415144-7415144 on Build GRCh38
rs973605937 Chr.5: 7414745-7414745 on Build GRCh38

View Full Product Details