Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33678129-33678581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F1
Entrez Gene ID
1869
Gene Name
E2F transcription factor 1
Gene Aliases
E2F-1, RBAP1, RBBP3, RBP3
UniGene
Hs.654393
Gene Chromosome Location
Chr.20: 33675486-33686404 on Build GRCh38
Transcripts
NM_005225.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1025971 c.588A>T p.T196T Chr.20: 33678338-33678338 on Build GRCh38
4950937 c.718A>C p.S240R Chr.20: 33678208-33678208 on Build GRCh38
6651006 c.654G>A p.Q218Q Chr.20: 33678272-33678272 on Build GRCh38
5964253 c.663C>T p.D221D Chr.20: 33678263-33678263 on Build GRCh38
6651009 c.631C>T p.R211* Chr.20: 33678295-33678295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754564586 Chr.20: 33678327-33678327 on Build GRCh38
rs774734833 Chr.20: 33678370-33678370 on Build GRCh38
rs35385772 Chr.20: 33678328-33678328 on Build GRCh38
rs753472103 Chr.20: 33678325-33678325 on Build GRCh38
rs775828609 Chr.20: 33678259-33678259 on Build GRCh38
rs764256438 Chr.20: 33678279-33678279 on Build GRCh38
rs751379954 Chr.20: 33678557-33678557 on Build GRCh38
rs373902244 Chr.20: 33678321-33678321 on Build GRCh38
rs750491197 Chr.20: 33678152-33678168 on Build GRCh38
rs1020849508 Chr.20: 33678384-33678384 on Build GRCh38
rs767675186 Chr.20: 33678375-33678375 on Build GRCh38
rs910329477 Chr.20: 33678151-33678151 on Build GRCh38
rs150303208 Chr.20: 33678418-33678418 on Build GRCh38
rs910290764 Chr.20: 33678413-33678413 on Build GRCh38
rs954007561 Chr.20: 33678541-33678541 on Build GRCh38
rs771638059 Chr.20: 33678331-33678331 on Build GRCh38
rs750422437 Chr.20: 33678312-33678312 on Build GRCh38
rs776055929 Chr.20: 33678351-33678351 on Build GRCh38
rs901724647 Chr.20: 33678517-33678517 on Build GRCh38
rs984233810 Chr.20: 33678550-33678550 on Build GRCh38
rs373477652 Chr.20: 33678374-33678374 on Build GRCh38
rs752993937 Chr.20: 33678168-33678168 on Build GRCh38
rs984199783 Chr.20: 33678207-33678207 on Build GRCh38
rs763333981 Chr.20: 33678278-33678278 on Build GRCh38
rs767306081 Chr.20: 33678294-33678294 on Build GRCh38
rs987188011 Chr.20: 33678406-33678406 on Build GRCh38
rs1028365325 Chr.20: 33678540-33678540 on Build GRCh38
rs917483838 Chr.20: 33678320-33678320 on Build GRCh38
rs891646505 Chr.20: 33678428-33678428 on Build GRCh38
rs771273717 Chr.20: 33678240-33678240 on Build GRCh38
rs540997640 Chr.20: 33678377-33678377 on Build GRCh38
rs779549616 Chr.20: 33678221-33678221 on Build GRCh38
rs758535355 Chr.20: 33678177-33678177 on Build GRCh38
rs757322704 Chr.20: 33678187-33678187 on Build GRCh38
rs759952955 Chr.20: 33678158-33678158 on Build GRCh38
rs747620421 Chr.20: 33678329-33678329 on Build GRCh38
rs118134345 Chr.20: 33678457-33678457 on Build GRCh38
rs199745489 Chr.20: 33678254-33678254 on Build GRCh38
rs377637237 Chr.20: 33678322-33678322 on Build GRCh38
rs765340568 Chr.20: 33678159-33678159 on Build GRCh38
rs546049774 Chr.20: 33678318-33678318 on Build GRCh38
rs746424387 Chr.20: 33678277-33678277 on Build GRCh38
rs1017002133 Chr.20: 33678552-33678552 on Build GRCh38
rs745608299 Chr.20: 33678201-33678201 on Build GRCh38
rs781204653 Chr.20: 33678189-33678189 on Build GRCh38
rs770049550 Chr.20: 33678346-33678346 on Build GRCh38
rs935776813 Chr.20: 33678417-33678417 on Build GRCh38
rs776800391 Chr.20: 33678155-33678155 on Build GRCh38
rs753560436 Chr.20: 33678398-33678398 on Build GRCh38
rs759773652 Chr.20: 33678253-33678253 on Build GRCh38
rs759867035 Chr.20: 33678434-33678434 on Build GRCh38
rs929484530 Chr.20: 33678518-33678518 on Build GRCh38
rs751633126 Chr.20: 33678287-33678287 on Build GRCh38
rs760780184 Chr.20: 33678390-33678390 on Build GRCh38
rs34954426 Chr.20: 33678534-33678534 on Build GRCh38
rs768935757 Chr.20: 33678368-33678368 on Build GRCh38
rs760719188 Chr.20: 33678389-33678389 on Build GRCh38
rs200658903 Chr.20: 33678291-33678291 on Build GRCh38
rs746432333 Chr.20: 33678341-33678341 on Build GRCh38
rs201920937 Chr.20: 33678236-33678236 on Build GRCh38
rs766488656 Chr.20: 33678404-33678404 on Build GRCh38
rs200904023 Chr.20: 33678305-33678305 on Build GRCh38
rs747667687 Chr.20: 33678239-33678239 on Build GRCh38
rs755958197 Chr.20: 33678215-33678215 on Build GRCh38
rs531113927 Chr.20: 33678232-33678232 on Build GRCh38
rs754795757 Chr.20: 33678401-33678401 on Build GRCh38
rs764309535 Chr.20: 33678184-33678184 on Build GRCh38
rs528731000 Chr.20: 33678332-33678332 on Build GRCh38
rs1047996645 Chr.20: 33678522-33678522 on Build GRCh38
rs996487460 Chr.20: 33678533-33678533 on Build GRCh38
rs371030754 Chr.20: 33678358-33678358 on Build GRCh38
rs1032145615 Chr.20: 33678355-33678355 on Build GRCh38

View Full Product Details