Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86603512-86604089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105374845
Entrez Gene ID
105374845
Gene Name
uncharacterized LOC105374845
Gene Aliases
-
UniGene
Hs.741633
Gene Chromosome Location
Chr.2: 86562345-86618766 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Gene Symbol
RNF103
Entrez Gene ID
7844
Gene Name
ring finger protein 103
Gene Aliases
HKF-1, KF-1, KF1, ZFP-103, ZFP103
UniGene
Hs.469199
Gene Chromosome Location
Chr.2: 86603393-86623877 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Gene Symbol
RNF103-CHMP3
Entrez Gene ID
100526767
Gene Name
RNF103-CHMP3 readthrough
Gene Aliases
RNF103-VPS24
UniGene
Hs.591582
Gene Chromosome Location
Chr.2: 86503430-86721122 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5397547 c.1886A>G p.E629G Chr.2: 86604015-86604015 on Build GRCh38
3840189 c.2010G>C p.K670N Chr.2: 86603891-86603891 on Build GRCh38
6767145 c.2017T>C p.Y673H Chr.2: 86603884-86603884 on Build GRCh38
4902310 c.1982T>A p.V661D Chr.2: 86603919-86603919 on Build GRCh38
2823157 c.2025A>G p.Q675Q Chr.2: 86603876-86603876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546004918 Chr.2: 86603673-86603673 on Build GRCh38
rs746516136 Chr.2: 86603900-86603900 on Build GRCh38
rs143128520 Chr.2: 86603796-86603796 on Build GRCh38
rs755732129 Chr.2: 86604054-86604054 on Build GRCh38
rs773307711 Chr.2: 86603809-86603809 on Build GRCh38
rs759794644 Chr.2: 86603992-86603992 on Build GRCh38
rs761941063 Chr.2: 86603822-86603822 on Build GRCh38
rs777018125 Chr.2: 86603982-86603982 on Build GRCh38
rs186230810 Chr.2: 86603601-86603601 on Build GRCh38
rs909389650 Chr.2: 86603632-86603632 on Build GRCh38
rs371702501 Chr.2: 86603880-86603880 on Build GRCh38
rs765651613 Chr.2: 86603876-86603876 on Build GRCh38
rs761823531 Chr.2: 86603766-86603766 on Build GRCh38
rs751538770 Chr.2: 86603892-86603892 on Build GRCh38
rs201068550 Chr.2: 86603855-86603855 on Build GRCh38
rs975334382 Chr.2: 86603667-86603667 on Build GRCh38
rs926278370 Chr.2: 86603889-86603889 on Build GRCh38
rs892333630 Chr.2: 86603914-86603914 on Build GRCh38
rs141930795 Chr.2: 86604057-86604057 on Build GRCh38
rs955224092 Chr.2: 86603733-86603733 on Build GRCh38
rs756358507 Chr.2: 86603998-86603998 on Build GRCh38
rs564562382 Chr.2: 86603702-86603702 on Build GRCh38
rs35926179 Chr.2: 86603966-86603966 on Build GRCh38
rs139382955 Chr.2: 86604064-86604064 on Build GRCh38
rs748854200 Chr.2: 86603887-86603889 on Build GRCh38
rs975169807 Chr.2: 86603544-86603544 on Build GRCh38
rs150313341 Chr.2: 86603954-86603954 on Build GRCh38
rs762824123 Chr.2: 86603807-86603807 on Build GRCh38
rs758946477 Chr.2: 86603891-86603891 on Build GRCh38
rs554442784 Chr.2: 86603561-86603561 on Build GRCh38
rs200630890 Chr.2: 86603934-86603934 on Build GRCh38
rs750106820 Chr.2: 86604053-86604053 on Build GRCh38
rs770846619 Chr.2: 86603960-86603960 on Build GRCh38
rs751945777 Chr.2: 86604011-86604011 on Build GRCh38
rs766608264 Chr.2: 86603827-86603827 on Build GRCh38
rs996885456 Chr.2: 86603994-86603994 on Build GRCh38
rs146507546 Chr.2: 86603901-86603901 on Build GRCh38
rs780516985 Chr.2: 86604045-86604045 on Build GRCh38
rs985327363 Chr.2: 86603594-86603594 on Build GRCh38
rs1803428 Chr.2: 86603670-86603670 on Build GRCh38
rs773218298 Chr.2: 86603953-86603953 on Build GRCh38
rs1041822558 Chr.2: 86604050-86604050 on Build GRCh38
rs768612101 Chr.2: 86603937-86603937 on Build GRCh38
rs1032981709 Chr.2: 86603623-86603623 on Build GRCh38
rs202185043 Chr.2: 86603999-86603999 on Build GRCh38
rs972349045 Chr.2: 86603706-86603706 on Build GRCh38
rs561735009 Chr.2: 86603904-86603904 on Build GRCh38
rs3755000 Chr.2: 86603753-86603753 on Build GRCh38
rs543456450 Chr.2: 86603873-86603873 on Build GRCh38
rs921040166 Chr.2: 86603549-86603549 on Build GRCh38
rs767524634 Chr.2: 86603808-86603808 on Build GRCh38
rs780903573 Chr.2: 86603902-86603902 on Build GRCh38
rs375516528 Chr.2: 86604013-86604013 on Build GRCh38
rs61760874 Chr.2: 86603928-86603928 on Build GRCh38
rs764659594 Chr.2: 86604008-86604008 on Build GRCh38
rs754744364 Chr.2: 86603891-86603892 on Build GRCh38
rs144162110 Chr.2: 86603899-86603899 on Build GRCh38
rs952505547 Chr.2: 86603557-86603557 on Build GRCh38
rs964320447 Chr.2: 86603637-86603637 on Build GRCh38
rs764350924 Chr.2: 86603681-86603681 on Build GRCh38
rs967385282 Chr.2: 86603548-86603548 on Build GRCh38
rs750896154 Chr.2: 86603938-86603938 on Build GRCh38
rs367610455 Chr.2: 86603823-86603823 on Build GRCh38
rs754219648 Chr.2: 86603861-86603861 on Build GRCh38
rs767152368 Chr.2: 86603769-86603769 on Build GRCh38
rs936772000 Chr.2: 86603874-86603876 on Build GRCh38
rs1000148956 Chr.2: 86603554-86603554 on Build GRCh38
rs775225683 Chr.2: 86603930-86603930 on Build GRCh38

View Full Product Details