Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90230145-90230662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIB1
Entrez Gene ID
10519
Gene Name
calcium and integrin binding 1
Gene Aliases
CIB, CIBP, KIP1, PRKDCIP, SIP2-28
UniGene
Hs.715556
Gene Chromosome Location
Chr.15: 90230245-90265759 on Build GRCh38
Transcripts
NM_001277764.1 NM_006384.3 NR_102427.1 NR_102428.1 XM_006720375.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4922514 c.566T>G p.I189S Chr.15: 90230494-90230494 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs563790691 Chr.15: 90230292-90230292 on Build GRCh38
rs987461328 Chr.15: 90230269-90230269 on Build GRCh38
rs926135951 Chr.15: 90230332-90230332 on Build GRCh38
rs897170963 Chr.15: 90230608-90230608 on Build GRCh38
rs571745594 Chr.15: 90230343-90230343 on Build GRCh38
rs1019622373 Chr.15: 90230281-90230281 on Build GRCh38
rs376599509 Chr.15: 90230388-90230388 on Build GRCh38
rs373892321 Chr.15: 90230458-90230458 on Build GRCh38
rs947785954 Chr.15: 90230572-90230572 on Build GRCh38
rs780392746 Chr.15: 90230564-90230564 on Build GRCh38
rs906483056 Chr.15: 90230413-90230413 on Build GRCh38
rs911373072 Chr.15: 90230493-90230493 on Build GRCh38
rs570959156 Chr.15: 90230603-90230603 on Build GRCh38
rs937264046 Chr.15: 90230389-90230389 on Build GRCh38
rs756336543 Chr.15: 90230465-90230466 on Build GRCh38
rs1001775470 Chr.15: 90230212-90230212 on Build GRCh38
rs376137827 Chr.15: 90230449-90230449 on Build GRCh38
rs975356539 Chr.15: 90230305-90230305 on Build GRCh38
rs920114216 Chr.15: 90230220-90230220 on Build GRCh38
rs374281114 Chr.15: 90230534-90230534 on Build GRCh38
rs369492809 Chr.15: 90230196-90230200 on Build GRCh38
rs761660913 Chr.15: 90230373-90230373 on Build GRCh38
rs1003482517 Chr.15: 90230602-90230602 on Build GRCh38
rs771152399 Chr.15: 90230494-90230494 on Build GRCh38
rs373499226 Chr.15: 90230438-90230438 on Build GRCh38
rs537611708 Chr.15: 90230624-90230624 on Build GRCh38
rs574694331 Chr.15: 90230264-90230264 on Build GRCh38
rs573429974 Chr.15: 90230370-90230370 on Build GRCh38
rs1023886371 Chr.15: 90230405-90230405 on Build GRCh38
rs774865095 Chr.15: 90230306-90230306 on Build GRCh38
rs1804804 Chr.15: 90230465-90230465 on Build GRCh38
rs556135608 Chr.15: 90230230-90230230 on Build GRCh38
rs994090496 Chr.15: 90230614-90230614 on Build GRCh38
rs1044936442 Chr.15: 90230573-90230573 on Build GRCh38
rs112061852 Chr.15: 90230416-90230416 on Build GRCh38
rs971166289 Chr.15: 90230597-90230597 on Build GRCh38
rs187381937 Chr.15: 90230333-90230333 on Build GRCh38
rs911440417 Chr.15: 90230293-90230293 on Build GRCh38
rs866212694 Chr.15: 90230546-90230546 on Build GRCh38
rs1038638094 Chr.15: 90230395-90230395 on Build GRCh38
rs557900089 Chr.15: 90230339-90230339 on Build GRCh38
rs994867236 Chr.15: 90230255-90230255 on Build GRCh38
rs923613809 Chr.15: 90230523-90230523 on Build GRCh38
rs552698988 Chr.15: 90230574-90230574 on Build GRCh38
rs760298374 Chr.15: 90230338-90230338 on Build GRCh38
rs370628711 Chr.15: 90230479-90230479 on Build GRCh38
rs1012040350 Chr.15: 90230396-90230396 on Build GRCh38
rs150632796 Chr.15: 90230470-90230470 on Build GRCh38
rs899288341 Chr.15: 90230245-90230245 on Build GRCh38
rs1026295296 Chr.15: 90230263-90230263 on Build GRCh38
rs544801127 Chr.15: 90230426-90230426 on Build GRCh38
rs993936791 Chr.15: 90230436-90230436 on Build GRCh38
rs1016763055 Chr.15: 90230233-90230233 on Build GRCh38
rs1026993352 Chr.15: 90230471-90230471 on Build GRCh38
rs952996095 Chr.15: 90230473-90230473 on Build GRCh38
rs750620059 Chr.15: 90230522-90230522 on Build GRCh38
rs190669499 Chr.15: 90230421-90230421 on Build GRCh38
rs2073706 Chr.15: 90230550-90230550 on Build GRCh38
rs74037064 Chr.15: 90230569-90230569 on Build GRCh38
rs944325168 Chr.15: 90230499-90230499 on Build GRCh38
rs563004130 Chr.15: 90230455-90230455 on Build GRCh38
rs534082575 Chr.15: 90230418-90230418 on Build GRCh38
rs531312201 Chr.15: 90230376-90230376 on Build GRCh38
rs906342688 Chr.15: 90230588-90230588 on Build GRCh38

View Full Product Details