Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 46919192-46919689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA4
Entrez Gene ID
2557
Gene Name
gamma-aminobutyric acid type A receptor alpha4 subunit
Gene Aliases
-
UniGene
Hs.248112
Gene Chromosome Location
Chr.4: 46918900-46994407 on Build GRCh38
Transcripts
NM_000809.3 NM_001204266.1 NM_001204267.1

SNP Detail

SNP ID Chromosome Location
rs921859299 Chr.4: 46919364-46919364 on Build GRCh38
rs920426272 Chr.4: 46919572-46919572 on Build GRCh38
rs1039075852 Chr.4: 46919607-46919607 on Build GRCh38
rs192656811 Chr.4: 46919507-46919507 on Build GRCh38
rs779387102 Chr.4: 46919383-46919383 on Build GRCh38
rs935987149 Chr.4: 46919323-46919323 on Build GRCh38
rs759772897 Chr.4: 46919585-46919585 on Build GRCh38
rs75561299 Chr.4: 46919268-46919268 on Build GRCh38
rs977204333 Chr.4: 46919248-46919249 on Build GRCh38
rs552636563 Chr.4: 46919615-46919615 on Build GRCh38
rs967518759 Chr.4: 46919279-46919279 on Build GRCh38
rs892002561 Chr.4: 46919503-46919503 on Build GRCh38
rs977461815 Chr.4: 46919638-46919641 on Build GRCh38
rs866703117 Chr.4: 46919318-46919318 on Build GRCh38
rs993031722 Chr.4: 46919252-46919252 on Build GRCh38
rs141339871 Chr.4: 46919594-46919594 on Build GRCh38
rs1001918137 Chr.4: 46919515-46919515 on Build GRCh38
rs950557378 Chr.4: 46919573-46919573 on Build GRCh38
rs1055024757 Chr.4: 46919448-46919448 on Build GRCh38
rs868573188 Chr.4: 46919251-46919251 on Build GRCh38
rs1004676230 Chr.4: 46919467-46919467 on Build GRCh38
rs544577575 Chr.4: 46919635-46919635 on Build GRCh38
rs1010332204 Chr.4: 46919654-46919654 on Build GRCh38
rs1053666658 Chr.4: 46919351-46919351 on Build GRCh38
rs977409617 Chr.4: 46919287-46919287 on Build GRCh38
rs890195390 Chr.4: 46919425-46919425 on Build GRCh38
rs1054658540 Chr.4: 46919645-46919645 on Build GRCh38
rs553759367 Chr.4: 46919602-46919602 on Build GRCh38
rs973381100 Chr.4: 46919564-46919564 on Build GRCh38
rs925980329 Chr.4: 46919321-46919321 on Build GRCh38
rs1010258752 Chr.4: 46919504-46919504 on Build GRCh38
rs900711408 Chr.4: 46919282-46919282 on Build GRCh38
rs1051627049 Chr.4: 46919382-46919382 on Build GRCh38
rs1000459203 Chr.4: 46919395-46919395 on Build GRCh38
rs1024509086 Chr.4: 46919600-46919600 on Build GRCh38
rs773150856 Chr.4: 46919237-46919237 on Build GRCh38
rs959518668 Chr.4: 46919529-46919529 on Build GRCh38
rs570299480 Chr.4: 46919362-46919362 on Build GRCh38
rs944472024 Chr.4: 46919595-46919595 on Build GRCh38
rs371030573 Chr.4: 46919616-46919616 on Build GRCh38
rs931783702 Chr.4: 46919379-46919379 on Build GRCh38
rs911544566 Chr.4: 46919226-46919226 on Build GRCh38
rs921651896 Chr.4: 46919614-46919614 on Build GRCh38
rs1034359010 Chr.4: 46919525-46919525 on Build GRCh38
rs577348641 Chr.4: 46919634-46919634 on Build GRCh38
rs112564086 Chr.4: 46919652-46919652 on Build GRCh38
rs546472754 Chr.4: 46919510-46919510 on Build GRCh38
rs537355926 Chr.4: 46919406-46919406 on Build GRCh38

View Full Product Details