Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41620822-41621390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DESI1
Entrez Gene ID
27351
Gene Name
desumoylating isopeptidase 1
Gene Aliases
D15Wsu75e, DESI2, DJ347H13.4, DeSI-1, FAM152B, PPPDE2
UniGene
Hs.570455
Gene Chromosome Location
Chr.22: 41598028-41621077 on Build GRCh38
Transcripts
NM_015704.2 XM_005261571.4 NM_001288976.1 NM_001288977.1 NM_001288978.1 NM_001469.4

Gene Symbol
XRCC6
Entrez Gene ID
2547
Gene Name
X-ray repair cross complementing 6
Gene Aliases
CTC75, CTCBF, G22P1, KU70, ML8, TLAA
UniGene
Hs.292493
Gene Chromosome Location
Chr.22: 41621163-41664048 on Build GRCh38
Transcripts
NM_015704.2 XM_005261571.4 NM_001288976.1 NM_001288977.1 NM_001288978.1 NM_001469.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1034654 c.10C>T p.P4S Chr.22: 41620830-41620830 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1037102561 Chr.22: 41620941-41620941 on Build GRCh38
rs28384703 Chr.22: 41620858-41620858 on Build GRCh38
rs772231151 Chr.22: 41621249-41621249 on Build GRCh38
rs28384704 Chr.22: 41620940-41620940 on Build GRCh38
rs569779120 Chr.22: 41621072-41621072 on Build GRCh38
rs1010649820 Chr.22: 41621100-41621100 on Build GRCh38
rs1022118107 Chr.22: 41620969-41620969 on Build GRCh38
rs547417018 Chr.22: 41621043-41621043 on Build GRCh38
rs951437530 Chr.22: 41621366-41621366 on Build GRCh38
rs1028086774 Chr.22: 41620957-41620957 on Build GRCh38
rs998311182 Chr.22: 41620948-41620948 on Build GRCh38
rs984153802 Chr.22: 41621371-41621371 on Build GRCh38
rs1003552510 Chr.22: 41621348-41621348 on Build GRCh38
rs759539018 Chr.22: 41621356-41621356 on Build GRCh38
rs376876886 Chr.22: 41621250-41621250 on Build GRCh38
rs982107234 Chr.22: 41621127-41621127 on Build GRCh38
rs36218585 Chr.22: 41621107-41621107 on Build GRCh38
rs752117830 Chr.22: 41620846-41620846 on Build GRCh38
rs132770 Chr.22: 41621260-41621260 on Build GRCh38
rs1048619543 Chr.22: 41620922-41620922 on Build GRCh38
rs889554766 Chr.22: 41620958-41620958 on Build GRCh38
rs916051223 Chr.22: 41620893-41620893 on Build GRCh38
rs992831956 Chr.22: 41621151-41621151 on Build GRCh38
rs745365643 Chr.22: 41620870-41620870 on Build GRCh38
rs982076093 Chr.22: 41621258-41621258 on Build GRCh38
rs910090610 Chr.22: 41621052-41621052 on Build GRCh38
rs924672219 Chr.22: 41620868-41620868 on Build GRCh38
rs1018032733 Chr.22: 41621075-41621076 on Build GRCh38
rs367967155 Chr.22: 41620998-41620998 on Build GRCh38
rs28384707 Chr.22: 41621276-41621276 on Build GRCh38
rs901884578 Chr.22: 41621126-41621145 on Build GRCh38
rs531853481 Chr.22: 41621302-41621302 on Build GRCh38
rs936774222 Chr.22: 41621029-41621029 on Build GRCh38
rs1036431513 Chr.22: 41620942-41620942 on Build GRCh38
rs373861057 Chr.22: 41621122-41621123 on Build GRCh38
rs900807698 Chr.22: 41621064-41621064 on Build GRCh38
rs781712836 Chr.22: 41620856-41620856 on Build GRCh38
rs755228400 Chr.22: 41620847-41620847 on Build GRCh38
rs961788070 Chr.22: 41621279-41621279 on Build GRCh38
rs1055564673 Chr.22: 41620934-41620934 on Build GRCh38
rs946432727 Chr.22: 41621216-41621216 on Build GRCh38
rs960080048 Chr.22: 41621006-41621006 on Build GRCh38
rs923701791 Chr.22: 41621337-41621337 on Build GRCh38
rs961580887 Chr.22: 41621183-41621183 on Build GRCh38
rs11342289 Chr.22: 41621122-41621122 on Build GRCh38
rs28384706 Chr.22: 41621243-41621243 on Build GRCh38
rs991276360 Chr.22: 41621121-41621121 on Build GRCh38
rs978035522 Chr.22: 41620988-41620988 on Build GRCh38
rs866199333 Chr.22: 41620909-41620909 on Build GRCh38
rs778748117 Chr.22: 41620866-41620866 on Build GRCh38
rs946401689 Chr.22: 41621326-41621326 on Build GRCh38
rs132770 Chr.22: 41621260-41621260 on Build GRCh38
rs77728367 Chr.22: 41621207-41621207 on Build GRCh38
rs933652493 Chr.22: 41621065-41621065 on Build GRCh38
rs146105341 Chr.22: 41621167-41621167 on Build GRCh38
rs966626627 Chr.22: 41620865-41620865 on Build GRCh38
rs571540057 Chr.22: 41621226-41621226 on Build GRCh38
rs374582556 Chr.22: 41620919-41620919 on Build GRCh38
rs768402475 Chr.22: 41620889-41620889 on Build GRCh38
rs1018399007 Chr.22: 41620962-41620962 on Build GRCh38
rs930002840 Chr.22: 41621259-41621259 on Build GRCh38
rs973373245 Chr.22: 41621185-41621185 on Build GRCh38
rs746972655 Chr.22: 41620884-41620884 on Build GRCh38
rs915980985 Chr.22: 41621021-41621021 on Build GRCh38
rs967178456 Chr.22: 41620991-41620991 on Build GRCh38
rs922800546 Chr.22: 41621303-41621303 on Build GRCh38
rs1049063629 Chr.22: 41621055-41621055 on Build GRCh38
rs749628206 Chr.22: 41621008-41621008 on Build GRCh38
rs1026707073 Chr.22: 41620954-41620954 on Build GRCh38
rs999349162 Chr.22: 41621230-41621230 on Build GRCh38
rs558003779 Chr.22: 41621071-41621071 on Build GRCh38
rs775234959 Chr.22: 41620881-41620881 on Build GRCh38
rs935111238 Chr.22: 41621341-41621341 on Build GRCh38
rs543994317 Chr.22: 41621328-41621328 on Build GRCh38
rs749746400 Chr.22: 41621114-41621114 on Build GRCh38
rs1034960988 Chr.22: 41621117-41621117 on Build GRCh38
rs528941317 Chr.22: 41621038-41621038 on Build GRCh38
rs967315581 Chr.22: 41621106-41621106 on Build GRCh38
rs972917607 Chr.22: 41621291-41621291 on Build GRCh38
rs568139169 Chr.22: 41620963-41620963 on Build GRCh38
rs897294723 Chr.22: 41620949-41620949 on Build GRCh38
rs752490256 Chr.22: 41621290-41621290 on Build GRCh38
rs565636585 Chr.22: 41621054-41621054 on Build GRCh38
rs965005519 Chr.22: 41621085-41621085 on Build GRCh38
rs1039922874 Chr.22: 41621298-41621298 on Build GRCh38
rs896397612 Chr.22: 41621246-41621246 on Build GRCh38
rs118087986 Chr.22: 41621257-41621257 on Build GRCh38
rs910857586 Chr.22: 41621325-41621325 on Build GRCh38
rs895379291 Chr.22: 41620935-41620935 on Build GRCh38
rs910017059 Chr.22: 41621182-41621182 on Build GRCh38
rs984207856 Chr.22: 41621173-41621173 on Build GRCh38
rs12485198 Chr.22: 41621056-41621056 on Build GRCh38
rs966593881 Chr.22: 41620970-41620970 on Build GRCh38
rs17002530 Chr.22: 41621146-41621146 on Build GRCh38
rs754503007 Chr.22: 41620848-41620849 on Build GRCh38
rs746927221 Chr.22: 41621275-41621275 on Build GRCh38
rs36217922 Chr.22: 41621258-41621258 on Build GRCh38
rs375770428 Chr.22: 41621080-41621080 on Build GRCh38
rs386821478 Chr.22: 41621258-41621260 on Build GRCh38
rs28384705 Chr.22: 41620952-41620956 on Build GRCh38
rs958139885 Chr.22: 41621110-41621110 on Build GRCh38
rs1046609486 Chr.22: 41621045-41621045 on Build GRCh38
rs1049382609 Chr.22: 41621203-41621203 on Build GRCh38
rs1039838549 Chr.22: 41621187-41621187 on Build GRCh38
rs902817181 Chr.22: 41621316-41621316 on Build GRCh38
rs999818562 Chr.22: 41621116-41621116 on Build GRCh38
rs1042456223 Chr.22: 41620903-41620903 on Build GRCh38
rs934270137 Chr.22: 41621197-41621197 on Build GRCh38
rs888690601 Chr.22: 41620936-41620936 on Build GRCh38
rs776617584 Chr.22: 41621304-41621304 on Build GRCh38
rs11557350 Chr.22: 41621320-41621320 on Build GRCh38
rs112105277 Chr.22: 41621333-41621333 on Build GRCh38
rs889523661 Chr.22: 41621075-41621075 on Build GRCh38
rs566521806 Chr.22: 41620943-41620943 on Build GRCh38
rs573923013 Chr.22: 41621240-41621240 on Build GRCh38
rs377088635 Chr.22: 41621315-41621315 on Build GRCh38
rs771670914 Chr.22: 41620880-41620880 on Build GRCh38
rs922865564 Chr.22: 41621186-41621186 on Build GRCh38
rs913157050 Chr.22: 41620984-41620984 on Build GRCh38
rs1023007715 Chr.22: 41621158-41621158 on Build GRCh38
rs997868430 Chr.22: 41621091-41621091 on Build GRCh38
rs17002533 Chr.22: 41621350-41621350 on Build GRCh38
rs1043467338 Chr.22: 41621224-41621224 on Build GRCh38
rs36218275 Chr.22: 41621313-41621313 on Build GRCh38

View Full Product Details