Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67344005-67344577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCH
Entrez Gene ID
64799
Gene Name
IQ motif containing H
Gene Aliases
NYDSP5
UniGene
Hs.657894
Gene Chromosome Location
Chr.15: 67254772-67501804 on Build GRCh38
Transcripts
NM_001031715.2 NM_001284347.1 NM_001284348.1 NM_001284349.1 NM_001322470.1 NM_001322471.1 NM_001322472.1 NM_001322473.1 NM_001322474.1 NM_001322475.1 XM_011521907.2 XM_011521908.2 XM_011521911.2 XM_011521912.2 XM_011521913.2 XM_011521914.2 XM_011521915.2 XM_011521916.2 XM_011521917.2 XM_011521918.2 XM_011521922.2 XM_017022491.1 XM_017022492.1 XM_017022493.1 XM_017022494.1 XM_017022495.1 XM_017022496.1 XM_017022497.1 XM_017022498.1 XM_017022499.1 XM_017022500.1 XM_017022501.1 XM_017022502.1 XM_017022503.1 XM_017022504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3420550 c.544C>T p.P182S Chr.15: 67344098-67344098 on Build GRCh38
2257469 c.603T>C p.H201H Chr.15: 67344157-67344157 on Build GRCh38
7313448 c.600G>T p.L200L Chr.15: 67344154-67344154 on Build GRCh38
2257470 c.626T>A p.I209N Chr.15: 67344180-67344180 on Build GRCh38
4500307 c.561C>T p.T187T Chr.15: 67344115-67344115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767445068 Chr.15: 67344173-67344173 on Build GRCh38
rs751663248 Chr.15: 67344151-67344151 on Build GRCh38
rs373313138 Chr.15: 67344064-67344064 on Build GRCh38
rs979979769 Chr.15: 67344142-67344142 on Build GRCh38
rs935933746 Chr.15: 67344188-67344188 on Build GRCh38
rs1040331697 Chr.15: 67344039-67344039 on Build GRCh38
rs773686106 Chr.15: 67344132-67344132 on Build GRCh38
rs528008481 Chr.15: 67344547-67344547 on Build GRCh38
rs749070080 Chr.15: 67344185-67344185 on Build GRCh38
rs770407983 Chr.15: 67344219-67344219 on Build GRCh38
rs1001332483 Chr.15: 67344364-67344364 on Build GRCh38
rs773919408 Chr.15: 67344228-67344228 on Build GRCh38
rs373846818 Chr.15: 67344416-67344416 on Build GRCh38
rs952255560 Chr.15: 67344378-67344378 on Build GRCh38
rs886471499 Chr.15: 67344400-67344400 on Build GRCh38
rs542928773 Chr.15: 67344126-67344126 on Build GRCh38
rs773934235 Chr.15: 67344110-67344110 on Build GRCh38
rs145087799 Chr.15: 67344367-67344367 on Build GRCh38
rs1024006795 Chr.15: 67344300-67344300 on Build GRCh38
rs528508316 Chr.15: 67344207-67344207 on Build GRCh38
rs757068537 Chr.15: 67344187-67344187 on Build GRCh38
rs775083091 Chr.15: 67344121-67344121 on Build GRCh38
rs370229786 Chr.15: 67344262-67344262 on Build GRCh38
rs899374731 Chr.15: 67344335-67344335 on Build GRCh38
rs950801413 Chr.15: 67344082-67344082 on Build GRCh38
rs756574138 Chr.15: 67344481-67344481 on Build GRCh38
rs373139972 Chr.15: 67344109-67344109 on Build GRCh38
rs200120948 Chr.15: 67344104-67344104 on Build GRCh38
rs565598723 Chr.15: 67344388-67344388 on Build GRCh38
rs749121911 Chr.15: 67344108-67344108 on Build GRCh38
rs912933208 Chr.15: 67344266-67344266 on Build GRCh38
rs766616525 Chr.15: 67344143-67344143 on Build GRCh38
rs755239683 Chr.15: 67344158-67344158 on Build GRCh38
rs868546980 Chr.15: 67344076-67344076 on Build GRCh38
rs931980370 Chr.15: 67344443-67344443 on Build GRCh38
rs148768360 Chr.15: 67344550-67344550 on Build GRCh38
rs375869262 Chr.15: 67344084-67344084 on Build GRCh38
rs908155010 Chr.15: 67344389-67344389 on Build GRCh38
rs147261195 Chr.15: 67344140-67344140 on Build GRCh38
rs549983889 Chr.15: 67344308-67344308 on Build GRCh38
rs781334433 Chr.15: 67344044-67344044 on Build GRCh38
rs775104627 Chr.15: 67344215-67344215 on Build GRCh38
rs550625321 Chr.15: 67344437-67344437 on Build GRCh38
rs763465278 Chr.15: 67344134-67344134 on Build GRCh38
rs141116177 Chr.15: 67344183-67344183 on Build GRCh38
rs184694281 Chr.15: 67344218-67344218 on Build GRCh38
rs11636980 Chr.15: 67344417-67344417 on Build GRCh38
rs749826960 Chr.15: 67344553-67344553 on Build GRCh38
rs11637008 Chr.15: 67344476-67344476 on Build GRCh38
rs144731544 Chr.15: 67344202-67344203 on Build GRCh38
rs935861697 Chr.15: 67344476-67344476 on Build GRCh38
rs759084381 Chr.15: 67344117-67344117 on Build GRCh38
rs560057226 Chr.15: 67344204-67344204 on Build GRCh38
rs945980762 Chr.15: 67344535-67344535 on Build GRCh38
rs765900957 Chr.15: 67344178-67344178 on Build GRCh38
rs986234056 Chr.15: 67344466-67344466 on Build GRCh38
rs932094943 Chr.15: 67344170-67344170 on Build GRCh38
rs113223282 Chr.15: 67344071-67344071 on Build GRCh38
rs778355852 Chr.15: 67344491-67344491 on Build GRCh38
rs1050854399 Chr.15: 67344276-67344276 on Build GRCh38
rs549023171 Chr.15: 67344371-67344371 on Build GRCh38
rs971045277 Chr.15: 67344339-67344339 on Build GRCh38
rs756014702 Chr.15: 67344176-67344176 on Build GRCh38
rs933713412 Chr.15: 67344274-67344274 on Build GRCh38
rs778776224 Chr.15: 67344202-67344202 on Build GRCh38
rs569731619 Chr.15: 67344323-67344323 on Build GRCh38
rs765880368 Chr.15: 67344131-67344131 on Build GRCh38
rs1039209811 Chr.15: 67344313-67344313 on Build GRCh38
rs943502649 Chr.15: 67344292-67344292 on Build GRCh38
rs577358386 Chr.15: 67344442-67344442 on Build GRCh38
rs771419111 Chr.15: 67344119-67344119 on Build GRCh38
rs766885771 Chr.15: 67344468-67344469 on Build GRCh38
rs867656894 Chr.15: 67344063-67344063 on Build GRCh38
rs769744411 Chr.15: 67344083-67344083 on Build GRCh38
rs371149945 Chr.15: 67344174-67344174 on Build GRCh38
rs763518713 Chr.15: 67344242-67344242 on Build GRCh38
rs189576498 Chr.15: 67344280-67344280 on Build GRCh38

View Full Product Details