Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 26212682-26213211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX3
Entrez Gene ID
11335
Gene Name
chromobox 3
Gene Aliases
HECH, HP1-GAMMA, HP1Hs-gamma
UniGene
Hs.381189
Gene Chromosome Location
Chr.7: 26201211-26213607 on Build GRCh38
Transcripts
NM_007276.4 NM_016587.3 XM_005249611.3

SNP Detail

SNP ID Chromosome Location
rs574579244 Chr.7: 26212934-26212934 on Build GRCh38
rs565672843 Chr.7: 26212814-26212814 on Build GRCh38
rs948205494 Chr.7: 26212885-26212885 on Build GRCh38
rs913229770 Chr.7: 26213146-26213146 on Build GRCh38
rs558543587 Chr.7: 26212727-26212727 on Build GRCh38
rs997980631 Chr.7: 26212886-26212886 on Build GRCh38
rs927467860 Chr.7: 26213079-26213079 on Build GRCh38
rs7186 Chr.7: 26213182-26213182 on Build GRCh38
rs11554894 Chr.7: 26213049-26213049 on Build GRCh38
rs1005455099 Chr.7: 26212920-26212920 on Build GRCh38
rs1049601389 Chr.7: 26212890-26212890 on Build GRCh38
rs957594392 Chr.7: 26213113-26213113 on Build GRCh38
rs543900752 Chr.7: 26212784-26212785 on Build GRCh38
rs1040423264 Chr.7: 26213168-26213168 on Build GRCh38
rs765035386 Chr.7: 26212996-26212996 on Build GRCh38
rs900962846 Chr.7: 26212874-26212874 on Build GRCh38
rs993893400 Chr.7: 26213006-26213007 on Build GRCh38
rs1024483282 Chr.7: 26213021-26213021 on Build GRCh38
rs1016594589 Chr.7: 26212933-26212933 on Build GRCh38
rs753810913 Chr.7: 26212881-26212881 on Build GRCh38
rs961376049 Chr.7: 26212938-26212939 on Build GRCh38
rs754532052 Chr.7: 26213141-26213141 on Build GRCh38
rs534280361 Chr.7: 26212877-26212877 on Build GRCh38
rs3735554 Chr.7: 26212837-26212837 on Build GRCh38
rs747720619 Chr.7: 26213177-26213177 on Build GRCh38
rs576678371 Chr.7: 26213024-26213024 on Build GRCh38
rs1040898134 Chr.7: 26212834-26212834 on Build GRCh38
rs946143970 Chr.7: 26212798-26212798 on Build GRCh38
rs767915869 Chr.7: 26212732-26212732 on Build GRCh38
rs990434915 Chr.7: 26213139-26213139 on Build GRCh38
rs543273759 Chr.7: 26212991-26212991 on Build GRCh38
rs780776847 Chr.7: 26213167-26213167 on Build GRCh38
rs752431269 Chr.7: 26213150-26213150 on Build GRCh38
rs565392435 Chr.7: 26213183-26213183 on Build GRCh38
rs189258793 Chr.7: 26212896-26212896 on Build GRCh38
rs559982933 Chr.7: 26213152-26213152 on Build GRCh38
rs549197600 Chr.7: 26213074-26213074 on Build GRCh38
rs980285796 Chr.7: 26213069-26213069 on Build GRCh38
rs913250934 Chr.7: 26212714-26212714 on Build GRCh38
rs374505442 Chr.7: 26212992-26212992 on Build GRCh38
rs889683873 Chr.7: 26212918-26212918 on Build GRCh38
rs375726042 Chr.7: 26213169-26213169 on Build GRCh38
rs971562271 Chr.7: 26213032-26213032 on Build GRCh38
rs10156063 Chr.7: 26212762-26212762 on Build GRCh38

View Full Product Details